Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152220288-152220561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HRNR
Entrez Gene ID
388697
Gene Name
hornerin
Gene Aliases
FLG3, S100A16, S100a18
UniGene
Hs.490162
Gene Chromosome Location
Chr.1: 152212076-152224196 on Build GRCh38
Transcripts
NM_001009931.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3399716 c.1240G>A p.G414S Chr.1: 152220389-152220389 on Build GRCh38
1472640 c.1186C>T p.Q396* Chr.1: 152220443-152220443 on Build GRCh38
109056 c.1113C>T p.S371S Chr.1: 152220516-152220516 on Build GRCh38
4463945 c.1311C>T p.P437P Chr.1: 152220318-152220318 on Build GRCh38
4896863 c.1133G>A p.G378E Chr.1: 152220496-152220496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772694111 Chr.1: 152220359-152220359 on Build GRCh38
rs1010127920 Chr.1: 152220399-152220399 on Build GRCh38
rs774116398 Chr.1: 152220378-152220378 on Build GRCh38
rs72477389 Chr.1: 152220337-152220337 on Build GRCh38
rs750139724 Chr.1: 152220513-152220513 on Build GRCh38
rs775708273 Chr.1: 152220334-152220334 on Build GRCh38
rs753485928 Chr.1: 152220523-152220523 on Build GRCh38
rs749067740 Chr.1: 152220405-152220405 on Build GRCh38
rs750838719 Chr.1: 152220462-152220462 on Build GRCh38
rs145543456 Chr.1: 152220329-152220329 on Build GRCh38
rs749493328 Chr.1: 152220500-152220500 on Build GRCh38
rs776569245 Chr.1: 152220420-152220420 on Build GRCh38
rs768820469 Chr.1: 152220411-152220411 on Build GRCh38
rs376465935 Chr.1: 152220535-152220535 on Build GRCh38
rs771898607 Chr.1: 152220501-152220501 on Build GRCh38
rs147325259 Chr.1: 152220348-152220348 on Build GRCh38
rs768782028 Chr.1: 152220369-152220369 on Build GRCh38
rs11204936 Chr.1: 152220351-152220351 on Build GRCh38
rs766065534 Chr.1: 152220339-152220339 on Build GRCh38
rs746444536 Chr.1: 152220355-152220355 on Build GRCh38
rs370983663 Chr.1: 152220311-152220311 on Build GRCh38
rs775728463 Chr.1: 152220362-152220362 on Build GRCh38
rs144722087 Chr.1: 152220446-152220446 on Build GRCh38
rs139453762 Chr.1: 152220489-152220489 on Build GRCh38
rs749602341 Chr.1: 152220316-152220316 on Build GRCh38
rs531035610 Chr.1: 152220407-152220407 on Build GRCh38
rs752141082 Chr.1: 152220346-152220346 on Build GRCh38
rs747043377 Chr.1: 152220365-152220365 on Build GRCh38
rs559764481 Chr.1: 152220376-152220376 on Build GRCh38
rs756856015 Chr.1: 152220393-152220393 on Build GRCh38
rs746178115 Chr.1: 152220394-152220394 on Build GRCh38
rs199929636 Chr.1: 152220375-152220375 on Build GRCh38
rs780734501 Chr.1: 152220345-152220345 on Build GRCh38
rs141848627 Chr.1: 152220516-152220516 on Build GRCh38
rs147288664 Chr.1: 152220451-152220451 on Build GRCh38
rs139351032 Chr.1: 152220347-152220347 on Build GRCh38
rs140273713 Chr.1: 152220391-152220391 on Build GRCh38
rs772319115 Chr.1: 152220333-152220333 on Build GRCh38
rs373911875 Chr.1: 152220472-152220472 on Build GRCh38
rs141069027 Chr.1: 152220416-152220416 on Build GRCh38
rs762690312 Chr.1: 152220519-152220519 on Build GRCh38
rs545053892 Chr.1: 152220445-152220445 on Build GRCh38
rs199656596 Chr.1: 152220335-152220335 on Build GRCh38
rs144898490 Chr.1: 152220479-152220479 on Build GRCh38
rs6666097 Chr.1: 152220349-152220349 on Build GRCh38
rs775706708 Chr.1: 152220312-152220312 on Build GRCh38
rs186903853 Chr.1: 152220373-152220373 on Build GRCh38
rs764322018 Chr.1: 152220336-152220336 on Build GRCh38
rs140309785 Chr.1: 152220531-152220531 on Build GRCh38
rs550473784 Chr.1: 152220422-152220422 on Build GRCh38
rs765482568 Chr.1: 152220537-152220537 on Build GRCh38
rs555346363 Chr.1: 152220505-152220505 on Build GRCh38
rs141454792 Chr.1: 152220343-152220343 on Build GRCh38
rs960352925 Chr.1: 152220424-152220424 on Build GRCh38
rs753274510 Chr.1: 152220456-152220456 on Build GRCh38
rs764918396 Chr.1: 152220460-152220460 on Build GRCh38
rs747860789 Chr.1: 152220307-152220307 on Build GRCh38
rs6587649 Chr.1: 152220502-152220502 on Build GRCh38
rs772226648 Chr.1: 152220338-152220338 on Build GRCh38
rs1022476504 Chr.1: 152220402-152220402 on Build GRCh38
rs764816260 Chr.1: 152220511-152220511 on Build GRCh38
rs752238836 Chr.1: 152220390-152220390 on Build GRCh38
rs555213189 Chr.1: 152220344-152220344 on Build GRCh38
rs921378070 Chr.1: 152220482-152220482 on Build GRCh38
rs142030922 Chr.1: 152220406-152220406 on Build GRCh38
rs761190409 Chr.1: 152220506-152220506 on Build GRCh38
rs753542098 Chr.1: 152220329-152220344 on Build GRCh38
rs1051116214 Chr.1: 152220496-152220496 on Build GRCh38
rs781342429 Chr.1: 152220486-152220486 on Build GRCh38
rs760722907 Chr.1: 152220503-152220503 on Build GRCh38
rs774876762 Chr.1: 152220327-152220327 on Build GRCh38
rs367548706 Chr.1: 152220350-152220350 on Build GRCh38
rs200559761 Chr.1: 152220475-152220475 on Build GRCh38
rs774133101 Chr.1: 152220440-152220440 on Build GRCh38
rs767373809 Chr.1: 152220389-152220389 on Build GRCh38
rs759480390 Chr.1: 152220388-152220388 on Build GRCh38
rs757163383 Chr.1: 152220401-152220401 on Build GRCh38
rs566122590 Chr.1: 152220490-152220490 on Build GRCh38
rs771375980 Chr.1: 152220322-152220322 on Build GRCh38
rs950682399 Chr.1: 152220442-152220442 on Build GRCh38
rs138689834 Chr.1: 152220428-152220428 on Build GRCh38
rs138118811 Chr.1: 152220392-152220392 on Build GRCh38
rs941466916 Chr.1: 152220370-152220370 on Build GRCh38
rs758881961 Chr.1: 152220467-152220467 on Build GRCh38
rs867365807 Chr.1: 152220497-152220497 on Build GRCh38

View Full Product Details