Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4408791-4409058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAF1A
Entrez Gene ID
10036
Gene Name
chromatin assembly factor 1 subunit A
Gene Aliases
CAF-1, CAF1, CAF1B, CAF1P150, P150
UniGene
Hs.79018
Gene Chromosome Location
Chr.19: 4402596-4448322 on Build GRCh38
Transcripts
NM_005483.2 XM_011527605.2 XM_011527606.2 XM_011527607.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6151605 c.107G>T p.R36L Chr.19: 4408906-4408906 on Build GRCh38
997642 c.237T>C p.C79C Chr.19: 4409036-4409036 on Build GRCh38
1304703 c.179C>T p.S60F Chr.19: 4408978-4408978 on Build GRCh38
5644156 c.194G>A p.S65N Chr.19: 4408993-4408993 on Build GRCh38
6839704 c.161C>G p.S54* Chr.19: 4408960-4408960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866626587 Chr.19: 4408912-4408912 on Build GRCh38
rs773021866 Chr.19: 4408993-4408993 on Build GRCh38
rs767161454 Chr.19: 4408937-4408937 on Build GRCh38
rs761728549 Chr.19: 4408879-4408879 on Build GRCh38
rs752386071 Chr.19: 4409032-4409032 on Build GRCh38
rs113787645 Chr.19: 4409013-4409013 on Build GRCh38
rs1022878838 Chr.19: 4408817-4408817 on Build GRCh38
rs756494953 Chr.19: 4408949-4408949 on Build GRCh38
rs891582638 Chr.19: 4409020-4409020 on Build GRCh38
rs772375979 Chr.19: 4408972-4408972 on Build GRCh38
rs374615768 Chr.19: 4408905-4408905 on Build GRCh38
rs369896445 Chr.19: 4408890-4408890 on Build GRCh38
rs1047745775 Chr.19: 4408834-4408834 on Build GRCh38
rs574909689 Chr.19: 4408854-4408854 on Build GRCh38
rs930717219 Chr.19: 4408827-4408827 on Build GRCh38
rs368639662 Chr.19: 4408964-4408964 on Build GRCh38
rs769755031 Chr.19: 4408980-4408980 on Build GRCh38
rs771080344 Chr.19: 4408866-4408866 on Build GRCh38
rs61729782 Chr.19: 4408965-4408965 on Build GRCh38
rs749870729 Chr.19: 4408952-4408952 on Build GRCh38
rs373142255 Chr.19: 4408956-4408956 on Build GRCh38
rs377165063 Chr.19: 4408872-4408872 on Build GRCh38
rs544391423 Chr.19: 4408841-4408841 on Build GRCh38
rs968368846 Chr.19: 4408818-4408818 on Build GRCh38
rs997164464 Chr.19: 4409007-4409007 on Build GRCh38
rs751161984 Chr.19: 4408933-4408933 on Build GRCh38
rs562687602 Chr.19: 4408844-4408844 on Build GRCh38
rs776498230 Chr.19: 4408870-4408870 on Build GRCh38
rs772432849 Chr.19: 4408869-4408869 on Build GRCh38
rs144933262 Chr.19: 4408913-4408913 on Build GRCh38
rs371002759 Chr.19: 4408978-4408978 on Build GRCh38
rs1038133005 Chr.19: 4408817-4408817 on Build GRCh38
rs370633320 Chr.19: 4408950-4408950 on Build GRCh38
rs919363683 Chr.19: 4408924-4408924 on Build GRCh38
rs767110124 Chr.19: 4409019-4409019 on Build GRCh38
rs754602845 Chr.19: 4408935-4408935 on Build GRCh38
rs757543664 Chr.19: 4408891-4408891 on Build GRCh38
rs8105764 Chr.19: 4408940-4408940 on Build GRCh38
rs372419158 Chr.19: 4408868-4408868 on Build GRCh38
rs775875692 Chr.19: 4408973-4408973 on Build GRCh38
rs2230633 Chr.19: 4409009-4409009 on Build GRCh38
rs763009084 Chr.19: 4408994-4408994 on Build GRCh38
rs376324554 Chr.19: 4408969-4408969 on Build GRCh38
rs765958815 Chr.19: 4408861-4408861 on Build GRCh38

View Full Product Details