Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 123624390-123624635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB15
Entrez Gene ID
142685
Gene Name
ankyrin repeat and SOCS box containing 15
Gene Aliases
-
UniGene
Hs.83916
Gene Chromosome Location
Chr.7: 123567003-123637900 on Build GRCh38
Transcripts
NM_001290258.1 NM_080928.4 XM_005250146.4 XM_011515816.2 XM_011515817.2 XM_011515818.2 XM_011515819.2 XM_011515820.2 XM_011515821.2 XM_017011756.1 XM_017011757.1 XM_017011758.1 NR_111922.1

Gene Symbol
LOC102724555
Entrez Gene ID
102724555
Gene Name
uncharacterized LOC102724555
Gene Aliases
-
UniGene
Hs.83916
Gene Chromosome Location
Chr.7: 123614161-123624957 on Build GRCh38
Transcripts
NM_001290258.1 NM_080928.4 XM_005250146.4 XM_011515816.2 XM_011515817.2 XM_011515818.2 XM_011515819.2 XM_011515820.2 XM_011515821.2 XM_017011756.1 XM_017011757.1 XM_017011758.1 NR_111922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5543734 c.464G>A p.G155D Chr.7: 123624581-123624581 on Build GRCh38
7356561 c.472G>A p.D158N Chr.7: 123624589-123624589 on Build GRCh38
3877726 c.518C>A p.P173H Chr.7: 123624635-123624635 on Build GRCh38
6605512 c.457delA p.K154fs*10 Chr.7: 123624574-123624574 on Build GRCh38
4663633 c.458A>G p.K153R Chr.7: 123624575-123624575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs267601261 Chr.7: 123624585-123624585 on Build GRCh38
rs149901686 Chr.7: 123624477-123624477 on Build GRCh38
rs537865484 Chr.7: 123624503-123624503 on Build GRCh38
rs569292163 Chr.7: 123624485-123624485 on Build GRCh38
rs200912565 Chr.7: 123624568-123624568 on Build GRCh38
rs558784382 Chr.7: 123624478-123624478 on Build GRCh38
rs756550709 Chr.7: 123624535-123624535 on Build GRCh38
rs769593680 Chr.7: 123624582-123624582 on Build GRCh38
rs761947941 Chr.7: 123624607-123624607 on Build GRCh38
rs764061642 Chr.7: 123624595-123624595 on Build GRCh38
rs201822867 Chr.7: 123624512-123624512 on Build GRCh38
rs753386880 Chr.7: 123624542-123624543 on Build GRCh38
rs762910488 Chr.7: 123624593-123624593 on Build GRCh38
rs778221995 Chr.7: 123624570-123624575 on Build GRCh38
rs750685654 Chr.7: 123624611-123624611 on Build GRCh38
rs774344855 Chr.7: 123624599-123624599 on Build GRCh38
rs770552026 Chr.7: 123624559-123624559 on Build GRCh38
rs772681696 Chr.7: 123624579-123624579 on Build GRCh38
rs781462962 Chr.7: 123624523-123624523 on Build GRCh38
rs755470063 Chr.7: 123624552-123624552 on Build GRCh38
rs574470030 Chr.7: 123624588-123624588 on Build GRCh38
rs777184880 Chr.7: 123624553-123624553 on Build GRCh38
rs750884067 Chr.7: 123624528-123624528 on Build GRCh38
rs780507846 Chr.7: 123624544-123624544 on Build GRCh38
rs949181898 Chr.7: 123624465-123624465 on Build GRCh38
rs747750225 Chr.7: 123624574-123624574 on Build GRCh38
rs749833044 Chr.7: 123624545-123624545 on Build GRCh38
rs767430385 Chr.7: 123624608-123624608 on Build GRCh38
rs746502188 Chr.7: 123624554-123624554 on Build GRCh38
rs754676417 Chr.7: 123624547-123624547 on Build GRCh38
rs887309184 Chr.7: 123624493-123624493 on Build GRCh38

View Full Product Details