Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14844837-14845080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
UniGene
Hs.554740
Gene Chromosome Location
Chr.X: 14690863-14873255 on Build GRCh38
Transcripts
NM_001018113.2 NM_001324162.1 NM_152633.3 XM_011545470.2 XM_017029355.1 XM_017029356.1

SNP Detail

SNP ID Chromosome Location
rs148560784 Chr.X: 14844966-14844966 on Build GRCh38
rs894355393 Chr.X: 14844916-14844916 on Build GRCh38
rs986844479 Chr.X: 14844977-14844977 on Build GRCh38
rs964099207 Chr.X: 14845016-14845016 on Build GRCh38
rs1021143964 Chr.X: 14844946-14844946 on Build GRCh38
rs754392611 Chr.X: 14845038-14845038 on Build GRCh38
rs1032511086 Chr.X: 14844910-14844910 on Build GRCh38
rs144764663 Chr.X: 14844945-14844945 on Build GRCh38
rs975151235 Chr.X: 14845018-14845018 on Build GRCh38
rs771950360 Chr.X: 14845047-14845047 on Build GRCh38
rs750639868 Chr.X: 14844937-14844937 on Build GRCh38
rs149387792 Chr.X: 14844889-14844889 on Build GRCh38
rs756702783 Chr.X: 14844997-14844997 on Build GRCh38
rs745472535 Chr.X: 14845028-14845028 on Build GRCh38
rs142959373 Chr.X: 14845014-14845014 on Build GRCh38
rs200948146 Chr.X: 14844883-14844883 on Build GRCh38
rs763366953 Chr.X: 14844942-14844942 on Build GRCh38
rs752308935 Chr.X: 14844995-14844995 on Build GRCh38
rs772005838 Chr.X: 14844868-14844868 on Build GRCh38
rs770548473 Chr.X: 14845027-14845027 on Build GRCh38
rs772448306 Chr.X: 14845029-14845029 on Build GRCh38
rs781690574 Chr.X: 14844972-14844972 on Build GRCh38
rs751885600 Chr.X: 14844949-14844949 on Build GRCh38
rs748958265 Chr.X: 14845024-14845024 on Build GRCh38
rs776713934 Chr.X: 14844871-14844871 on Build GRCh38

View Full Product Details