Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50382411-50382682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKK
Entrez Gene ID
139189
Gene Name
diacylglycerol kinase kappa
Gene Aliases
-
UniGene
Hs.38132
Gene Chromosome Location
Chr.X: 50365407-50470850 on Build GRCh38
Transcripts
NM_001013742.3 XM_017029268.1

SNP Detail

SNP ID Chromosome Location
rs781878595 Chr.X: 50382509-50382509 on Build GRCh38
rs782495115 Chr.X: 50382474-50382474 on Build GRCh38
rs782585742 Chr.X: 50382571-50382571 on Build GRCh38
rs541765470 Chr.X: 50382567-50382567 on Build GRCh38
rs782402579 Chr.X: 50382603-50382603 on Build GRCh38
rs782274310 Chr.X: 50382547-50382547 on Build GRCh38
rs945166912 Chr.X: 50382476-50382476 on Build GRCh38
rs900811629 Chr.X: 50382584-50382584 on Build GRCh38
rs1015868933 Chr.X: 50382631-50382631 on Build GRCh38
rs199995789 Chr.X: 50382635-50382635 on Build GRCh38
rs782290115 Chr.X: 50382653-50382653 on Build GRCh38
rs187259548 Chr.X: 50382630-50382630 on Build GRCh38
rs375771283 Chr.X: 50382523-50382523 on Build GRCh38
rs782685434 Chr.X: 50382525-50382525 on Build GRCh38
rs782726582 Chr.X: 50382457-50382457 on Build GRCh38
rs782330201 Chr.X: 50382566-50382566 on Build GRCh38
rs782230209 Chr.X: 50382573-50382573 on Build GRCh38
rs368247344 Chr.X: 50382601-50382601 on Build GRCh38
rs996470723 Chr.X: 50382595-50382595 on Build GRCh38
rs782599525 Chr.X: 50382493-50382493 on Build GRCh38
rs371175247 Chr.X: 50382452-50382452 on Build GRCh38

View Full Product Details