Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68756561-68757065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPT1A
Entrez Gene ID
1374
Gene Name
carnitine palmitoyltransferase 1A
Gene Aliases
CPT1, CPT1-L, L-CPT1
UniGene
Hs.503043
Gene Chromosome Location
Chr.11: 68754620-68844410 on Build GRCh38
Transcripts
NM_001031847.2 NM_001876.3 XM_005273762.2 XM_005273763.1

SNP Detail

SNP ID Chromosome Location
rs530083614 Chr.11: 68756846-68756846 on Build GRCh38
rs533401453 Chr.11: 68756916-68756916 on Build GRCh38
rs1025920403 Chr.11: 68756914-68756914 on Build GRCh38
rs906406348 Chr.11: 68756596-68756596 on Build GRCh38
rs1055942722 Chr.11: 68756848-68756848 on Build GRCh38
rs1053756084 Chr.11: 68756637-68756637 on Build GRCh38
rs565493475 Chr.11: 68756792-68756792 on Build GRCh38
rs771243449 Chr.11: 68756814-68756814 on Build GRCh38
rs749467869 Chr.11: 68756693-68756693 on Build GRCh38
rs532797427 Chr.11: 68756821-68756821 on Build GRCh38
rs961647173 Chr.11: 68756880-68756880 on Build GRCh38
rs566652518 Chr.11: 68756881-68756881 on Build GRCh38
rs544584298 Chr.11: 68756842-68756842 on Build GRCh38
rs952611924 Chr.11: 68756616-68756616 on Build GRCh38
rs562899721 Chr.11: 68756843-68756843 on Build GRCh38
rs964028794 Chr.11: 68756709-68756709 on Build GRCh38
rs895987150 Chr.11: 68756645-68756645 on Build GRCh38
rs778043060 Chr.11: 68756595-68756595 on Build GRCh38
rs964416093 Chr.11: 68757013-68757013 on Build GRCh38
rs946268134 Chr.11: 68756809-68756809 on Build GRCh38
rs373835418 Chr.11: 68756789-68756789 on Build GRCh38
rs1026612349 Chr.11: 68756882-68756882 on Build GRCh38
rs976855927 Chr.11: 68756774-68756774 on Build GRCh38
rs371693398 Chr.11: 68756600-68756600 on Build GRCh38
rs548263723 Chr.11: 68756856-68756856 on Build GRCh38
rs993893279 Chr.11: 68756892-68756892 on Build GRCh38
rs772527141 Chr.11: 68756951-68756951 on Build GRCh38
rs984016172 Chr.11: 68756628-68756628 on Build GRCh38
rs1025442343 Chr.11: 68756608-68756608 on Build GRCh38
rs954576920 Chr.11: 68756967-68756967 on Build GRCh38
rs772298375 Chr.11: 68756750-68756752 on Build GRCh38
rs149870808 Chr.11: 68756602-68756603 on Build GRCh38
rs978451266 Chr.11: 68757039-68757039 on Build GRCh38
rs546355771 Chr.11: 68757044-68757044 on Build GRCh38
rs894842128 Chr.11: 68756850-68756850 on Build GRCh38
rs527801838 Chr.11: 68756976-68756976 on Build GRCh38
rs888254452 Chr.11: 68756920-68756920 on Build GRCh38
rs947722203 Chr.11: 68756858-68756858 on Build GRCh38
rs922674099 Chr.11: 68756776-68756776 on Build GRCh38
rs964078432 Chr.11: 68757021-68757021 on Build GRCh38
rs774523688 Chr.11: 68756859-68756859 on Build GRCh38
rs2278907 Chr.11: 68756585-68756585 on Build GRCh38
rs796270955 Chr.11: 68756679-68756679 on Build GRCh38
rs910325643 Chr.11: 68756991-68756992 on Build GRCh38

View Full Product Details