Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 71996469-71996966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF121
Entrez Gene ID
55298
Gene Name
ring finger protein 121
Gene Aliases
-
UniGene
Hs.368554
Gene Chromosome Location
Chr.11: 71928701-71997597 on Build GRCh38
Transcripts
NM_001300926.1 NM_018320.4 NR_024147.1 NR_024148.1 XM_006718629.1 XM_006718630.1 XM_006718632.1 XM_011545150.1

SNP Detail

SNP ID Chromosome Location
rs191733941 Chr.11: 71996936-71996936 on Build GRCh38
rs181124629 Chr.11: 71996668-71996668 on Build GRCh38
rs757558784 Chr.11: 71996530-71996530 on Build GRCh38
rs570442062 Chr.11: 71996932-71996932 on Build GRCh38
rs186780651 Chr.11: 71996830-71996830 on Build GRCh38
rs1035971181 Chr.11: 71996736-71996736 on Build GRCh38
rs980041165 Chr.11: 71996634-71996634 on Build GRCh38
rs995077991 Chr.11: 71996654-71996654 on Build GRCh38
rs137972494 Chr.11: 71996772-71996772 on Build GRCh38
rs897608317 Chr.11: 71996748-71996748 on Build GRCh38
rs113699392 Chr.11: 71996812-71996812 on Build GRCh38
rs926806889 Chr.11: 71996639-71996639 on Build GRCh38
rs1056686208 Chr.11: 71996689-71996689 on Build GRCh38
rs940263799 Chr.11: 71996857-71996857 on Build GRCh38
rs532364485 Chr.11: 71996713-71996713 on Build GRCh38
rs556778298 Chr.11: 71996791-71996791 on Build GRCh38
rs914011844 Chr.11: 71996595-71996595 on Build GRCh38
rs538908113 Chr.11: 71996527-71996527 on Build GRCh38
rs772613481 Chr.11: 71996579-71996579 on Build GRCh38
rs11235416 Chr.11: 71996773-71996773 on Build GRCh38
rs771978247 Chr.11: 71996690-71996690 on Build GRCh38
rs953454934 Chr.11: 71996494-71996494 on Build GRCh38
rs916929649 Chr.11: 71996696-71996696 on Build GRCh38
rs1046078810 Chr.11: 71996798-71996798 on Build GRCh38
rs566540209 Chr.11: 71996792-71996792 on Build GRCh38
rs368646464 Chr.11: 71996562-71996562 on Build GRCh38
rs371295709 Chr.11: 71996715-71996715 on Build GRCh38
rs34646174 Chr.11: 71996579-71996579 on Build GRCh38
rs565286944 Chr.11: 71996709-71996709 on Build GRCh38
rs918921659 Chr.11: 71996909-71996909 on Build GRCh38
rs542364922 Chr.11: 71996938-71996938 on Build GRCh38
rs1034273010 Chr.11: 71996821-71996821 on Build GRCh38
rs867053062 Chr.11: 71996757-71996757 on Build GRCh38
rs968109251 Chr.11: 71996598-71996598 on Build GRCh38
rs1046448369 Chr.11: 71996560-71996560 on Build GRCh38
rs898912968 Chr.11: 71996830-71996833 on Build GRCh38
rs1009255520 Chr.11: 71996888-71996888 on Build GRCh38
rs565880584 Chr.11: 71996526-71996526 on Build GRCh38
rs550680412 Chr.11: 71996750-71996750 on Build GRCh38
rs918194787 Chr.11: 71996855-71996855 on Build GRCh38
rs947753534 Chr.11: 71996544-71996544 on Build GRCh38
rs1018853716 Chr.11: 71996918-71996918 on Build GRCh38
rs994690393 Chr.11: 71996856-71996856 on Build GRCh38
rs369628902 Chr.11: 71996580-71996580 on Build GRCh38
rs959541247 Chr.11: 71996823-71996823 on Build GRCh38
rs979500237 Chr.11: 71996813-71996813 on Build GRCh38
rs992840915 Chr.11: 71996825-71996825 on Build GRCh38
rs971738690 Chr.11: 71996906-71996906 on Build GRCh38
rs539719796 Chr.11: 71996883-71996883 on Build GRCh38
rs1049407026 Chr.11: 71996752-71996752 on Build GRCh38
rs568984392 Chr.11: 71996762-71996762 on Build GRCh38
rs899029618 Chr.11: 71996617-71996617 on Build GRCh38
rs888917241 Chr.11: 71996754-71996754 on Build GRCh38
rs953385544 Chr.11: 71996669-71996669 on Build GRCh38

View Full Product Details