Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72685359-72685851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARAP1
Entrez Gene ID
116985
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1
Gene Aliases
CENTD2
UniGene
Hs.503165
Gene Chromosome Location
Chr.11: 72685069-72752389 on Build GRCh38
Transcripts
NM_001040118.2 NM_001135190.1 NM_015242.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3967822 c.4343G>C p.R1448P Chr.11: 72685674-72685674 on Build GRCh38
3967823 c.3608G>C p.R1203P Chr.11: 72685674-72685674 on Build GRCh38
5373553 c.4352G>A p.*1451* Chr.11: 72685665-72685665 on Build GRCh38
5373554 c.3617G>A p.*1206* Chr.11: 72685665-72685665 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs36094095 Chr.11: 72685608-72685608 on Build GRCh38
rs894104604 Chr.11: 72685534-72685534 on Build GRCh38
rs781087301 Chr.11: 72685709-72685709 on Build GRCh38
rs765021123 Chr.11: 72685678-72685678 on Build GRCh38
rs12802369 Chr.11: 72685755-72685755 on Build GRCh38
rs367927606 Chr.11: 72685619-72685619 on Build GRCh38
rs554373802 Chr.11: 72685455-72685455 on Build GRCh38
rs1013159681 Chr.11: 72685567-72685567 on Build GRCh38
rs747706673 Chr.11: 72685765-72685765 on Build GRCh38
rs1009415682 Chr.11: 72685785-72685785 on Build GRCh38
rs536409167 Chr.11: 72685431-72685431 on Build GRCh38
rs761907677 Chr.11: 72685824-72685824 on Build GRCh38
rs925746449 Chr.11: 72685736-72685736 on Build GRCh38
rs568978971 Chr.11: 72685410-72685410 on Build GRCh38
rs752563680 Chr.11: 72685818-72685818 on Build GRCh38
rs762040438 Chr.11: 72685505-72685505 on Build GRCh38
rs924369841 Chr.11: 72685720-72685720 on Build GRCh38
rs576893580 Chr.11: 72685598-72685598 on Build GRCh38
rs375398526 Chr.11: 72685731-72685731 on Build GRCh38
rs750867288 Chr.11: 72685574-72685574 on Build GRCh38
rs540312548 Chr.11: 72685754-72685754 on Build GRCh38
rs1045943544 Chr.11: 72685750-72685750 on Build GRCh38
rs775780604 Chr.11: 72685760-72685760 on Build GRCh38
rs963949823 Chr.11: 72685632-72685632 on Build GRCh38
rs768413125 Chr.11: 72685629-72685629 on Build GRCh38
rs932640016 Chr.11: 72685416-72685416 on Build GRCh38
rs1056129254 Chr.11: 72685511-72685511 on Build GRCh38
rs779165098 Chr.11: 72685407-72685407 on Build GRCh38
rs1045480175 Chr.11: 72685550-72685550 on Build GRCh38
rs779351850 Chr.11: 72685762-72685762 on Build GRCh38
rs757217540 Chr.11: 72685602-72685602 on Build GRCh38
rs148980436 Chr.11: 72685746-72685754 on Build GRCh38
rs754388817 Chr.11: 72685450-72685450 on Build GRCh38
rs575765918 Chr.11: 72685701-72685701 on Build GRCh38
rs1024718160 Chr.11: 72685568-72685568 on Build GRCh38
rs151103578 Chr.11: 72685670-72685670 on Build GRCh38
rs765661985 Chr.11: 72685549-72685549 on Build GRCh38
rs1041609502 Chr.11: 72685451-72685452 on Build GRCh38
rs1051043472 Chr.11: 72685566-72685566 on Build GRCh38
rs183975270 Chr.11: 72685393-72685393 on Build GRCh38
rs572847266 Chr.11: 72685504-72685504 on Build GRCh38
rs781342501 Chr.11: 72685607-72685607 on Build GRCh38
rs759390723 Chr.11: 72685675-72685675 on Build GRCh38
rs1000289998 Chr.11: 72685471-72685471 on Build GRCh38
rs111787770 Chr.11: 72685732-72685732 on Build GRCh38
rs933661510 Chr.11: 72685409-72685409 on Build GRCh38
rs766345289 Chr.11: 72685672-72685672 on Build GRCh38
rs760282346 Chr.11: 72685777-72685777 on Build GRCh38
rs770362708 Chr.11: 72685611-72685611 on Build GRCh38
rs372111341 Chr.11: 72685663-72685663 on Build GRCh38
rs977501998 Chr.11: 72685719-72685719 on Build GRCh38
rs1041430169 Chr.11: 72685413-72685413 on Build GRCh38
rs771787599 Chr.11: 72685766-72685766 on Build GRCh38
rs771426987 Chr.11: 72685656-72685656 on Build GRCh38
rs375393530 Chr.11: 72685597-72685597 on Build GRCh38
rs558628310 Chr.11: 72685573-72685573 on Build GRCh38
rs749722879 Chr.11: 72685628-72685628 on Build GRCh38
rs764962729 Chr.11: 72685816-72685816 on Build GRCh38
rs763245592 Chr.11: 72685778-72685781 on Build GRCh38
rs759482592 Chr.11: 72685815-72685815 on Build GRCh38
rs781057706 Chr.11: 72685784-72685784 on Build GRCh38
rs776482508 Chr.11: 72685789-72685789 on Build GRCh38
rs191460924 Chr.11: 72685744-72685744 on Build GRCh38
rs774219037 Chr.11: 72685651-72685651 on Build GRCh38
rs766731509 Chr.11: 72685818-72685847 on Build GRCh38
rs967477093 Chr.11: 72685691-72685691 on Build GRCh38
rs199584464 Chr.11: 72685674-72685674 on Build GRCh38
rs1054549113 Chr.11: 72685740-72685740 on Build GRCh38
rs561230618 Chr.11: 72685741-72685741 on Build GRCh38
rs761535213 Chr.11: 72685655-72685655 on Build GRCh38
rs543166752 Chr.11: 72685726-72685726 on Build GRCh38
rs1019026655 Chr.11: 72685676-72685676 on Build GRCh38
rs750420721 Chr.11: 72685408-72685408 on Build GRCh38
rs187762518 Chr.11: 72685572-72685572 on Build GRCh38
rs111571611 Chr.11: 72685799-72685799 on Build GRCh38
rs925839975 Chr.11: 72685461-72685461 on Build GRCh38

View Full Product Details