Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 76446809-76447314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMSY
Entrez Gene ID
56946
Gene Name
EMSY, BRCA2 interacting transcriptional repressor
Gene Aliases
C11orf30, GL002
UniGene
Hs.352588
Gene Chromosome Location
Chr.11: 76444928-76552901 on Build GRCh38
Transcripts
NM_001300942.1 NM_001300943.1 NM_001300944.1 NM_020193.4 XM_005274112.4 XM_006718637.3 XM_006718638.3 XM_006718639.3 XM_011545173.2 XM_011545174.2 XM_011545175.2 XM_011545177.2 XM_011545178.2 XM_011545179.2 XM_017018021.1 XM_017018022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274196 c.53G>T p.R18I Chr.11: 76446991-76446991 on Build GRCh38
3953813 c.61C>T p.R21* Chr.11: 76446999-76446999 on Build GRCh38
380842 c.53G>C p.R18T Chr.11: 76446991-76446991 on Build GRCh38
3810447 c.58C>G p.L20V Chr.11: 76446996-76446996 on Build GRCh38
4567423 c.69_70GG>AA p.E24K Chr.11: 76447007-76447008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908203810 Chr.11: 76447135-76447135 on Build GRCh38
rs955352479 Chr.11: 76447045-76447045 on Build GRCh38
rs760683065 Chr.11: 76446946-76446946 on Build GRCh38
rs764999766 Chr.11: 76446980-76446980 on Build GRCh38
rs965329428 Chr.11: 76446943-76446943 on Build GRCh38
rs1055689493 Chr.11: 76447269-76447269 on Build GRCh38
rs774181297 Chr.11: 76446879-76446879 on Build GRCh38
rs749400485 Chr.11: 76446909-76446909 on Build GRCh38
rs780208701 Chr.11: 76446901-76446901 on Build GRCh38
rs937675175 Chr.11: 76447166-76447166 on Build GRCh38
rs887267644 Chr.11: 76446870-76446870 on Build GRCh38
rs767903513 Chr.11: 76447005-76447005 on Build GRCh38
rs542818656 Chr.11: 76447035-76447035 on Build GRCh38
rs988462104 Chr.11: 76447085-76447085 on Build GRCh38
rs1047752061 Chr.11: 76447252-76447252 on Build GRCh38
rs779027843 Chr.11: 76447047-76447047 on Build GRCh38
rs899885839 Chr.11: 76447287-76447287 on Build GRCh38
rs926349951 Chr.11: 76447146-76447148 on Build GRCh38
rs748001924 Chr.11: 76446921-76446921 on Build GRCh38
rs1056097541 Chr.11: 76447171-76447171 on Build GRCh38
rs770722638 Chr.11: 76446834-76446834 on Build GRCh38
rs983893758 Chr.11: 76447110-76447110 on Build GRCh38
rs931035730 Chr.11: 76447090-76447090 on Build GRCh38
rs756112263 Chr.11: 76446893-76446893 on Build GRCh38
rs778711970 Chr.11: 76447050-76447050 on Build GRCh38
rs555518643 Chr.11: 76447193-76447193 on Build GRCh38
rs367735381 Chr.11: 76446879-76446879 on Build GRCh38
rs772102626 Chr.11: 76446922-76446922 on Build GRCh38
rs567243197 Chr.11: 76447285-76447285 on Build GRCh38
rs921110049 Chr.11: 76447072-76447072 on Build GRCh38
rs547077035 Chr.11: 76447261-76447261 on Build GRCh38
rs1025843323 Chr.11: 76446859-76446859 on Build GRCh38
rs370227811 Chr.11: 76446923-76446923 on Build GRCh38
rs1014843151 Chr.11: 76446852-76446853 on Build GRCh38
rs1046072320 Chr.11: 76447194-76447194 on Build GRCh38
rs776210053 Chr.11: 76446960-76446960 on Build GRCh38
rs745973291 Chr.11: 76447134-76447134 on Build GRCh38
rs753482765 Chr.11: 76447021-76447022 on Build GRCh38
rs939715589 Chr.11: 76447137-76447137 on Build GRCh38
rs1019918238 Chr.11: 76446887-76446887 on Build GRCh38
rs531133991 Chr.11: 76447262-76447262 on Build GRCh38
rs950367800 Chr.11: 76447235-76447235 on Build GRCh38
rs756662085 Chr.11: 76447015-76447015 on Build GRCh38
rs779316533 Chr.11: 76447033-76447033 on Build GRCh38
rs745781013 Chr.11: 76446957-76446957 on Build GRCh38
rs765175953 Chr.11: 76447268-76447268 on Build GRCh38
rs533870483 Chr.11: 76446993-76446993 on Build GRCh38
rs762207865 Chr.11: 76447000-76447000 on Build GRCh38
rs572857622 Chr.11: 76447102-76447102 on Build GRCh38
rs894001416 Chr.11: 76447273-76447273 on Build GRCh38
rs750884526 Chr.11: 76447013-76447013 on Build GRCh38
rs768985956 Chr.11: 76446918-76446918 on Build GRCh38
rs906755084 Chr.11: 76447213-76447213 on Build GRCh38
rs756997261 Chr.11: 76447027-76447028 on Build GRCh38
rs552798502 Chr.11: 76447094-76447094 on Build GRCh38
rs745738791 Chr.11: 76447061-76447062 on Build GRCh38
rs566964458 Chr.11: 76447289-76447289 on Build GRCh38
rs759107180 Chr.11: 76446968-76446968 on Build GRCh38
rs544721487 Chr.11: 76447144-76447144 on Build GRCh38
rs766361879 Chr.11: 76447016-76447016 on Build GRCh38
rs1056467793 Chr.11: 76446830-76446830 on Build GRCh38
rs913760870 Chr.11: 76447098-76447098 on Build GRCh38
rs754004345 Chr.11: 76447020-76447020 on Build GRCh38
rs750473954 Chr.11: 76446890-76446890 on Build GRCh38
rs1030002675 Chr.11: 76446963-76446963 on Build GRCh38
rs755130440 Chr.11: 76447032-76447032 on Build GRCh38
rs778835986 Chr.11: 76446919-76446919 on Build GRCh38
rs143879211 Chr.11: 76446937-76446937 on Build GRCh38

View Full Product Details