Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 106021922-106022451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSANTD4
Entrez Gene ID
84437
Gene Name
Myb/SANT DNA binding domain containing 4 with coiled-coils
Gene Aliases
KIAA1826
UniGene
Hs.266782
Gene Chromosome Location
Chr.11: 106007920-106022287 on Build GRCh38
Transcripts
NM_001318747.1 NM_001318748.1 NM_001318749.1 NM_001318750.1 NM_032424.2 XM_011543022.2 XM_017018416.1 XM_017018417.1

SNP Detail

SNP ID Chromosome Location
rs946404849 Chr.11: 106022416-106022416 on Build GRCh38
rs964783109 Chr.11: 106021959-106021959 on Build GRCh38
rs902238375 Chr.11: 106022237-106022237 on Build GRCh38
rs915188287 Chr.11: 106022378-106022378 on Build GRCh38
rs559020417 Chr.11: 106022041-106022041 on Build GRCh38
rs559579408 Chr.11: 106022360-106022360 on Build GRCh38
rs955298644 Chr.11: 106022164-106022164 on Build GRCh38
rs532225958 Chr.11: 106022274-106022274 on Build GRCh38
rs944072068 Chr.11: 106022417-106022417 on Build GRCh38
rs1009342101 Chr.11: 106022167-106022167 on Build GRCh38
rs117177843 Chr.11: 106022119-106022119 on Build GRCh38
rs1041154929 Chr.11: 106022422-106022422 on Build GRCh38
rs979049462 Chr.11: 106022431-106022431 on Build GRCh38
rs150695004 Chr.11: 106022234-106022234 on Build GRCh38
rs910663711 Chr.11: 106021956-106021956 on Build GRCh38
rs928215828 Chr.11: 106022019-106022019 on Build GRCh38
rs543578723 Chr.11: 106022039-106022039 on Build GRCh38
rs952984546 Chr.11: 106022255-106022255 on Build GRCh38
rs570582794 Chr.11: 106022097-106022097 on Build GRCh38
rs1020405736 Chr.11: 106022029-106022029 on Build GRCh38
rs569374062 Chr.11: 106022261-106022261 on Build GRCh38
rs961122695 Chr.11: 106022409-106022409 on Build GRCh38
rs928232481 Chr.11: 106022224-106022224 on Build GRCh38
rs982939036 Chr.11: 106022182-106022182 on Build GRCh38
rs945182833 Chr.11: 106022398-106022398 on Build GRCh38
rs372949171 Chr.11: 106022147-106022147 on Build GRCh38
rs997470775 Chr.11: 106022248-106022248 on Build GRCh38
rs748898471 Chr.11: 106022089-106022089 on Build GRCh38
rs893647703 Chr.11: 106021964-106021964 on Build GRCh38
rs200534507 Chr.11: 106022169-106022169 on Build GRCh38
rs994815041 Chr.11: 106022313-106022313 on Build GRCh38
rs530064449 Chr.11: 106022423-106022423 on Build GRCh38
rs999789233 Chr.11: 106022239-106022239 on Build GRCh38
rs553135240 Chr.11: 106022207-106022207 on Build GRCh38
rs796075528 Chr.11: 106022322-106022322 on Build GRCh38
rs73554710 Chr.11: 106022298-106022298 on Build GRCh38
rs920764872 Chr.11: 106022011-106022011 on Build GRCh38
rs981833794 Chr.11: 106022343-106022343 on Build GRCh38
rs1048462870 Chr.11: 106022016-106022016 on Build GRCh38
rs770800466 Chr.11: 106022075-106022075 on Build GRCh38
rs916803815 Chr.11: 106022415-106022415 on Build GRCh38
rs139950727 Chr.11: 106022262-106022262 on Build GRCh38
rs766447498 Chr.11: 106022341-106022341 on Build GRCh38
rs112207730 Chr.11: 106021994-106021994 on Build GRCh38
rs565015904 Chr.11: 106022264-106022264 on Build GRCh38
rs908321604 Chr.11: 106022205-106022205 on Build GRCh38
rs900515816 Chr.11: 106022312-106022312 on Build GRCh38
rs973493337 Chr.11: 106022196-106022196 on Build GRCh38
rs1027279942 Chr.11: 106022250-106022250 on Build GRCh38
rs939614077 Chr.11: 106022227-106022227 on Build GRCh38
rs929458925 Chr.11: 106022012-106022012 on Build GRCh38
rs570031447 Chr.11: 106022037-106022037 on Build GRCh38
rs377153195 Chr.11: 106022232-106022232 on Build GRCh38
rs1027763314 Chr.11: 106022337-106022337 on Build GRCh38
rs113072171 Chr.11: 106022032-106022032 on Build GRCh38
rs773169900 Chr.11: 106022278-106022278 on Build GRCh38
rs926251709 Chr.11: 106022356-106022356 on Build GRCh38
rs984056698 Chr.11: 106022400-106022400 on Build GRCh38
rs370396505 Chr.11: 106022129-106022129 on Build GRCh38
rs1006813850 Chr.11: 106022282-106022282 on Build GRCh38
rs950965219 Chr.11: 106022208-106022208 on Build GRCh38
rs192460292 Chr.11: 106022233-106022233 on Build GRCh38
rs991041177 Chr.11: 106022410-106022410 on Build GRCh38
rs937730477 Chr.11: 106022365-106022365 on Build GRCh38
rs548334848 Chr.11: 106022033-106022033 on Build GRCh38
rs964669314 Chr.11: 106022191-106022191 on Build GRCh38
rs1017615978 Chr.11: 106022185-106022185 on Build GRCh38
rs987466290 Chr.11: 106021946-106021946 on Build GRCh38
rs1035194653 Chr.11: 106022402-106022402 on Build GRCh38
rs983618705 Chr.11: 106022216-106022216 on Build GRCh38
rs71486038 Chr.11: 106022292-106022292 on Build GRCh38
rs1043470573 Chr.11: 106022236-106022236 on Build GRCh38

View Full Product Details