Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123642355-123642850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN3B
Entrez Gene ID
55800
Gene Name
sodium voltage-gated channel beta subunit 3
Gene Aliases
ATFB16, BRGDA7, HSA243396, SCNB3
UniGene
Hs.4865
Gene Chromosome Location
Chr.11: 123629187-123654607 on Build GRCh38
Transcripts
NM_001040151.1 NM_018400.3 XM_011542897.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
686762 c.304G>A p.D102N Chr.11: 123642587-123642587 on Build GRCh38
4724091 c.271C>T p.R91C Chr.11: 123642620-123642620 on Build GRCh38
1352182 c.371G>A p.R124Q Chr.11: 123642520-123642520 on Build GRCh38
76352 c.394C>T p.R132W Chr.11: 123642497-123642497 on Build GRCh38
5360987 c.353A>G p.Y118C Chr.11: 123642538-123642538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145148682 Chr.11: 123642819-123642819 on Build GRCh38
rs940598603 Chr.11: 123642809-123642809 on Build GRCh38
rs748870408 Chr.11: 123642421-123642421 on Build GRCh38
rs979787502 Chr.11: 123642784-123642784 on Build GRCh38
rs765466963 Chr.11: 123642555-123642555 on Build GRCh38
rs72552144 Chr.11: 123642477-123642478 on Build GRCh38
rs747452360 Chr.11: 123642521-123642521 on Build GRCh38
rs754912359 Chr.11: 123642487-123642487 on Build GRCh38
rs770801747 Chr.11: 123642475-123642475 on Build GRCh38
rs879253730 Chr.11: 123642468-123642468 on Build GRCh38
rs371050389 Chr.11: 123642631-123642631 on Build GRCh38
rs752683274 Chr.11: 123642579-123642579 on Build GRCh38
rs752593287 Chr.11: 123642495-123642495 on Build GRCh38
rs988405275 Chr.11: 123642802-123642802 on Build GRCh38
rs745365889 Chr.11: 123642507-123642507 on Build GRCh38
rs886038881 Chr.11: 123642528-123642528 on Build GRCh38
rs778618728 Chr.11: 123642620-123642620 on Build GRCh38
rs1783901 Chr.11: 123642798-123642798 on Build GRCh38
rs745437143 Chr.11: 123642621-123642621 on Build GRCh38
rs954926590 Chr.11: 123642471-123642471 on Build GRCh38
rs751464407 Chr.11: 123642398-123642398 on Build GRCh38
rs890414796 Chr.11: 123642732-123642732 on Build GRCh38
rs755967151 Chr.11: 123642496-123642496 on Build GRCh38
rs767283681 Chr.11: 123642576-123642576 on Build GRCh38
rs774536378 Chr.11: 123642698-123642698 on Build GRCh38
rs147205617 Chr.11: 123642563-123642563 on Build GRCh38
rs375545539 Chr.11: 123642658-123642658 on Build GRCh38
rs777551635 Chr.11: 123642418-123642418 on Build GRCh38
rs199937307 Chr.11: 123642564-123642564 on Build GRCh38
rs548044819 Chr.11: 123642455-123642455 on Build GRCh38
rs763747625 Chr.11: 123642733-123642733 on Build GRCh38
rs761964055 Chr.11: 123642478-123642478 on Build GRCh38
rs753656773 Chr.11: 123642596-123642596 on Build GRCh38
rs764015014 Chr.11: 123642585-123642585 on Build GRCh38
rs899269347 Chr.11: 123642651-123642651 on Build GRCh38
rs767416466 Chr.11: 123642566-123642566 on Build GRCh38
rs374062139 Chr.11: 123642759-123642759 on Build GRCh38
rs148484744 Chr.11: 123642501-123642501 on Build GRCh38
rs770348120 Chr.11: 123642425-123642425 on Build GRCh38
rs587777556 Chr.11: 123642502-123642502 on Build GRCh38
rs777020833 Chr.11: 123642645-123642645 on Build GRCh38
rs569206517 Chr.11: 123642417-123642417 on Build GRCh38
rs117419983 Chr.11: 123642412-123642412 on Build GRCh38
rs35174956 Chr.11: 123642601-123642601 on Build GRCh38
rs199974089 Chr.11: 123642642-123642642 on Build GRCh38
rs368054375 Chr.11: 123642476-123642476 on Build GRCh38
rs1031706489 Chr.11: 123642746-123642746 on Build GRCh38
rs142613556 Chr.11: 123642552-123642552 on Build GRCh38
rs989631957 Chr.11: 123642773-123642773 on Build GRCh38
rs201894537 Chr.11: 123642520-123642520 on Build GRCh38
rs776930951 Chr.11: 123642529-123642529 on Build GRCh38
rs750476444 Chr.11: 123642481-123642481 on Build GRCh38
rs1014663608 Chr.11: 123642641-123642641 on Build GRCh38
rs753313403 Chr.11: 123642778-123642778 on Build GRCh38
rs554843195 Chr.11: 123642780-123642780 on Build GRCh38
rs980121043 Chr.11: 123642390-123642390 on Build GRCh38
rs759023978 Chr.11: 123642823-123642823 on Build GRCh38
rs747919576 Chr.11: 123642416-123642416 on Build GRCh38
rs764587765 Chr.11: 123642659-123642659 on Build GRCh38
rs759996960 Chr.11: 123642438-123642438 on Build GRCh38
rs748523362 Chr.11: 123642657-123642657 on Build GRCh38
rs763016117 Chr.11: 123642666-123642666 on Build GRCh38
rs769024174 Chr.11: 123642527-123642527 on Build GRCh38
rs1275085 Chr.11: 123642453-123642453 on Build GRCh38
rs765254611 Chr.11: 123642480-123642480 on Build GRCh38
rs772513589 Chr.11: 123642513-123642513 on Build GRCh38
rs773474783 Chr.11: 123642665-123642665 on Build GRCh38
rs371558196 Chr.11: 123642497-123642497 on Build GRCh38
rs201799741 Chr.11: 123642633-123642633 on Build GRCh38
rs1037554685 Chr.11: 123642813-123642813 on Build GRCh38
rs996152549 Chr.11: 123642443-123642443 on Build GRCh38
rs926963558 Chr.11: 123642796-123642796 on Build GRCh38
rs760736732 Chr.11: 123642722-123642722 on Build GRCh38
rs2280582 Chr.11: 123642803-123642803 on Build GRCh38
rs956964922 Chr.11: 123642760-123642760 on Build GRCh38
rs756913203 Chr.11: 123642619-123642619 on Build GRCh38
rs72552180 Chr.11: 123642822-123642823 on Build GRCh38
rs541943652 Chr.11: 123642779-123642779 on Build GRCh38
rs964185029 Chr.11: 123642788-123642788 on Build GRCh38
rs773866969 Chr.11: 123642434-123642434 on Build GRCh38
rs72552167 Chr.11: 123642804-123642804 on Build GRCh38
rs760254403 Chr.11: 123642692-123642692 on Build GRCh38
rs574416973 Chr.11: 123642763-123642763 on Build GRCh38
rs149273271 Chr.11: 123642410-123642410 on Build GRCh38
rs754065169 Chr.11: 123642477-123642477 on Build GRCh38
rs951613326 Chr.11: 123642723-123642723 on Build GRCh38
rs960288438 Chr.11: 123642430-123642430 on Build GRCh38
rs781163714 Chr.11: 123642494-123642494 on Build GRCh38
rs72552143 Chr.11: 123642387-123642387 on Build GRCh38
rs974544911 Chr.11: 123642790-123642790 on Build GRCh38
rs775194985 Chr.11: 123642582-123642582 on Build GRCh38

View Full Product Details