Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125063271-125063763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC37A2
Entrez Gene ID
219855
Gene Name
solute carrier family 37 member 2
Gene Aliases
pp11662
UniGene
Hs.352661
Gene Chromosome Location
Chr.11: 125063067-125090516 on Build GRCh38
Transcripts
NM_001145290.1 NM_198277.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3791301 c.33C>T p.F11F Chr.11: 125063400-125063400 on Build GRCh38
6781908 c.31T>C p.F11L Chr.11: 125063398-125063398 on Build GRCh38
1243615 c.43T>G p.F15V Chr.11: 125063410-125063410 on Build GRCh38
305421 c.59G>A p.W20* Chr.11: 125063426-125063426 on Build GRCh38
6781907 c.31T>C p.F11L Chr.11: 125063398-125063398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749304984 Chr.11: 125063330-125063330 on Build GRCh38
rs760306626 Chr.11: 125063477-125063477 on Build GRCh38
rs777548440 Chr.11: 125063329-125063329 on Build GRCh38
rs768893099 Chr.11: 125063453-125063453 on Build GRCh38
rs899989153 Chr.11: 125063622-125063622 on Build GRCh38
rs926578334 Chr.11: 125063738-125063738 on Build GRCh38
rs950592862 Chr.11: 125063650-125063650 on Build GRCh38
rs1004242118 Chr.11: 125063653-125063653 on Build GRCh38
rs200478457 Chr.11: 125063357-125063357 on Build GRCh38
rs1014760002 Chr.11: 125063546-125063546 on Build GRCh38
rs767126437 Chr.11: 125063397-125063397 on Build GRCh38
rs747096242 Chr.11: 125063338-125063338 on Build GRCh38
rs11820425 Chr.11: 125063686-125063686 on Build GRCh38
rs763453343 Chr.11: 125063378-125063378 on Build GRCh38
rs538309169 Chr.11: 125063449-125063449 on Build GRCh38
rs1051359305 Chr.11: 125063501-125063501 on Build GRCh38
rs540820206 Chr.11: 125063551-125063551 on Build GRCh38
rs386758459 Chr.11: 125063632-125063633 on Build GRCh38
rs1044662803 Chr.11: 125063604-125063604 on Build GRCh38
rs780847014 Chr.11: 125063318-125063318 on Build GRCh38
rs377630439 Chr.11: 125063387-125063387 on Build GRCh38
rs758227051 Chr.11: 125063510-125063510 on Build GRCh38
rs769917530 Chr.11: 125063457-125063457 on Build GRCh38
rs759026427 Chr.11: 125063471-125063471 on Build GRCh38
rs372324419 Chr.11: 125063458-125063458 on Build GRCh38
rs767071319 Chr.11: 125063475-125063475 on Build GRCh38
rs905690086 Chr.11: 125063614-125063614 on Build GRCh38
rs775119465 Chr.11: 125063476-125063476 on Build GRCh38
rs932930519 Chr.11: 125063490-125063490 on Build GRCh38
rs73625329 Chr.11: 125063304-125063304 on Build GRCh38
rs774878582 Chr.11: 125063470-125063470 on Build GRCh38
rs1022785811 Chr.11: 125063300-125063300 on Build GRCh38
rs376420600 Chr.11: 125063375-125063375 on Build GRCh38
rs530446789 Chr.11: 125063734-125063734 on Build GRCh38
rs565950118 Chr.11: 125063629-125063629 on Build GRCh38
rs201925893 Chr.11: 125063407-125063407 on Build GRCh38
rs766812330 Chr.11: 125063380-125063380 on Build GRCh38
rs1017085646 Chr.11: 125063388-125063388 on Build GRCh38
rs770254575 Chr.11: 125063372-125063372 on Build GRCh38
rs772038350 Chr.11: 125063335-125063335 on Build GRCh38
rs887120533 Chr.11: 125063564-125063564 on Build GRCh38
rs780112789 Chr.11: 125063336-125063336 on Build GRCh38
rs201307672 Chr.11: 125063328-125063330 on Build GRCh38
rs746445945 Chr.11: 125063516-125063516 on Build GRCh38
rs532443355 Chr.11: 125063461-125063461 on Build GRCh38
rs575044682 Chr.11: 125063511-125063511 on Build GRCh38
rs913566400 Chr.11: 125063469-125063469 on Build GRCh38
rs768948240 Chr.11: 125063348-125063348 on Build GRCh38
rs921310024 Chr.11: 125063733-125063733 on Build GRCh38
rs373725918 Chr.11: 125063327-125063327 on Build GRCh38
rs753577596 Chr.11: 125063421-125063421 on Build GRCh38
rs763252639 Chr.11: 125063460-125063460 on Build GRCh38
rs759079909 Chr.11: 125063390-125063390 on Build GRCh38
rs12791554 Chr.11: 125063455-125063455 on Build GRCh38
rs1047026966 Chr.11: 125063560-125063560 on Build GRCh38
rs778650450 Chr.11: 125063417-125063417 on Build GRCh38
rs1056276834 Chr.11: 125063522-125063522 on Build GRCh38
rs1015591392 Chr.11: 125063654-125063654 on Build GRCh38
rs1003087719 Chr.11: 125063631-125063631 on Build GRCh38
rs1011307843 Chr.11: 125063554-125063554 on Build GRCh38
rs371403159 Chr.11: 125063404-125063404 on Build GRCh38
rs577589406 Chr.11: 125063633-125063633 on Build GRCh38
rs777020080 Chr.11: 125063356-125063356 on Build GRCh38
rs771277756 Chr.11: 125063471-125063472 on Build GRCh38
rs568870835 Chr.11: 125063737-125063737 on Build GRCh38
rs1005784134 Chr.11: 125063565-125063565 on Build GRCh38
rs1052691529 Chr.11: 125063547-125063547 on Build GRCh38
rs752449606 Chr.11: 125063326-125063326 on Build GRCh38
rs201411153 Chr.11: 125063426-125063426 on Build GRCh38
rs866810416 Chr.11: 125063574-125063574 on Build GRCh38
rs560711253 Chr.11: 125063552-125063552 on Build GRCh38
rs1005815401 Chr.11: 125063350-125063350 on Build GRCh38
rs955116566 Chr.11: 125063669-125063669 on Build GRCh38
rs139904225 Chr.11: 125063725-125063725 on Build GRCh38
rs778735087 Chr.11: 125063427-125063427 on Build GRCh38
rs951572370 Chr.11: 125063340-125063340 on Build GRCh38
rs895941674 Chr.11: 125063534-125063534 on Build GRCh38
rs777690109 Chr.11: 125063408-125063408 on Build GRCh38
rs558890234 Chr.11: 125063412-125063412 on Build GRCh38

View Full Product Details