Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 126406075-126406572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST3GAL4
Entrez Gene ID
6484
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 4
Gene Aliases
CGS23, NANTA3, SAT3, SIAT4, SIAT4C, ST3GalIV, STZ
UniGene
Hs.591947
Gene Chromosome Location
Chr.11: 126355645-126459601 on Build GRCh38
Transcripts
NM_001254757.1 NM_001254758.1 NM_001254759.1 NM_006278.2 XM_011542959.1 XM_011542960.2 XM_017018134.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1352436 c.45G>T p.L15L Chr.11: 126406501-126406501 on Build GRCh38
376219 c.51G>T p.L17L Chr.11: 126406507-126406507 on Build GRCh38
924694 c.89+3A>G p.? Chr.11: 126406548-126406548 on Build GRCh38
389019 c.79C>T p.R27W Chr.11: 126406535-126406535 on Build GRCh38
5826917 c.45G>T p.L15L Chr.11: 126406501-126406501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042352973 Chr.11: 126406238-126406238 on Build GRCh38
rs553463181 Chr.11: 126406396-126406396 on Build GRCh38
rs374434343 Chr.11: 126406327-126406327 on Build GRCh38
rs571511242 Chr.11: 126406303-126406303 on Build GRCh38
rs575877902 Chr.11: 126406435-126406435 on Build GRCh38
rs767454453 Chr.11: 126406154-126406154 on Build GRCh38
rs752532606 Chr.11: 126406162-126406162 on Build GRCh38
rs781738277 Chr.11: 126406196-126406196 on Build GRCh38
rs780341196 Chr.11: 126406185-126406185 on Build GRCh38
rs371604660 Chr.11: 126406223-126406223 on Build GRCh38
rs200880433 Chr.11: 126406511-126406511 on Build GRCh38
rs763213129 Chr.11: 126406446-126406446 on Build GRCh38
rs1037205499 Chr.11: 126406296-126406296 on Build GRCh38
rs201871612 Chr.11: 126406510-126406510 on Build GRCh38
rs773322033 Chr.11: 126406517-126406517 on Build GRCh38
rs376871531 Chr.11: 126406237-126406237 on Build GRCh38
rs543161044 Chr.11: 126406385-126406385 on Build GRCh38
rs924499536 Chr.11: 126406121-126406121 on Build GRCh38
rs955941382 Chr.11: 126406365-126406365 on Build GRCh38
rs199935524 Chr.11: 126406449-126406449 on Build GRCh38
rs959999316 Chr.11: 126406322-126406322 on Build GRCh38
rs528933775 Chr.11: 126406165-126406165 on Build GRCh38
rs187273010 Chr.11: 126406186-126406186 on Build GRCh38
rs763126899 Chr.11: 126406519-126406519 on Build GRCh38
rs758617896 Chr.11: 126406171-126406171 on Build GRCh38
rs998222818 Chr.11: 126406279-126406279 on Build GRCh38
rs146779997 Chr.11: 126406536-126406536 on Build GRCh38
rs573853432 Chr.11: 126406381-126406381 on Build GRCh38
rs766594250 Chr.11: 126406447-126406447 on Build GRCh38
rs555032192 Chr.11: 126406103-126406103 on Build GRCh38
rs1041012797 Chr.11: 126406147-126406147 on Build GRCh38
rs924489642 Chr.11: 126406349-126406349 on Build GRCh38
rs573277695 Chr.11: 126406403-126406403 on Build GRCh38
rs771711394 Chr.11: 126406421-126406421 on Build GRCh38
rs190884962 Chr.11: 126406111-126406111 on Build GRCh38
rs889745722 Chr.11: 126406299-126406299 on Build GRCh38
rs557768568 Chr.11: 126406272-126406272 on Build GRCh38
rs775422823 Chr.11: 126406422-126406422 on Build GRCh38
rs770302853 Chr.11: 126406498-126406498 on Build GRCh38
rs760543459 Chr.11: 126406431-126406431 on Build GRCh38
rs773011450 Chr.11: 126406102-126406102 on Build GRCh38
rs190622122 Chr.11: 126406200-126406200 on Build GRCh38
rs750993800 Chr.11: 126406489-126406489 on Build GRCh38
rs201136801 Chr.11: 126406190-126406190 on Build GRCh38
rs1025891415 Chr.11: 126406336-126406336 on Build GRCh38
rs976658973 Chr.11: 126406397-126406397 on Build GRCh38
rs769389712 Chr.11: 126406101-126406101 on Build GRCh38
rs902430751 Chr.11: 126406264-126406264 on Build GRCh38
rs1012603948 Chr.11: 126406324-126406324 on Build GRCh38
rs748606910 Chr.11: 126406197-126406197 on Build GRCh38
rs148736743 Chr.11: 126406499-126406499 on Build GRCh38
rs747379391 Chr.11: 126406187-126406187 on Build GRCh38
rs372066765 Chr.11: 126406418-126406418 on Build GRCh38
rs981624352 Chr.11: 126406376-126406376 on Build GRCh38
rs756724395 Chr.11: 126406540-126406540 on Build GRCh38
rs1035350072 Chr.11: 126406320-126406320 on Build GRCh38
rs914960722 Chr.11: 126406442-126406442 on Build GRCh38
rs781343777 Chr.11: 126406467-126406467 on Build GRCh38
rs367862665 Chr.11: 126406417-126406417 on Build GRCh38
rs927652088 Chr.11: 126406383-126406383 on Build GRCh38
rs557098937 Chr.11: 126406367-126406367 on Build GRCh38
rs753088020 Chr.11: 126406538-126406538 on Build GRCh38
rs750492435 Chr.11: 126406445-126406445 on Build GRCh38
rs767301667 Chr.11: 126406502-126406502 on Build GRCh38
rs766342195 Chr.11: 126406108-126406108 on Build GRCh38
rs772842973 Chr.11: 126406222-126406222 on Build GRCh38
rs978528171 Chr.11: 126406341-126406341 on Build GRCh38
rs1003831699 Chr.11: 126406314-126406314 on Build GRCh38
rs1005689152 Chr.11: 126406269-126406269 on Build GRCh38
rs545381257 Chr.11: 126406404-126406404 on Build GRCh38
rs778451724 Chr.11: 126406547-126406547 on Build GRCh38
rs968448811 Chr.11: 126406330-126406330 on Build GRCh38
rs144340959 Chr.11: 126406491-126406491 on Build GRCh38
rs200598125 Chr.11: 126406473-126406473 on Build GRCh38
rs766323001 Chr.11: 126406529-126406529 on Build GRCh38
rs755393088 Chr.11: 126406456-126406456 on Build GRCh38
rs768560304 Chr.11: 126406515-126406515 on Build GRCh38
rs184073997 Chr.11: 126406112-126406112 on Build GRCh38
rs756673733 Chr.11: 126406490-126406490 on Build GRCh38
rs1042238173 Chr.11: 126406479-126406479 on Build GRCh38
rs150025921 Chr.11: 126406163-126406163 on Build GRCh38
rs367840776 Chr.11: 126406201-126406201 on Build GRCh38
rs748774231 Chr.11: 126406493-126406493 on Build GRCh38
rs11559148 Chr.11: 126406535-126406535 on Build GRCh38
rs934584191 Chr.11: 126406134-126406134 on Build GRCh38

View Full Product Details