Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 132419503-132419998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPCML
Entrez Gene ID
4978
Gene Name
opioid binding protein/cell adhesion molecule like
Gene Aliases
IGLON1, OBCAM, OPCM
UniGene
Hs.4817
Gene Chromosome Location
Chr.11: 132403361-133532983 on Build GRCh38
Transcripts
NM_001012393.2 NM_001319103.1 NM_001319104.1 NM_001319105.1 NM_001319106.1 NM_002545.4 XM_006718846.2 XM_011542856.2

SNP Detail

SNP ID Chromosome Location
rs556968110 Chr.11: 132419843-132419843 on Build GRCh38
rs761546946 Chr.11: 132419914-132419914 on Build GRCh38
rs148936914 Chr.11: 132419912-132419912 on Build GRCh38
rs1003992913 Chr.11: 132419536-132419536 on Build GRCh38
rs946871019 Chr.11: 132419890-132419890 on Build GRCh38
rs1033654345 Chr.11: 132419585-132419585 on Build GRCh38
rs966746227 Chr.11: 132419956-132419956 on Build GRCh38
rs1011197519 Chr.11: 132419544-132419544 on Build GRCh38
rs913972890 Chr.11: 132419619-132419619 on Build GRCh38
rs962618655 Chr.11: 132419575-132419575 on Build GRCh38
rs1050748773 Chr.11: 132419763-132419763 on Build GRCh38
rs933635276 Chr.11: 132419760-132419760 on Build GRCh38
rs556480315 Chr.11: 132419670-132419670 on Build GRCh38
rs771884523 Chr.11: 132419931-132419931 on Build GRCh38
rs185002423 Chr.11: 132419813-132419813 on Build GRCh38
rs960684293 Chr.11: 132419908-132419908 on Build GRCh38
rs1002103570 Chr.11: 132419855-132419855 on Build GRCh38
rs1016762443 Chr.11: 132419554-132419554 on Build GRCh38
rs1040245230 Chr.11: 132419893-132419893 on Build GRCh38
rs561454369 Chr.11: 132419833-132419833 on Build GRCh38
rs992181786 Chr.11: 132419604-132419604 on Build GRCh38
rs747554068 Chr.11: 132419692-132419692 on Build GRCh38
rs973331333 Chr.11: 132419719-132419719 on Build GRCh38
rs145703455 Chr.11: 132419854-132419854 on Build GRCh38
rs537854856 Chr.11: 132419614-132419614 on Build GRCh38
rs140817622 Chr.11: 132419706-132419706 on Build GRCh38
rs759441010 Chr.11: 132419632-132419632 on Build GRCh38
rs1039976870 Chr.11: 132419550-132419550 on Build GRCh38
rs3360 Chr.11: 132419867-132419867 on Build GRCh38
rs982147214 Chr.11: 132419798-132419798 on Build GRCh38
rs577947992 Chr.11: 132419702-132419702 on Build GRCh38
rs144768353 Chr.11: 132419794-132419794 on Build GRCh38
rs760616968 Chr.11: 132419932-132419932 on Build GRCh38
rs961007560 Chr.11: 132419602-132419602 on Build GRCh38
rs767332074 Chr.11: 132419658-132419658 on Build GRCh38
rs757436134 Chr.11: 132419682-132419682 on Build GRCh38
rs567734305 Chr.11: 132419591-132419591 on Build GRCh38
rs1002538176 Chr.11: 132419584-132419584 on Build GRCh38
rs745994250 Chr.11: 132419713-132419713 on Build GRCh38
rs966820263 Chr.11: 132419636-132419636 on Build GRCh38
rs916503389 Chr.11: 132419756-132419756 on Build GRCh38
rs886883790 Chr.11: 132419531-132419531 on Build GRCh38
rs1051464691 Chr.11: 132419945-132419945 on Build GRCh38
rs532767514 Chr.11: 132419946-132419946 on Build GRCh38
rs752501875 Chr.11: 132419673-132419673 on Build GRCh38
rs375410063 Chr.11: 132419799-132419799 on Build GRCh38

View Full Product Details