Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 550830-551339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT3
Entrez Gene ID
283358
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 3
Gene Aliases
-
UniGene
Hs.504416
Gene Chromosome Location
Chr.12: 460377-561892 on Build GRCh38
Transcripts
NM_173593.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4639375 c.1088G>A p.R363H Chr.12: 551012-551012 on Build GRCh38
127413 c.1017G>A p.S339S Chr.12: 550941-550941 on Build GRCh38
3987155 c.1087C>T p.R363C Chr.12: 551011-551011 on Build GRCh38
6346392 c.1035G>T p.L345L Chr.12: 550959-550959 on Build GRCh38
7090399 c.1081C>A p.L361M Chr.12: 551005-551005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770350846 Chr.12: 550960-550960 on Build GRCh38
rs776962080 Chr.12: 550898-550898 on Build GRCh38
rs975595377 Chr.12: 551236-551236 on Build GRCh38
rs763923882 Chr.12: 551011-551011 on Build GRCh38
rs144889879 Chr.12: 550948-550948 on Build GRCh38
rs747142418 Chr.12: 550952-550952 on Build GRCh38
rs756028898 Chr.12: 551010-551010 on Build GRCh38
rs769571599 Chr.12: 551289-551291 on Build GRCh38
rs751059231 Chr.12: 550909-550909 on Build GRCh38
rs934701661 Chr.12: 551095-551095 on Build GRCh38
rs35944123 Chr.12: 551121-551122 on Build GRCh38
rs1003643446 Chr.12: 551148-551148 on Build GRCh38
rs766025363 Chr.12: 550985-550985 on Build GRCh38
rs906332248 Chr.12: 551143-551143 on Build GRCh38
rs145875846 Chr.12: 550953-550953 on Build GRCh38
rs765639281 Chr.12: 550901-550901 on Build GRCh38
rs71447496 Chr.12: 550906-550906 on Build GRCh38
rs548753456 Chr.12: 551303-551303 on Build GRCh38
rs369763840 Chr.12: 550967-550967 on Build GRCh38
rs984710418 Chr.12: 551294-551294 on Build GRCh38
rs1023003229 Chr.12: 551197-551197 on Build GRCh38
rs763888330 Chr.12: 551287-551287 on Build GRCh38
rs966078040 Chr.12: 551225-551225 on Build GRCh38
rs922780071 Chr.12: 551256-551256 on Build GRCh38
rs750105211 Chr.12: 550902-550902 on Build GRCh38
rs915736024 Chr.12: 551296-551296 on Build GRCh38
rs557288943 Chr.12: 551155-551155 on Build GRCh38
rs747142632 Chr.12: 551028-551028 on Build GRCh38
rs143781587 Chr.12: 551009-551009 on Build GRCh38
rs373263690 Chr.12: 551043-551043 on Build GRCh38
rs771456159 Chr.12: 551019-551019 on Build GRCh38
rs12423166 Chr.12: 550899-550899 on Build GRCh38
rs933072018 Chr.12: 550974-550974 on Build GRCh38
rs987207095 Chr.12: 551295-551295 on Build GRCh38
rs1010894242 Chr.12: 551195-551195 on Build GRCh38
rs762664031 Chr.12: 551235-551235 on Build GRCh38
rs773753652 Chr.12: 551082-551082 on Build GRCh38
rs764604850 Chr.12: 551154-551154 on Build GRCh38
rs368156322 Chr.12: 550942-550942 on Build GRCh38
rs759112894 Chr.12: 550990-550990 on Build GRCh38
rs554743820 Chr.12: 551073-551073 on Build GRCh38
rs748723319 Chr.12: 551069-551069 on Build GRCh38
rs199925734 Chr.12: 550922-550922 on Build GRCh38
rs201331783 Chr.12: 550878-550878 on Build GRCh38
rs781417739 Chr.12: 551056-551056 on Build GRCh38
rs762343462 Chr.12: 550959-550959 on Build GRCh38
rs754914848 Chr.12: 550917-550917 on Build GRCh38
rs1026614042 Chr.12: 551279-551279 on Build GRCh38
rs561094157 Chr.12: 551204-551204 on Build GRCh38
rs777208563 Chr.12: 550954-550954 on Build GRCh38
rs773852783 Chr.12: 550894-550899 on Build GRCh38
rs561592718 Chr.12: 551207-551207 on Build GRCh38
rs761132813 Chr.12: 550882-550882 on Build GRCh38
rs267603543 Chr.12: 550969-550969 on Build GRCh38
rs753595445 Chr.12: 551012-551012 on Build GRCh38
rs775097734 Chr.12: 551224-551224 on Build GRCh38
rs893402295 Chr.12: 551111-551111 on Build GRCh38
rs528993208 Chr.12: 551277-551277 on Build GRCh38
rs747419605 Chr.12: 550993-550993 on Build GRCh38
rs899076245 Chr.12: 551124-551124 on Build GRCh38
rs755088583 Chr.12: 551032-551032 on Build GRCh38
rs762745746 Chr.12: 550977-550977 on Build GRCh38
rs200885614 Chr.12: 550894-550894 on Build GRCh38
rs373776692 Chr.12: 550898-550900 on Build GRCh38
rs756960351 Chr.12: 551023-551023 on Build GRCh38
rs570749950 Chr.12: 550879-550879 on Build GRCh38
rs779690550 Chr.12: 551077-551077 on Build GRCh38
rs1023761427 Chr.12: 550893-550893 on Build GRCh38
rs767116275 Chr.12: 551019-551020 on Build GRCh38
rs879083731 Chr.12: 550940-550940 on Build GRCh38
rs575139341 Chr.12: 551027-551027 on Build GRCh38
rs141135800 Chr.12: 550934-550934 on Build GRCh38
rs148974990 Chr.12: 550987-550987 on Build GRCh38
rs951965463 Chr.12: 551286-551286 on Build GRCh38
rs755920743 Chr.12: 550941-550941 on Build GRCh38
rs762690539 Chr.12: 550903-550903 on Build GRCh38
rs928613028 Chr.12: 551039-551039 on Build GRCh38
rs1012534361 Chr.12: 551127-551127 on Build GRCh38
rs191679082 Chr.12: 550975-550975 on Build GRCh38
rs907558345 Chr.12: 551103-551103 on Build GRCh38
rs140849879 Chr.12: 551140-551140 on Build GRCh38
rs765865272 Chr.12: 550904-550904 on Build GRCh38
rs572954261 Chr.12: 551109-551109 on Build GRCh38
rs186841043 Chr.12: 550949-550949 on Build GRCh38

View Full Product Details