Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6914351-6914855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO2
Entrez Gene ID
2026
Gene Name
enolase 2
Gene Aliases
HEL-S-279, NSE
UniGene
Hs.511915
Gene Chromosome Location
Chr.12: 6914450-6923696 on Build GRCh38
Transcripts
NM_001975.2

SNP Detail

SNP ID Chromosome Location
rs915826586 Chr.12: 6914706-6914706 on Build GRCh38
rs968470690 Chr.12: 6914501-6914501 on Build GRCh38
rs998402480 Chr.12: 6914392-6914392 on Build GRCh38
rs992739510 Chr.12: 6914519-6914519 on Build GRCh38
rs782233473 Chr.12: 6914634-6914634 on Build GRCh38
rs1028570059 Chr.12: 6914397-6914397 on Build GRCh38
rs781963007 Chr.12: 6914783-6914783 on Build GRCh38
rs2239165 Chr.12: 6914702-6914702 on Build GRCh38
rs34551558 Chr.12: 6914640-6914641 on Build GRCh38
rs998527644 Chr.12: 6914676-6914676 on Build GRCh38
rs915811435 Chr.12: 6914529-6914529 on Build GRCh38
rs909975787 Chr.12: 6914579-6914579 on Build GRCh38
rs901207373 Chr.12: 6914611-6914611 on Build GRCh38
rs890054757 Chr.12: 6914812-6914812 on Build GRCh38
rs954394495 Chr.12: 6914459-6914459 on Build GRCh38
rs375750776 Chr.12: 6914405-6914405 on Build GRCh38
rs1048197805 Chr.12: 6914575-6914575 on Build GRCh38
rs1037040742 Chr.12: 6914594-6914594 on Build GRCh38
rs1022440269 Chr.12: 6914656-6914656 on Build GRCh38
rs1031375929 Chr.12: 6914684-6914684 on Build GRCh38
rs938609375 Chr.12: 6914516-6914516 on Build GRCh38
rs187073633 Chr.12: 6914621-6914621 on Build GRCh38
rs782526261 Chr.12: 6914607-6914608 on Build GRCh38
rs959738870 Chr.12: 6914692-6914692 on Build GRCh38
rs922661065 Chr.12: 6914790-6914790 on Build GRCh38
rs1011119442 Chr.12: 6914814-6914814 on Build GRCh38
rs760975884 Chr.12: 6914811-6914811 on Build GRCh38
rs782253628 Chr.12: 6914671-6914671 on Build GRCh38
rs1011507631 Chr.12: 6914642-6914642 on Build GRCh38
rs1021824731 Chr.12: 6914484-6914484 on Build GRCh38
rs989647544 Chr.12: 6914477-6914477 on Build GRCh38
rs1037155465 Chr.12: 6914778-6914778 on Build GRCh38
rs984363882 Chr.12: 6914742-6914742 on Build GRCh38
rs909680341 Chr.12: 6914760-6914760 on Build GRCh38
rs781849081 Chr.12: 6914521-6914521 on Build GRCh38
rs782332633 Chr.12: 6914746-6914746 on Build GRCh38
rs782683985 Chr.12: 6914750-6914750 on Build GRCh38
rs977075962 Chr.12: 6914502-6914502 on Build GRCh38
rs782774097 Chr.12: 6914802-6914802 on Build GRCh38
rs139729747 Chr.12: 6914657-6914657 on Build GRCh38
rs969976841 Chr.12: 6914714-6914714 on Build GRCh38
rs782022738 Chr.12: 6914732-6914732 on Build GRCh38
rs939825847 Chr.12: 6914765-6914765 on Build GRCh38
rs2239164 Chr.12: 6914640-6914640 on Build GRCh38
rs782337592 Chr.12: 6914791-6914791 on Build GRCh38
rs1050029617 Chr.12: 6914639-6914639 on Build GRCh38
rs948587350 Chr.12: 6914557-6914557 on Build GRCh38

View Full Product Details