Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10160699-10161202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLR1
Entrez Gene ID
4973
Gene Name
oxidized low density lipoprotein receptor 1
Gene Aliases
CLEC8A, LOX1, LOXIN, SCARE1, SLOX1
UniGene
Hs.412484
Gene Chromosome Location
Chr.12: 10158300-10172191 on Build GRCh38
Transcripts
NM_001172632.1 NM_001172633.1 NM_002543.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1218443 c.443G>A p.W148* Chr.12: 10160907-10160907 on Build GRCh38
6135545 c.508G>T p.E170* Chr.12: 10160842-10160842 on Build GRCh38
5456254 c.564+10C>T p.? Chr.12: 10160776-10160776 on Build GRCh38
3810807 c.434C>A p.P145Q Chr.12: 10160916-10160916 on Build GRCh38
3455192 c.540G>A p.L180L Chr.12: 10160810-10160810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768691963 Chr.12: 10160871-10160871 on Build GRCh38
rs950622824 Chr.12: 10161148-10161148 on Build GRCh38
rs778334737 Chr.12: 10161036-10161037 on Build GRCh38
rs386760246 Chr.12: 10160757-10160759 on Build GRCh38
rs767441031 Chr.12: 10161068-10161068 on Build GRCh38
rs974247060 Chr.12: 10161031-10161042 on Build GRCh38
rs35586937 Chr.12: 10161073-10161073 on Build GRCh38
rs779696999 Chr.12: 10160966-10160966 on Build GRCh38
rs540589720 Chr.12: 10161146-10161146 on Build GRCh38
rs1053402561 Chr.12: 10161021-10161021 on Build GRCh38
rs752811946 Chr.12: 10161093-10161093 on Build GRCh38
rs767044631 Chr.12: 10160959-10160959 on Build GRCh38
rs1015241355 Chr.12: 10161017-10161017 on Build GRCh38
rs569613279 Chr.12: 10161120-10161120 on Build GRCh38
rs573578085 Chr.12: 10160955-10160955 on Build GRCh38
rs755861636 Chr.12: 10160852-10160852 on Build GRCh38
rs761012790 Chr.12: 10160903-10160903 on Build GRCh38
rs11053647 Chr.12: 10161123-10161123 on Build GRCh38
rs749276327 Chr.12: 10160748-10160748 on Build GRCh38
rs778112459 Chr.12: 10161130-10161130 on Build GRCh38
rs960648542 Chr.12: 10161014-10161014 on Build GRCh38
rs754260673 Chr.12: 10160822-10160822 on Build GRCh38
rs751972509 Chr.12: 10160995-10160995 on Build GRCh38
rs926480697 Chr.12: 10160774-10160774 on Build GRCh38
rs759947861 Chr.12: 10160916-10160916 on Build GRCh38
rs1050444516 Chr.12: 10160831-10160831 on Build GRCh38
rs11053646 Chr.12: 10160849-10160849 on Build GRCh38
rs778851250 Chr.12: 10160875-10160875 on Build GRCh38
rs780871480 Chr.12: 10160846-10160846 on Build GRCh38
rs183144177 Chr.12: 10161010-10161010 on Build GRCh38
rs577879012 Chr.12: 10161132-10161132 on Build GRCh38
rs201079254 Chr.12: 10160947-10160947 on Build GRCh38
rs773520812 Chr.12: 10160887-10160887 on Build GRCh38
rs749130285 Chr.12: 10160978-10160978 on Build GRCh38
rs369614046 Chr.12: 10160778-10160778 on Build GRCh38
rs776358905 Chr.12: 10160754-10160754 on Build GRCh38
rs1021690240 Chr.12: 10160941-10160941 on Build GRCh38
rs536324555 Chr.12: 10160963-10160963 on Build GRCh38
rs765840296 Chr.12: 10160925-10160925 on Build GRCh38
rs34807991 Chr.12: 10160757-10160757 on Build GRCh38
rs761172569 Chr.12: 10160799-10160799 on Build GRCh38
rs1026204893 Chr.12: 10161145-10161145 on Build GRCh38
rs1033557097 Chr.12: 10160964-10160964 on Build GRCh38
rs1012100018 Chr.12: 10160938-10160938 on Build GRCh38
rs1001948462 Chr.12: 10161129-10161129 on Build GRCh38
rs766875651 Chr.12: 10160809-10160809 on Build GRCh38
rs34418327 Chr.12: 10161016-10161016 on Build GRCh38
rs372771543 Chr.12: 10160878-10160878 on Build GRCh38
rs564346133 Chr.12: 10161125-10161125 on Build GRCh38
rs748871918 Chr.12: 10161031-10161036 on Build GRCh38
rs1009164624 Chr.12: 10161083-10161083 on Build GRCh38
rs749120239 Chr.12: 10160870-10160870 on Build GRCh38
rs756870922 Chr.12: 10160839-10160839 on Build GRCh38
rs1039179771 Chr.12: 10160982-10160982 on Build GRCh38
rs764862595 Chr.12: 10160798-10160798 on Build GRCh38
rs768733049 Chr.12: 10161131-10161131 on Build GRCh38
rs773684434 Chr.12: 10160793-10160793 on Build GRCh38
rs201129982 Chr.12: 10160853-10160853 on Build GRCh38
rs763455847 Chr.12: 10160825-10160825 on Build GRCh38
rs761924125 Chr.12: 10160771-10160771 on Build GRCh38
rs751140676 Chr.12: 10160834-10160834 on Build GRCh38
rs1049112315 Chr.12: 10160898-10160898 on Build GRCh38
rs763279630 Chr.12: 10160915-10160915 on Build GRCh38
rs750008035 Chr.12: 10160961-10160961 on Build GRCh38
rs902050559 Chr.12: 10160991-10160991 on Build GRCh38
rs3736232 Chr.12: 10160759-10160759 on Build GRCh38
rs772406020 Chr.12: 10160885-10160885 on Build GRCh38

View Full Product Details