Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12118357-12118904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRP6
Entrez Gene ID
4040
Gene Name
LDL receptor related protein 6
Gene Aliases
ADCAD2, STHAG7
UniGene
Hs.584775
Gene Chromosome Location
Chr.12: 12116025-12267021 on Build GRCh38
Transcripts
NM_002336.2 XM_006719078.3 XM_011520671.2 XM_017019304.1

SNP Detail

SNP ID Chromosome Location
rs751465489 Chr.12: 12118845-12118845 on Build GRCh38
rs992734158 Chr.12: 12118551-12118551 on Build GRCh38
rs917206614 Chr.12: 12118792-12118792 on Build GRCh38
rs71457130 Chr.12: 12118640-12118640 on Build GRCh38
rs1055966674 Chr.12: 12118400-12118400 on Build GRCh38
rs962266881 Chr.12: 12118801-12118801 on Build GRCh38
rs2241181 Chr.12: 12118789-12118789 on Build GRCh38
rs1020899644 Chr.12: 12118831-12118831 on Build GRCh38
rs763936483 Chr.12: 12118842-12118842 on Build GRCh38
rs996518201 Chr.12: 12118802-12118802 on Build GRCh38
rs886751904 Chr.12: 12118490-12118490 on Build GRCh38
rs968799759 Chr.12: 12118731-12118731 on Build GRCh38
rs1014471254 Chr.12: 12118420-12118420 on Build GRCh38
rs543219143 Chr.12: 12118879-12118879 on Build GRCh38
rs930361235 Chr.12: 12118630-12118630 on Build GRCh38
rs946093070 Chr.12: 12118631-12118631 on Build GRCh38
rs1055977840 Chr.12: 12118706-12118706 on Build GRCh38
rs772034790 Chr.12: 12118484-12118484 on Build GRCh38
rs950365692 Chr.12: 12118727-12118727 on Build GRCh38
rs779799394 Chr.12: 12118574-12118574 on Build GRCh38
rs549185346 Chr.12: 12118566-12118566 on Build GRCh38
rs965551804 Chr.12: 12118412-12118412 on Build GRCh38
rs1003914020 Chr.12: 12118745-12118745 on Build GRCh38
rs940500460 Chr.12: 12118580-12118580 on Build GRCh38
rs987020984 Chr.12: 12118814-12118814 on Build GRCh38
rs772842977 Chr.12: 12118824-12118824 on Build GRCh38
rs962428267 Chr.12: 12118516-12118516 on Build GRCh38
rs140578950 Chr.12: 12118834-12118836 on Build GRCh38
rs1007572409 Chr.12: 12118654-12118654 on Build GRCh38
rs80081247 Chr.12: 12118761-12118761 on Build GRCh38
rs535183961 Chr.12: 12118700-12118700 on Build GRCh38
rs920413211 Chr.12: 12118809-12118809 on Build GRCh38
rs1020871404 Chr.12: 12118712-12118712 on Build GRCh38
rs901466466 Chr.12: 12118873-12118873 on Build GRCh38
rs368958971 Chr.12: 12118723-12118723 on Build GRCh38
rs968768291 Chr.12: 12118462-12118462 on Build GRCh38
rs761534286 Chr.12: 12118741-12118741 on Build GRCh38
rs989304906 Chr.12: 12118436-12118436 on Build GRCh38
rs869292776 Chr.12: 12118545-12118546 on Build GRCh38
rs926063560 Chr.12: 12118679-12118679 on Build GRCh38
rs746826805 Chr.12: 12118581-12118581 on Build GRCh38
rs1016552647 Chr.12: 12118762-12118762 on Build GRCh38
rs1036595769 Chr.12: 12118603-12118603 on Build GRCh38
rs1020336689 Chr.12: 12118455-12118455 on Build GRCh38
rs887343355 Chr.12: 12118744-12118744 on Build GRCh38
rs557874147 Chr.12: 12118514-12118514 on Build GRCh38
rs1004266067 Chr.12: 12118506-12118506 on Build GRCh38
rs1019826353 Chr.12: 12118698-12118698 on Build GRCh38
rs1040437695 Chr.12: 12118872-12118872 on Build GRCh38
rs553499367 Chr.12: 12118709-12118709 on Build GRCh38
rs760835178 Chr.12: 12118549-12118551 on Build GRCh38
rs1050149428 Chr.12: 12118832-12118832 on Build GRCh38
rs975255384 Chr.12: 12118518-12118518 on Build GRCh38
rs545780740 Chr.12: 12118481-12118481 on Build GRCh38
rs576308362 Chr.12: 12118844-12118844 on Build GRCh38
rs911899915 Chr.12: 12118596-12118596 on Build GRCh38
rs551777337 Chr.12: 12118406-12118406 on Build GRCh38
rs11054696 Chr.12: 12118805-12118805 on Build GRCh38
rs1050117028 Chr.12: 12118638-12118638 on Build GRCh38
rs898954288 Chr.12: 12118612-12118612 on Build GRCh38
rs768295879 Chr.12: 12118608-12118608 on Build GRCh38
rs955185674 Chr.12: 12118811-12118811 on Build GRCh38

View Full Product Details