Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12119542-12120085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRP6
Entrez Gene ID
4040
Gene Name
LDL receptor related protein 6
Gene Aliases
ADCAD2, STHAG7
UniGene
Hs.584775
Gene Chromosome Location
Chr.12: 12116025-12267021 on Build GRCh38
Transcripts
NM_002336.2 XM_006719078.3 XM_011520671.2 XM_017019304.1

SNP Detail

SNP ID Chromosome Location
rs561282852 Chr.12: 12119888-12119888 on Build GRCh38
rs545037698 Chr.12: 12119827-12119827 on Build GRCh38
rs559340355 Chr.12: 12119638-12119638 on Build GRCh38
rs981903798 Chr.12: 12119683-12119683 on Build GRCh38
rs943816698 Chr.12: 12119833-12119833 on Build GRCh38
rs184058212 Chr.12: 12119724-12119724 on Build GRCh38
rs569884328 Chr.12: 12119971-12119971 on Build GRCh38
rs550587604 Chr.12: 12120026-12120043 on Build GRCh38
rs1002856625 Chr.12: 12119923-12119923 on Build GRCh38
rs1022978907 Chr.12: 12119884-12119884 on Build GRCh38
rs35025692 Chr.12: 12119722-12119722 on Build GRCh38
rs868146560 Chr.12: 12120012-12120012 on Build GRCh38
rs369329930 Chr.12: 12120004-12120004 on Build GRCh38
rs760606023 Chr.12: 12119869-12119869 on Build GRCh38
rs902317513 Chr.12: 12119573-12119573 on Build GRCh38
rs191816977 Chr.12: 12119664-12119664 on Build GRCh38
rs528642777 Chr.12: 12119945-12119945 on Build GRCh38
rs1053613553 Chr.12: 12119861-12119861 on Build GRCh38
rs12827309 Chr.12: 12119988-12119988 on Build GRCh38
rs567315765 Chr.12: 12119987-12119988 on Build GRCh38
rs530641317 Chr.12: 12119832-12119832 on Build GRCh38
rs772413878 Chr.12: 12119778-12119778 on Build GRCh38
rs950229427 Chr.12: 12119877-12119877 on Build GRCh38
rs186428247 Chr.12: 12119846-12119846 on Build GRCh38
rs985096338 Chr.12: 12119812-12119812 on Build GRCh38
rs1032287611 Chr.12: 12119619-12119619 on Build GRCh38
rs1023348172 Chr.12: 12119645-12119645 on Build GRCh38
rs961867963 Chr.12: 12119699-12119699 on Build GRCh38
rs902245186 Chr.12: 12119855-12119855 on Build GRCh38
rs868322967 Chr.12: 12119924-12119924 on Build GRCh38
rs774429262 Chr.12: 12119991-12120028 on Build GRCh38
rs542907054 Chr.12: 12119876-12119876 on Build GRCh38
rs979088989 Chr.12: 12119805-12119805 on Build GRCh38
rs1007471365 Chr.12: 12119643-12119643 on Build GRCh38
rs936521747 Chr.12: 12119856-12119856 on Build GRCh38
rs1012959559 Chr.12: 12119639-12119639 on Build GRCh38
rs12827301 Chr.12: 12119980-12119980 on Build GRCh38
rs61195540 Chr.12: 12120043-12120044 on Build GRCh38
rs867378060 Chr.12: 12120042-12120042 on Build GRCh38
rs769555593 Chr.12: 12119756-12119756 on Build GRCh38
rs139152304 Chr.12: 12119698-12119698 on Build GRCh38
rs35043292 Chr.12: 12120033-12120034 on Build GRCh38
rs991677524 Chr.12: 12119839-12119839 on Build GRCh38
rs1042509095 Chr.12: 12119835-12119835 on Build GRCh38
rs371290168 Chr.12: 12120006-12120006 on Build GRCh38
rs868464277 Chr.12: 12120014-12120014 on Build GRCh38
rs932902515 Chr.12: 12119735-12119735 on Build GRCh38
rs773799808 Chr.12: 12119779-12119779 on Build GRCh38
rs762525885 Chr.12: 12119907-12119907 on Build GRCh38
rs1000681092 Chr.12: 12119580-12119580 on Build GRCh38
rs865837392 Chr.12: 12120010-12120010 on Build GRCh38
rs1045894085 Chr.12: 12119893-12119893 on Build GRCh38
rs867167380 Chr.12: 12120040-12120040 on Build GRCh38

View Full Product Details