Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12989101-12989606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM234B
Entrez Gene ID
57613
Gene Name
family with sequence similarity 234 member B
Gene Aliases
KIAA1467
UniGene
Hs.132660
Gene Chromosome Location
Chr.12: 12927680-13083449 on Build GRCh38
Transcripts
XM_017019706.1 NM_015987.4

Gene Symbol
HEBP1
Entrez Gene ID
50865
Gene Name
heme binding protein 1
Gene Aliases
HBP, HEBP
UniGene
Hs.642618
Gene Chromosome Location
Chr.12: 12974864-13000309 on Build GRCh38
Transcripts
XM_017019706.1 NM_015987.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6135877 c.203G>A p.G68D Chr.12: 12989291-12989291 on Build GRCh38
2228538 c.84A>G p.E28E Chr.12: 12989410-12989410 on Build GRCh38
347534 c.116G>A p.G39D Chr.12: 12989378-12989378 on Build GRCh38
2228534 c.192G>C p.K64N Chr.12: 12989302-12989302 on Build GRCh38
6678659 c.167G>T p.R56L Chr.12: 12989327-12989327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550185396 Chr.12: 12989495-12989495 on Build GRCh38
rs758646938 Chr.12: 12989265-12989265 on Build GRCh38
rs140357176 Chr.12: 12989385-12989385 on Build GRCh38
rs201900227 Chr.12: 12989379-12989379 on Build GRCh38
rs753922468 Chr.12: 12989244-12989244 on Build GRCh38
rs555766842 Chr.12: 12989168-12989168 on Build GRCh38
rs756587233 Chr.12: 12989335-12989335 on Build GRCh38
rs752801907 Chr.12: 12989311-12989311 on Build GRCh38
rs746576485 Chr.12: 12989363-12989363 on Build GRCh38
rs141002469 Chr.12: 12989296-12989296 on Build GRCh38
rs770790009 Chr.12: 12989365-12989365 on Build GRCh38
rs777107379 Chr.12: 12989412-12989412 on Build GRCh38
rs759351075 Chr.12: 12989307-12989307 on Build GRCh38
rs113772561 Chr.12: 12989574-12989574 on Build GRCh38
rs756678921 Chr.12: 12989402-12989402 on Build GRCh38
rs367955716 Chr.12: 12989386-12989386 on Build GRCh38
rs199829664 Chr.12: 12989380-12989380 on Build GRCh38
rs371773825 Chr.12: 12989433-12989433 on Build GRCh38
rs893654980 Chr.12: 12989492-12989492 on Build GRCh38
rs761905397 Chr.12: 12989389-12989389 on Build GRCh38
rs781347916 Chr.12: 12989328-12989328 on Build GRCh38
rs750717922 Chr.12: 12989401-12989401 on Build GRCh38
rs780522020 Chr.12: 12989404-12989404 on Build GRCh38
rs767318923 Chr.12: 12989227-12989227 on Build GRCh38
rs964905034 Chr.12: 12989343-12989343 on Build GRCh38
rs760204191 Chr.12: 12989462-12989463 on Build GRCh38
rs771432932 Chr.12: 12989274-12989274 on Build GRCh38
rs771679813 Chr.12: 12989294-12989294 on Build GRCh38
rs935401558 Chr.12: 12989439-12989439 on Build GRCh38
rs922715378 Chr.12: 12989434-12989434 on Build GRCh38
rs770572537 Chr.12: 12989297-12989297 on Build GRCh38
rs747842976 Chr.12: 12989293-12989293 on Build GRCh38
rs757659059 Chr.12: 12989327-12989327 on Build GRCh38
rs746306932 Chr.12: 12989283-12989283 on Build GRCh38
rs750958688 Chr.12: 12989329-12989329 on Build GRCh38
rs757906860 Chr.12: 12989406-12989406 on Build GRCh38
rs2232243 Chr.12: 12989375-12989375 on Build GRCh38
rs767970203 Chr.12: 12989392-12989392 on Build GRCh38
rs850918 Chr.12: 12989240-12989240 on Build GRCh38
rs1035210637 Chr.12: 12989306-12989306 on Build GRCh38
rs747649421 Chr.12: 12989351-12989351 on Build GRCh38
rs1045408589 Chr.12: 12989504-12989504 on Build GRCh38
rs969476157 Chr.12: 12989302-12989302 on Build GRCh38
rs182080474 Chr.12: 12989319-12989319 on Build GRCh38
rs774012252 Chr.12: 12989253-12989253 on Build GRCh38
rs772965491 Chr.12: 12989237-12989237 on Build GRCh38
rs757775150 Chr.12: 12989276-12989276 on Build GRCh38
rs778331263 Chr.12: 12989267-12989267 on Build GRCh38
rs376480527 Chr.12: 12989438-12989438 on Build GRCh38
rs532073911 Chr.12: 12989338-12989338 on Build GRCh38
rs977055984 Chr.12: 12989204-12989204 on Build GRCh38
rs768219771 Chr.12: 12989290-12989290 on Build GRCh38
rs183604156 Chr.12: 12989505-12989505 on Build GRCh38
rs533935823 Chr.12: 12989566-12989566 on Build GRCh38
rs778341967 Chr.12: 12989291-12989291 on Build GRCh38
rs781717766 Chr.12: 12989277-12989277 on Build GRCh38
rs763309173 Chr.12: 12989314-12989314 on Build GRCh38
rs776635117 Chr.12: 12989304-12989304 on Build GRCh38
rs774800633 Chr.12: 12989417-12989418 on Build GRCh38
rs1054296504 Chr.12: 12989245-12989245 on Build GRCh38
rs560452444 Chr.12: 12989243-12989243 on Build GRCh38
rs764407715 Chr.12: 12989315-12989315 on Build GRCh38
rs1010719628 Chr.12: 12989500-12989500 on Build GRCh38
rs753000071 Chr.12: 12989263-12989263 on Build GRCh38
rs990079248 Chr.12: 12989487-12989487 on Build GRCh38
rs756837762 Chr.12: 12989446-12989446 on Build GRCh38
rs981780646 Chr.12: 12989303-12989303 on Build GRCh38
rs866985250 Chr.12: 12989378-12989378 on Build GRCh38
rs935936342 Chr.12: 12989471-12989471 on Build GRCh38
rs763247049 Chr.12: 12989226-12989227 on Build GRCh38
rs751815832 Chr.12: 12989387-12989387 on Build GRCh38
rs754284050 Chr.12: 12989405-12989405 on Build GRCh38
rs529343020 Chr.12: 12989525-12989525 on Build GRCh38
rs781081357 Chr.12: 12989467-12989467 on Build GRCh38
rs764796571 Chr.12: 12989483-12989483 on Build GRCh38
rs189428822 Chr.12: 12989260-12989260 on Build GRCh38
rs372106634 Chr.12: 12989308-12989308 on Build GRCh38
rs148078457 Chr.12: 12989173-12989173 on Build GRCh38
rs777493570 Chr.12: 12989354-12989354 on Build GRCh38
rs778269562 Chr.12: 12989418-12989421 on Build GRCh38
rs771768178 Chr.12: 12989353-12989353 on Build GRCh38
rs1052481576 Chr.12: 12989477-12989477 on Build GRCh38

View Full Product Details