Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 15322556-15323064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRO
Entrez Gene ID
5800
Gene Name
protein tyrosine phosphatase, receptor type O
Gene Aliases
GLEPP1, NPHS6, PTP-OC, PTP-U2, PTPROT, PTPU2, R-PTP-O
UniGene
Hs.160871
Gene Chromosome Location
Chr.12: 15322257-15598331 on Build GRCh38
Transcripts
NM_002848.3 NM_030667.2 XM_017019725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1705225 c.19G>A p.G7R Chr.12: 15322745-15322745 on Build GRCh38
5659074 c.12G>T p.L4L Chr.12: 15322738-15322738 on Build GRCh38
5051486 c.27C>T p.H9H Chr.12: 15322753-15322753 on Build GRCh38
7157485 c.49C>T p.L17L Chr.12: 15322775-15322775 on Build GRCh38
4719284 c.4G>T p.G2W Chr.12: 15322730-15322730 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375272425 Chr.12: 15322686-15322686 on Build GRCh38
rs751954059 Chr.12: 15322693-15322693 on Build GRCh38
rs750982167 Chr.12: 15322754-15322754 on Build GRCh38
rs544374940 Chr.12: 15322585-15322585 on Build GRCh38
rs542092081 Chr.12: 15322740-15322740 on Build GRCh38
rs376929238 Chr.12: 15322834-15322834 on Build GRCh38
rs962638651 Chr.12: 15322905-15322905 on Build GRCh38
rs759448037 Chr.12: 15322680-15322680 on Build GRCh38
rs775401657 Chr.12: 15323004-15323004 on Build GRCh38
rs1032542202 Chr.12: 15323019-15323019 on Build GRCh38
rs1013052000 Chr.12: 15322654-15322654 on Build GRCh38
rs531564836 Chr.12: 15322799-15322799 on Build GRCh38
rs369405874 Chr.12: 15322716-15322716 on Build GRCh38
rs756776270 Chr.12: 15322786-15322786 on Build GRCh38
rs749433233 Chr.12: 15322810-15322810 on Build GRCh38
rs756519574 Chr.12: 15322713-15322713 on Build GRCh38
rs373358416 Chr.12: 15322794-15322794 on Build GRCh38
rs971288955 Chr.12: 15322887-15322887 on Build GRCh38
rs779916135 Chr.12: 15322788-15322789 on Build GRCh38
rs776601178 Chr.12: 15322677-15322677 on Build GRCh38
rs1024147493 Chr.12: 15322662-15322662 on Build GRCh38
rs997308395 Chr.12: 15322636-15322636 on Build GRCh38
rs957742336 Chr.12: 15323030-15323030 on Build GRCh38
rs1048317603 Chr.12: 15322883-15322883 on Build GRCh38
rs770122069 Chr.12: 15322681-15322681 on Build GRCh38
rs772201493 Chr.12: 15322828-15322828 on Build GRCh38
rs777347588 Chr.12: 15322720-15322720 on Build GRCh38
rs372854549 Chr.12: 15322719-15322719 on Build GRCh38
rs761836162 Chr.12: 15322748-15322748 on Build GRCh38
rs774519030 Chr.12: 15322747-15322747 on Build GRCh38
rs764240215 Chr.12: 15322687-15322687 on Build GRCh38
rs936262416 Chr.12: 15322990-15322990 on Build GRCh38
rs566898556 Chr.12: 15322873-15322873 on Build GRCh38
rs912067810 Chr.12: 15322800-15322800 on Build GRCh38
rs767985777 Chr.12: 15322751-15322751 on Build GRCh38
rs376103175 Chr.12: 15322730-15322730 on Build GRCh38
rs533937231 Chr.12: 15322910-15322910 on Build GRCh38
rs900393754 Chr.12: 15322633-15322633 on Build GRCh38
rs748241677 Chr.12: 15322823-15322823 on Build GRCh38
rs745646041 Chr.12: 15322732-15322732 on Build GRCh38
rs200496219 Chr.12: 15322787-15322787 on Build GRCh38
rs1015542301 Chr.12: 15322897-15322897 on Build GRCh38
rs775478150 Chr.12: 15322894-15322894 on Build GRCh38
rs751237661 Chr.12: 15322845-15322845 on Build GRCh38
rs761023590 Chr.12: 15322755-15322755 on Build GRCh38
rs973957512 Chr.12: 15322912-15322912 on Build GRCh38
rs199577382 Chr.12: 15322830-15322830 on Build GRCh38
rs942043819 Chr.12: 15322901-15322902 on Build GRCh38
rs774482419 Chr.12: 15322822-15322822 on Build GRCh38
rs767827661 Chr.12: 15322707-15322707 on Build GRCh38
rs745809506 Chr.12: 15322789-15322789 on Build GRCh38
rs568228715 Chr.12: 15322839-15322839 on Build GRCh38
rs763253930 Chr.12: 15322736-15322736 on Build GRCh38
rs570839503 Chr.12: 15322998-15322998 on Build GRCh38
rs769779448 Chr.12: 15322735-15322735 on Build GRCh38
rs1041852349 Chr.12: 15322975-15322975 on Build GRCh38
rs559035358 Chr.12: 15322958-15322958 on Build GRCh38
rs527253479 Chr.12: 15322761-15322761 on Build GRCh38
rs139015332 Chr.12: 15322757-15322757 on Build GRCh38
rs900446354 Chr.12: 15322946-15322946 on Build GRCh38
rs746870773 Chr.12: 15322725-15322725 on Build GRCh38
rs750753678 Chr.12: 15322709-15322709 on Build GRCh38
rs139244830 Chr.12: 15323037-15323037 on Build GRCh38
rs1054595541 Chr.12: 15323017-15323017 on Build GRCh38
rs752232017 Chr.12: 15322758-15322758 on Build GRCh38
rs770873561 Chr.12: 15322728-15322728 on Build GRCh38
rs920603127 Chr.12: 15322913-15322913 on Build GRCh38
rs999366852 Chr.12: 15323006-15323006 on Build GRCh38
rs547332760 Chr.12: 15322865-15322865 on Build GRCh38
rs773642552 Chr.12: 15322829-15322829 on Build GRCh38
rs958465451 Chr.12: 15322632-15322632 on Build GRCh38
rs770570916 Chr.12: 15322676-15322676 on Build GRCh38
rs574807123 Chr.12: 15322718-15322718 on Build GRCh38
rs911957340 Chr.12: 15322955-15322955 on Build GRCh38
rs986273374 Chr.12: 15322945-15322945 on Build GRCh38
rs1023275605 Chr.12: 15323035-15323035 on Build GRCh38
rs1038961018 Chr.12: 15322610-15322610 on Build GRCh38
rs758090360 Chr.12: 15322760-15322760 on Build GRCh38
rs577850597 Chr.12: 15323040-15323040 on Build GRCh38
rs894178530 Chr.12: 15323031-15323031 on Build GRCh38
rs747135922 Chr.12: 15322822-15322823 on Build GRCh38
rs953464908 Chr.12: 15322927-15322927 on Build GRCh38
rs999810961 Chr.12: 15322598-15322598 on Build GRCh38
rs1032754043 Chr.12: 15322624-15322624 on Build GRCh38
rs562875174 Chr.12: 15322587-15322587 on Build GRCh38
rs370419176 Chr.12: 15322813-15322813 on Build GRCh38
rs529204700 Chr.12: 15322850-15322850 on Build GRCh38
rs999711808 Chr.12: 15322726-15322726 on Build GRCh38
rs766679098 Chr.12: 15322833-15322833 on Build GRCh38
rs924751850 Chr.12: 15322988-15322988 on Build GRCh38
rs545926860 Chr.12: 15322777-15322777 on Build GRCh38
rs762407319 Chr.12: 15322706-15322706 on Build GRCh38

View Full Product Details