Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 22195241-22195736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST8SIA1
Entrez Gene ID
6489
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1
Gene Aliases
GD3S, SIAT8, SIAT8-A, SIAT8A, ST8SiaI
UniGene
Hs.408614
Gene Chromosome Location
Chr.12: 22193391-22334714 on Build GRCh38
Transcripts
NM_001304450.1 NM_003034.3

SNP Detail

SNP ID Chromosome Location
rs1038419650 Chr.12: 22195501-22195501 on Build GRCh38
rs187791721 Chr.12: 22195411-22195411 on Build GRCh38
rs998478781 Chr.12: 22195424-22195424 on Build GRCh38
rs565319929 Chr.12: 22195564-22195564 on Build GRCh38
rs369364659 Chr.12: 22195279-22195279 on Build GRCh38
rs975722555 Chr.12: 22195440-22195440 on Build GRCh38
rs965726428 Chr.12: 22195370-22195370 on Build GRCh38
rs927340591 Chr.12: 22195617-22195617 on Build GRCh38
rs201596276 Chr.12: 22195418-22195419 on Build GRCh38
rs6487294 Chr.12: 22195566-22195566 on Build GRCh38
rs950087622 Chr.12: 22195575-22195575 on Build GRCh38
rs908606093 Chr.12: 22195340-22195340 on Build GRCh38
rs535418036 Chr.12: 22195365-22195365 on Build GRCh38
rs373711406 Chr.12: 22195457-22195461 on Build GRCh38
rs530678930 Chr.12: 22195306-22195306 on Build GRCh38
rs199894680 Chr.12: 22195435-22195439 on Build GRCh38
rs955403070 Chr.12: 22195446-22195446 on Build GRCh38
rs1054698130 Chr.12: 22195273-22195273 on Build GRCh38
rs545010439 Chr.12: 22195435-22195444 on Build GRCh38
rs796117509 Chr.12: 22195487-22195487 on Build GRCh38
rs961623665 Chr.12: 22195503-22195503 on Build GRCh38
rs566144273 Chr.12: 22195534-22195534 on Build GRCh38
rs986623372 Chr.12: 22195452-22195452 on Build GRCh38
rs768103829 Chr.12: 22195457-22195458 on Build GRCh38
rs1051598236 Chr.12: 22195289-22195289 on Build GRCh38
rs995913423 Chr.12: 22195388-22195388 on Build GRCh38
rs1055334809 Chr.12: 22195387-22195387 on Build GRCh38
rs980152121 Chr.12: 22195603-22195603 on Build GRCh38
rs940017738 Chr.12: 22195361-22195361 on Build GRCh38
rs919919688 Chr.12: 22195515-22195515 on Build GRCh38
rs202155885 Chr.12: 22195502-22195502 on Build GRCh38
rs949590274 Chr.12: 22195589-22195589 on Build GRCh38
rs967065698 Chr.12: 22195422-22195422 on Build GRCh38
rs192084775 Chr.12: 22195568-22195568 on Build GRCh38
rs553908392 Chr.12: 22195498-22195498 on Build GRCh38
rs1024486607 Chr.12: 22195419-22195419 on Build GRCh38
rs535144027 Chr.12: 22195541-22195541 on Build GRCh38
rs1042493204 Chr.12: 22195608-22195608 on Build GRCh38
rs1010581523 Chr.12: 22195364-22195364 on Build GRCh38
rs954193020 Chr.12: 22195416-22195416 on Build GRCh38
rs936094278 Chr.12: 22195641-22195641 on Build GRCh38
rs73264086 Chr.12: 22195673-22195673 on Build GRCh38
rs921130771 Chr.12: 22195443-22195443 on Build GRCh38
rs973417393 Chr.12: 22195527-22195527 on Build GRCh38
rs772667275 Chr.12: 22195335-22195335 on Build GRCh38
rs1000023645 Chr.12: 22195271-22195271 on Build GRCh38
rs957318392 Chr.12: 22195434-22195435 on Build GRCh38
rs550893247 Chr.12: 22195339-22195339 on Build GRCh38
rs1032636608 Chr.12: 22195427-22195428 on Build GRCh38
rs891674893 Chr.12: 22195294-22195294 on Build GRCh38
rs1038347130 Chr.12: 22195362-22195362 on Build GRCh38

View Full Product Details