Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27081850-27082345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf71
Entrez Gene ID
728858
Gene Name
chromosome 12 open reading frame 71
Gene Aliases
-
UniGene
Hs.334003
Gene Chromosome Location
Chr.12: 27081057-27084548 on Build GRCh38
Transcripts
NM_001080406.1 XM_011520847.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6072143 c.362A>G p.K121R Chr.12: 27082122-27082122 on Build GRCh38
3871414 c.146C>T p.P49L Chr.12: 27082338-27082338 on Build GRCh38
6072142 c.340C>A p.L114M Chr.12: 27082144-27082144 on Build GRCh38
3931607 c.503C>T p.P168L Chr.12: 27081981-27081981 on Build GRCh38
6436017 c.403G>T p.V135L Chr.12: 27082081-27082081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747776775 Chr.12: 27082248-27082248 on Build GRCh38
rs549676884 Chr.12: 27082144-27082144 on Build GRCh38
rs1012227960 Chr.12: 27081915-27081915 on Build GRCh38
rs61741740 Chr.12: 27081991-27081991 on Build GRCh38
rs372491000 Chr.12: 27082082-27082082 on Build GRCh38
rs186143933 Chr.12: 27082083-27082083 on Build GRCh38
rs755522442 Chr.12: 27081892-27081892 on Build GRCh38
rs757041791 Chr.12: 27081999-27081999 on Build GRCh38
rs369070910 Chr.12: 27082080-27082080 on Build GRCh38
rs765466815 Chr.12: 27082126-27082126 on Build GRCh38
rs753780825 Chr.12: 27082233-27082233 on Build GRCh38
rs756068119 Chr.12: 27082140-27082140 on Build GRCh38
rs745348940 Chr.12: 27081911-27081911 on Build GRCh38
rs778239185 Chr.12: 27082252-27082252 on Build GRCh38
rs758021995 Chr.12: 27082322-27082322 on Build GRCh38
rs748803775 Chr.12: 27082143-27082143 on Build GRCh38
rs370734099 Chr.12: 27082205-27082205 on Build GRCh38
rs781083916 Chr.12: 27082003-27082003 on Build GRCh38
rs199571805 Chr.12: 27082191-27082191 on Build GRCh38
rs540561313 Chr.12: 27081905-27081905 on Build GRCh38
rs752898958 Chr.12: 27082196-27082196 on Build GRCh38
rs746427345 Chr.12: 27082271-27082271 on Build GRCh38
rs773919567 Chr.12: 27082038-27082038 on Build GRCh38
rs762189922 Chr.12: 27081951-27081951 on Build GRCh38
rs538607959 Chr.12: 27082315-27082315 on Build GRCh38
rs377393417 Chr.12: 27082204-27082204 on Build GRCh38
rs754509391 Chr.12: 27082239-27082239 on Build GRCh38
rs770922820 Chr.12: 27081894-27081894 on Build GRCh38
rs758142785 Chr.12: 27082250-27082250 on Build GRCh38
rs771471009 Chr.12: 27082259-27082259 on Build GRCh38
rs370338167 Chr.12: 27082287-27082287 on Build GRCh38
rs776062388 Chr.12: 27082102-27082103 on Build GRCh38
rs772410901 Chr.12: 27082166-27082166 on Build GRCh38
rs544686402 Chr.12: 27082065-27082065 on Build GRCh38
rs773721194 Chr.12: 27082173-27082173 on Build GRCh38
rs768041484 Chr.12: 27081975-27081975 on Build GRCh38
rs200299981 Chr.12: 27082189-27082189 on Build GRCh38
rs754881895 Chr.12: 27082310-27082310 on Build GRCh38
rs747345893 Chr.12: 27082253-27082253 on Build GRCh38
rs763098046 Chr.12: 27082202-27082202 on Build GRCh38
rs778616967 Chr.12: 27082154-27082154 on Build GRCh38
rs543742737 Chr.12: 27082232-27082232 on Build GRCh38
rs751019695 Chr.12: 27081979-27081979 on Build GRCh38
rs779503098 Chr.12: 27082141-27082141 on Build GRCh38
rs762599214 Chr.12: 27081935-27081935 on Build GRCh38
rs763196274 Chr.12: 27082279-27082279 on Build GRCh38
rs774608629 Chr.12: 27082291-27082291 on Build GRCh38
rs149107126 Chr.12: 27081949-27081949 on Build GRCh38
rs760453270 Chr.12: 27082306-27082306 on Build GRCh38
rs544874543 Chr.12: 27082077-27082077 on Build GRCh38
rs867431250 Chr.12: 27082301-27082301 on Build GRCh38
rs756246538 Chr.12: 27081980-27081980 on Build GRCh38
rs376451189 Chr.12: 27082124-27082124 on Build GRCh38
rs555270232 Chr.12: 27081919-27081919 on Build GRCh38
rs754440913 Chr.12: 27081873-27081873 on Build GRCh38
rs866975780 Chr.12: 27081955-27081955 on Build GRCh38
rs747819090 Chr.12: 27082157-27082157 on Build GRCh38
rs779554039 Chr.12: 27082031-27082031 on Build GRCh38
rs74075111 Chr.12: 27081945-27081945 on Build GRCh38
rs768389453 Chr.12: 27082037-27082037 on Build GRCh38
rs777316548 Chr.12: 27082190-27082190 on Build GRCh38
rs752174905 Chr.12: 27082316-27082316 on Build GRCh38
rs745875375 Chr.12: 27082024-27082024 on Build GRCh38
rs554894534 Chr.12: 27082115-27082115 on Build GRCh38
rs61741737 Chr.12: 27081981-27081981 on Build GRCh38
rs764720630 Chr.12: 27082244-27082245 on Build GRCh38
rs1045116328 Chr.12: 27081926-27081926 on Build GRCh38
rs766230920 Chr.12: 27082308-27082308 on Build GRCh38
rs779948963 Chr.12: 27082228-27082228 on Build GRCh38
rs746667729 Chr.12: 27082213-27082213 on Build GRCh38
rs750359070 Chr.12: 27082257-27082258 on Build GRCh38
rs75158113 Chr.12: 27082193-27082193 on Build GRCh38
rs752910392 Chr.12: 27082127-27082127 on Build GRCh38
rs376311282 Chr.12: 27081932-27081932 on Build GRCh38
rs748890156 Chr.12: 27082036-27082036 on Build GRCh38
rs762219231 Chr.12: 27082043-27082043 on Build GRCh38
rs761211082 Chr.12: 27082187-27082187 on Build GRCh38
rs761390291 Chr.12: 27082241-27082241 on Build GRCh38
rs761266145 Chr.12: 27082073-27082073 on Build GRCh38
rs189420548 Chr.12: 27082264-27082264 on Build GRCh38
rs201692205 Chr.12: 27082278-27082278 on Build GRCh38
rs746851563 Chr.12: 27081995-27081995 on Build GRCh38
rs757059798 Chr.12: 27082135-27082135 on Build GRCh38
rs708167 Chr.12: 27082066-27082066 on Build GRCh38
rs200731094 Chr.12: 27082029-27082029 on Build GRCh38
rs780971670 Chr.12: 27082138-27082138 on Build GRCh38
rs755237630 Chr.12: 27081992-27081992 on Build GRCh38

View Full Product Details