Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27470543-27471053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMCO2
Entrez Gene ID
341346
Gene Name
single-pass membrane protein with coiled-coil domains 2
Gene Aliases
C12orf70
UniGene
Hs.282121
Gene Chromosome Location
Chr.12: 27446482-27506911 on Build GRCh38
Transcripts
NM_001145010.1 XM_011520634.2 XM_011520635.2 XM_011520636.2 XM_017019254.1 XM_017019255.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6300061 c.54G>T p.M18I Chr.12: 27470685-27470685 on Build GRCh38
1361079 c.12G>A p.T4T Chr.12: 27470643-27470643 on Build GRCh38
6553101 c.21C>T p.N7N Chr.12: 27470652-27470652 on Build GRCh38
4935875 c.81G>T p.K27N Chr.12: 27470712-27470712 on Build GRCh38
6571286 c.114G>A p.V38V Chr.12: 27470745-27470745 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937851657 Chr.12: 27470815-27470815 on Build GRCh38
rs977916324 Chr.12: 27470878-27470878 on Build GRCh38
rs116271792 Chr.12: 27470583-27470583 on Build GRCh38
rs1019678888 Chr.12: 27471008-27471008 on Build GRCh38
rs773242213 Chr.12: 27470809-27470809 on Build GRCh38
rs544593871 Chr.12: 27470842-27470842 on Build GRCh38
rs898539810 Chr.12: 27470817-27470817 on Build GRCh38
rs972754947 Chr.12: 27470721-27470721 on Build GRCh38
rs891413509 Chr.12: 27470789-27470789 on Build GRCh38
rs987133506 Chr.12: 27470987-27470988 on Build GRCh38
rs1033681609 Chr.12: 27470601-27470601 on Build GRCh38
rs373506540 Chr.12: 27470643-27470643 on Build GRCh38
rs760527205 Chr.12: 27470593-27470593 on Build GRCh38
rs921708936 Chr.12: 27470907-27470907 on Build GRCh38
rs546151440 Chr.12: 27470821-27470821 on Build GRCh38
rs766544796 Chr.12: 27470815-27470818 on Build GRCh38
rs73086401 Chr.12: 27470725-27470725 on Build GRCh38
rs558131580 Chr.12: 27470642-27470642 on Build GRCh38
rs377557693 Chr.12: 27470695-27470695 on Build GRCh38
rs936424699 Chr.12: 27470664-27470664 on Build GRCh38
rs411432 Chr.12: 27470818-27470818 on Build GRCh38
rs202054477 Chr.12: 27470758-27470758 on Build GRCh38
rs752188747 Chr.12: 27470658-27470658 on Build GRCh38
rs366980 Chr.12: 27470883-27470883 on Build GRCh38
rs758995977 Chr.12: 27470640-27470640 on Build GRCh38
rs912673384 Chr.12: 27470684-27470684 on Build GRCh38
rs867770729 Chr.12: 27470946-27470946 on Build GRCh38
rs868301276 Chr.12: 27470844-27470844 on Build GRCh38
rs395098 Chr.12: 27470625-27470625 on Build GRCh38
rs555243671 Chr.12: 27470675-27470675 on Build GRCh38
rs396407 Chr.12: 27470795-27470795 on Build GRCh38
rs543229874 Chr.12: 27471018-27471018 on Build GRCh38
rs75360866 Chr.12: 27470986-27470986 on Build GRCh38
rs376726851 Chr.12: 27470609-27470609 on Build GRCh38
rs763204446 Chr.12: 27470904-27470904 on Build GRCh38
rs577264223 Chr.12: 27470837-27470837 on Build GRCh38
rs1029000146 Chr.12: 27470950-27470950 on Build GRCh38
rs60834107 Chr.12: 27470729-27470729 on Build GRCh38
rs772010350 Chr.12: 27470717-27470717 on Build GRCh38
rs746401543 Chr.12: 27470759-27470759 on Build GRCh38
rs12819243 Chr.12: 27470960-27470960 on Build GRCh38
rs556126313 Chr.12: 27470831-27470831 on Build GRCh38
rs957783893 Chr.12: 27470897-27470897 on Build GRCh38
rs1051270520 Chr.12: 27470744-27470744 on Build GRCh38
rs1009226655 Chr.12: 27470604-27470604 on Build GRCh38
rs930518417 Chr.12: 27470774-27470774 on Build GRCh38
rs1013616008 Chr.12: 27470832-27470832 on Build GRCh38
rs1021143914 Chr.12: 27470619-27470619 on Build GRCh38
rs1008853030 Chr.12: 27470970-27470970 on Build GRCh38
rs959250850 Chr.12: 27470620-27470620 on Build GRCh38
rs1001083836 Chr.12: 27470899-27470899 on Build GRCh38
rs527960445 Chr.12: 27470969-27470969 on Build GRCh38
rs57008581 Chr.12: 27470731-27470731 on Build GRCh38
rs561292253 Chr.12: 27471027-27471027 on Build GRCh38
rs192829459 Chr.12: 27470659-27470659 on Build GRCh38
rs757701331 Chr.12: 27470748-27470748 on Build GRCh38

View Full Product Details