Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 29502424-29502931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMTC1
Entrez Gene ID
83857
Gene Name
transmembrane and tetratricopeptide repeat containing 1
Gene Aliases
ARG99, OLF, TMTC1A
UniGene
Hs.401954
Gene Chromosome Location
Chr.12: 29500813-29784759 on Build GRCh38
Transcripts
NM_001193451.1 NM_175861.3 XM_005253499.4 XM_017020003.1 XM_017020004.1 XM_017020005.1 XM_017020007.1

SNP Detail

SNP ID Chromosome Location
rs906862329 Chr.12: 29502690-29502690 on Build GRCh38
rs181960245 Chr.12: 29502890-29502890 on Build GRCh38
rs114327726 Chr.12: 29502787-29502787 on Build GRCh38
rs577696305 Chr.12: 29502621-29502621 on Build GRCh38
rs1021263781 Chr.12: 29502705-29502705 on Build GRCh38
rs1035467170 Chr.12: 29502695-29502695 on Build GRCh38
rs867177618 Chr.12: 29502668-29502668 on Build GRCh38
rs995734205 Chr.12: 29502651-29502651 on Build GRCh38
rs981191301 Chr.12: 29502508-29502508 on Build GRCh38
rs891193622 Chr.12: 29502588-29502588 on Build GRCh38
rs867397749 Chr.12: 29502680-29502680 on Build GRCh38
rs966002388 Chr.12: 29502877-29502877 on Build GRCh38
rs368472731 Chr.12: 29502563-29502563 on Build GRCh38
rs4931197 Chr.12: 29502682-29502682 on Build GRCh38
rs1048191699 Chr.12: 29502791-29502791 on Build GRCh38
rs940064662 Chr.12: 29502615-29502615 on Build GRCh38
rs138209909 Chr.12: 29502616-29502616 on Build GRCh38
rs974661235 Chr.12: 29502782-29502782 on Build GRCh38
rs1042404272 Chr.12: 29502684-29502684 on Build GRCh38
rs10332 Chr.12: 29502704-29502704 on Build GRCh38
rs3759198 Chr.12: 29502543-29502543 on Build GRCh38
rs990308588 Chr.12: 29502664-29502664 on Build GRCh38
rs906001914 Chr.12: 29502663-29502663 on Build GRCh38
rs553624962 Chr.12: 29502817-29502817 on Build GRCh38
rs898759320 Chr.12: 29502648-29502648 on Build GRCh38
rs922047076 Chr.12: 29502661-29502661 on Build GRCh38
rs183756514 Chr.12: 29502481-29502481 on Build GRCh38
rs955769418 Chr.12: 29502696-29502696 on Build GRCh38
rs138396720 Chr.12: 29502545-29502545 on Build GRCh38
rs940075569 Chr.12: 29502857-29502857 on Build GRCh38
rs777331655 Chr.12: 29502672-29502672 on Build GRCh38
rs946306122 Chr.12: 29502456-29502456 on Build GRCh38
rs144424029 Chr.12: 29502560-29502560 on Build GRCh38
rs531601352 Chr.12: 29502848-29502848 on Build GRCh38
rs1036987149 Chr.12: 29502630-29502630 on Build GRCh38
rs983112480 Chr.12: 29502589-29502589 on Build GRCh38
rs914734007 Chr.12: 29502670-29502670 on Build GRCh38
rs371940010 Chr.12: 29502603-29502603 on Build GRCh38
rs898478126 Chr.12: 29502891-29502891 on Build GRCh38
rs955719388 Chr.12: 29502511-29502511 on Build GRCh38
rs928301372 Chr.12: 29502509-29502509 on Build GRCh38
rs111349446 Chr.12: 29502870-29502872 on Build GRCh38
rs1023572471 Chr.12: 29502659-29502659 on Build GRCh38
rs756384026 Chr.12: 29502730-29502730 on Build GRCh38
rs912680798 Chr.12: 29502849-29502849 on Build GRCh38
rs879810063 Chr.12: 29502591-29502591 on Build GRCh38
rs1004129906 Chr.12: 29502847-29502847 on Build GRCh38
rs778556406 Chr.12: 29502535-29502535 on Build GRCh38
rs538066270 Chr.12: 29502565-29502565 on Build GRCh38
rs192555527 Chr.12: 29502467-29502467 on Build GRCh38
rs149250122 Chr.12: 29502550-29502550 on Build GRCh38
rs189579469 Chr.12: 29502466-29502466 on Build GRCh38
rs1023100106 Chr.12: 29502487-29502487 on Build GRCh38
rs914530862 Chr.12: 29502544-29502544 on Build GRCh38
rs933192971 Chr.12: 29502831-29502831 on Build GRCh38
rs371982730 Chr.12: 29502746-29502746 on Build GRCh38
rs189305214 Chr.12: 29502874-29502874 on Build GRCh38
rs886943915 Chr.12: 29502819-29502819 on Build GRCh38
rs1051218564 Chr.12: 29502582-29502582 on Build GRCh38
rs931569625 Chr.12: 29502907-29502907 on Build GRCh38
rs970214541 Chr.12: 29502504-29502504 on Build GRCh38
rs544761780 Chr.12: 29502660-29502660 on Build GRCh38

View Full Product Details