Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48694147-48694646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNT1
Entrez Gene ID
904
Gene Name
cyclin T1
Gene Aliases
CCNT, CYCT1, HIVE1
UniGene
Hs.92308
Gene Chromosome Location
Chr.12: 48688458-48716998 on Build GRCh38
Transcripts
NM_001240.3 NM_001277842.1 XM_017020197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3986951 c.988C>T p.P330S Chr.12: 48694226-48694226 on Build GRCh38
4820931 c.935C>G p.S312C Chr.12: 48694279-48694279 on Build GRCh38
5641776 c.1038T>G p.T346T Chr.12: 48694176-48694176 on Build GRCh38
5500539 c.859A>G p.I287V Chr.12: 48694355-48694355 on Build GRCh38
6072557 c.831G>T p.K277N Chr.12: 48694383-48694383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755560513 Chr.12: 48694267-48694267 on Build GRCh38
rs769068390 Chr.12: 48694585-48694585 on Build GRCh38
rs754521618 Chr.12: 48694422-48694422 on Build GRCh38
rs757605829 Chr.12: 48694244-48694244 on Build GRCh38
rs763463430 Chr.12: 48694292-48694292 on Build GRCh38
rs770192921 Chr.12: 48694220-48694220 on Build GRCh38
rs555137940 Chr.12: 48694495-48694495 on Build GRCh38
rs923760801 Chr.12: 48694226-48694226 on Build GRCh38
rs960729876 Chr.12: 48694367-48694367 on Build GRCh38
rs538166862 Chr.12: 48694469-48694469 on Build GRCh38
rs779541732 Chr.12: 48694278-48694278 on Build GRCh38
rs558154621 Chr.12: 48694393-48694393 on Build GRCh38
rs771829615 Chr.12: 48694289-48694289 on Build GRCh38
rs747942854 Chr.12: 48694288-48694288 on Build GRCh38
rs970415376 Chr.12: 48694556-48694556 on Build GRCh38
rs372886439 Chr.12: 48694428-48694428 on Build GRCh38
rs560534520 Chr.12: 48694568-48694568 on Build GRCh38
rs778621038 Chr.12: 48694427-48694427 on Build GRCh38
rs768371612 Chr.12: 48694173-48694173 on Build GRCh38
rs754644478 Chr.12: 48694280-48694280 on Build GRCh38
rs979595178 Chr.12: 48694538-48694538 on Build GRCh38
rs775736756 Chr.12: 48694294-48694294 on Build GRCh38
rs199856816 Chr.12: 48694217-48694217 on Build GRCh38
rs866825070 Chr.12: 48694175-48694175 on Build GRCh38
rs746894129 Chr.12: 48694446-48694446 on Build GRCh38
rs575488596 Chr.12: 48694399-48694399 on Build GRCh38
rs144308598 Chr.12: 48694549-48694549 on Build GRCh38
rs923708687 Chr.12: 48694608-48694608 on Build GRCh38
rs751934361 Chr.12: 48694242-48694242 on Build GRCh38
rs201440153 Chr.12: 48694371-48694371 on Build GRCh38
rs749376617 Chr.12: 48694465-48694465 on Build GRCh38
rs750953240 Chr.12: 48694266-48694266 on Build GRCh38
rs1006159634 Chr.12: 48694508-48694508 on Build GRCh38
rs374791846 Chr.12: 48694216-48694216 on Build GRCh38
rs772022667 Chr.12: 48694200-48694200 on Build GRCh38
rs150867063 Chr.12: 48694272-48694272 on Build GRCh38
rs761942365 Chr.12: 48694470-48694470 on Build GRCh38
rs933860655 Chr.12: 48694247-48694247 on Build GRCh38
rs140020979 Chr.12: 48694301-48694301 on Build GRCh38
rs574959746 Chr.12: 48694525-48694525 on Build GRCh38
rs770765539 Chr.12: 48694460-48694464 on Build GRCh38
rs912893605 Chr.12: 48694583-48694583 on Build GRCh38
rs566116309 Chr.12: 48694360-48694360 on Build GRCh38
rs775789988 Chr.12: 48694222-48694222 on Build GRCh38
rs768972559 Chr.12: 48694330-48694330 on Build GRCh38
rs770972200 Chr.12: 48694454-48694454 on Build GRCh38
rs774159319 Chr.12: 48694178-48694178 on Build GRCh38
rs866673208 Chr.12: 48694331-48694331 on Build GRCh38
rs778965689 Chr.12: 48694466-48694474 on Build GRCh38
rs111231208 Chr.12: 48694417-48694417 on Build GRCh38
rs748896346 Chr.12: 48694279-48694279 on Build GRCh38
rs777460626 Chr.12: 48694431-48694431 on Build GRCh38
rs969946788 Chr.12: 48694518-48694518 on Build GRCh38
rs546318630 Chr.12: 48694353-48694353 on Build GRCh38
rs538696433 Chr.12: 48694290-48694290 on Build GRCh38
rs140318462 Chr.12: 48694185-48694185 on Build GRCh38
rs957068104 Chr.12: 48694565-48694565 on Build GRCh38
rs765823227 Chr.12: 48694414-48694414 on Build GRCh38
rs767790293 Chr.12: 48694370-48694370 on Build GRCh38
rs781321741 Chr.12: 48694456-48694456 on Build GRCh38
rs775724017 Chr.12: 48694232-48694232 on Build GRCh38
rs763301494 Chr.12: 48694231-48694231 on Build GRCh38
rs753320814 Chr.12: 48694418-48694418 on Build GRCh38
rs758153792 Chr.12: 48694430-48694430 on Build GRCh38
rs369135021 Chr.12: 48694239-48694239 on Build GRCh38
rs768066318 Chr.12: 48694265-48694265 on Build GRCh38

View Full Product Details