Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49097448-49097948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMBR1L
Entrez Gene ID
55716
Gene Name
limb development membrane protein 1 like
Gene Aliases
LIMR
UniGene
Hs.272838
Gene Chromosome Location
Chr.12: 49096551-49111535 on Build GRCh38
Transcripts
NM_001300750.1 NM_001300751.1 NM_018113.3 XM_011538563.2 XM_011538564.2 XM_011538565.2 XM_011538566.2 XM_017019626.1 XM_017019627.1 XM_017019628.1 XM_017019629.1 XM_017019630.1 XM_017019631.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6280997 c.1388G>A p.G463E Chr.12: 49097739-49097739 on Build GRCh38
6502989 c.1448C>A p.S483Y Chr.12: 49097694-49097694 on Build GRCh38
5730883 c.1425C>T p.P475P Chr.12: 49097702-49097702 on Build GRCh38
940105 c.1399C>T p.L467L Chr.12: 49097728-49097728 on Build GRCh38
6701708 c.1411G>A p.V471I Chr.12: 49097716-49097716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780315300 Chr.12: 49097724-49097724 on Build GRCh38
rs141299860 Chr.12: 49097710-49097710 on Build GRCh38
rs552872781 Chr.12: 49097475-49097475 on Build GRCh38
rs575143385 Chr.12: 49097723-49097723 on Build GRCh38
rs984110052 Chr.12: 49097480-49097480 on Build GRCh38
rs756200315 Chr.12: 49097910-49097910 on Build GRCh38
rs991178880 Chr.12: 49097575-49097575 on Build GRCh38
rs915615257 Chr.12: 49097822-49097822 on Build GRCh38
rs574819296 Chr.12: 49097664-49097664 on Build GRCh38
rs978534000 Chr.12: 49097845-49097845 on Build GRCh38
rs774628704 Chr.12: 49097846-49097846 on Build GRCh38
rs750408798 Chr.12: 49097721-49097721 on Build GRCh38
rs369859624 Chr.12: 49097787-49097787 on Build GRCh38
rs901436718 Chr.12: 49097702-49097702 on Build GRCh38
rs946187690 Chr.12: 49097875-49097875 on Build GRCh38
rs200146930 Chr.12: 49097623-49097623 on Build GRCh38
rs758224854 Chr.12: 49097717-49097717 on Build GRCh38
rs751706882 Chr.12: 49097743-49097747 on Build GRCh38
rs375664220 Chr.12: 49097895-49097895 on Build GRCh38
rs915607403 Chr.12: 49097605-49097605 on Build GRCh38
rs760171023 Chr.12: 49097768-49097768 on Build GRCh38
rs530644210 Chr.12: 49097598-49097598 on Build GRCh38
rs544910447 Chr.12: 49097711-49097711 on Build GRCh38
rs386376451 Chr.12: 49097915-49097916 on Build GRCh38
rs372096261 Chr.12: 49097716-49097716 on Build GRCh38
rs767086535 Chr.12: 49097701-49097701 on Build GRCh38
rs367823171 Chr.12: 49097687-49097687 on Build GRCh38
rs148501287 Chr.12: 49097915-49097915 on Build GRCh38
rs759970811 Chr.12: 49097910-49097910 on Build GRCh38
rs540826142 Chr.12: 49097659-49097659 on Build GRCh38
rs748711677 Chr.12: 49097743-49097743 on Build GRCh38
rs35864667 Chr.12: 49097736-49097737 on Build GRCh38
rs375344594 Chr.12: 49097779-49097779 on Build GRCh38
rs931334411 Chr.12: 49097479-49097479 on Build GRCh38
rs111446209 Chr.12: 49097921-49097921 on Build GRCh38
rs34184319 Chr.12: 49097767-49097768 on Build GRCh38
rs763593679 Chr.12: 49097499-49097499 on Build GRCh38
rs779058769 Chr.12: 49097636-49097636 on Build GRCh38
rs772548434 Chr.12: 49097704-49097704 on Build GRCh38
rs927537852 Chr.12: 49097490-49097490 on Build GRCh38
rs952826681 Chr.12: 49097771-49097771 on Build GRCh38
rs538009920 Chr.12: 49097555-49097555 on Build GRCh38
rs371523859 Chr.12: 49097662-49097662 on Build GRCh38
rs779264177 Chr.12: 49097918-49097918 on Build GRCh38
rs928574556 Chr.12: 49097520-49097520 on Build GRCh38
rs889464610 Chr.12: 49097917-49097917 on Build GRCh38
rs760390520 Chr.12: 49097705-49097705 on Build GRCh38
rs544069213 Chr.12: 49097774-49097774 on Build GRCh38
rs1005626611 Chr.12: 49097722-49097722 on Build GRCh38
rs762199890 Chr.12: 49097696-49097696 on Build GRCh38
rs779213151 Chr.12: 49097734-49097734 on Build GRCh38
rs918569709 Chr.12: 49097473-49097473 on Build GRCh38
rs375349450 Chr.12: 49097755-49097755 on Build GRCh38
rs546649352 Chr.12: 49097839-49097839 on Build GRCh38
rs781350852 Chr.12: 49097719-49097719 on Build GRCh38
rs780079604 Chr.12: 49097911-49097911 on Build GRCh38
rs775843708 Chr.12: 49097673-49097673 on Build GRCh38
rs768852221 Chr.12: 49097688-49097688 on Build GRCh38
rs749493280 Chr.12: 49097728-49097728 on Build GRCh38
rs553899639 Chr.12: 49097709-49097709 on Build GRCh38
rs369732299 Chr.12: 49097767-49097767 on Build GRCh38
rs947198968 Chr.12: 49097616-49097616 on Build GRCh38
rs754971592 Chr.12: 49097628-49097628 on Build GRCh38
rs763868486 Chr.12: 49097904-49097904 on Build GRCh38
rs187490591 Chr.12: 49097550-49097550 on Build GRCh38
rs762957800 Chr.12: 49097898-49097898 on Build GRCh38
rs778199407 Chr.12: 49097924-49097924 on Build GRCh38
rs749722407 Chr.12: 49097683-49097683 on Build GRCh38
rs748395860 Chr.12: 49097919-49097919 on Build GRCh38
rs200520975 Chr.12: 49097905-49097905 on Build GRCh38
rs144688401 Chr.12: 49097833-49097833 on Build GRCh38
rs769052852 Chr.12: 49097732-49097732 on Build GRCh38
rs958909908 Chr.12: 49097823-49097823 on Build GRCh38
rs192672535 Chr.12: 49097665-49097665 on Build GRCh38
rs927337247 Chr.12: 49097817-49097817 on Build GRCh38
rs111934971 Chr.12: 49097788-49097788 on Build GRCh38

View Full Product Details