Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3405359-3405852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM16
Entrez Gene ID
63976
Gene Name
PR domain 16
Gene Aliases
CMD1LL, LVNC8, MEL1, PFM13
UniGene
Hs.99500
Gene Chromosome Location
Chr.1: 3069025-3438621 on Build GRCh38
Transcripts
NM_022114.3 NM_199454.2 XM_005244772.4 XM_005244773.4 XM_005244774.4 XM_006710814.3 XM_011541945.2 XM_017002050.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6755092 c.1098C>T p.H366H Chr.1: 3405560-3405560 on Build GRCh38
6755094 c.1045C>A p.P349T Chr.1: 3405507-3405507 on Build GRCh38
2078234 c.1090C>T p.R364W Chr.1: 3405552-3405552 on Build GRCh38
4031511 c.1128C>T p.A376A Chr.1: 3405590-3405590 on Build GRCh38
4748897 c.1039A>T p.T347S Chr.1: 3405501-3405501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370090853 Chr.1: 3405485-3405485 on Build GRCh38
rs372711563 Chr.1: 3405471-3405471 on Build GRCh38
rs904710033 Chr.1: 3405776-3405776 on Build GRCh38
rs774486846 Chr.1: 3405467-3405467 on Build GRCh38
rs1056081266 Chr.1: 3405811-3405811 on Build GRCh38
rs374018370 Chr.1: 3405451-3405451 on Build GRCh38
rs775537180 Chr.1: 3405444-3405444 on Build GRCh38
rs1019838568 Chr.1: 3405821-3405821 on Build GRCh38
rs375762774 Chr.1: 3405689-3405689 on Build GRCh38
rs746868217 Chr.1: 3405502-3405502 on Build GRCh38
rs374836387 Chr.1: 3405379-3405379 on Build GRCh38
rs745769134 Chr.1: 3405588-3405588 on Build GRCh38
rs1034401350 Chr.1: 3405554-3405554 on Build GRCh38
rs768571449 Chr.1: 3405528-3405528 on Build GRCh38
rs182903782 Chr.1: 3405509-3405509 on Build GRCh38
rs1049463147 Chr.1: 3405487-3405487 on Build GRCh38
rs778504357 Chr.1: 3405481-3405481 on Build GRCh38
rs545898678 Chr.1: 3405775-3405775 on Build GRCh38
rs774523031 Chr.1: 3405531-3405531 on Build GRCh38
rs756894589 Chr.1: 3405800-3405800 on Build GRCh38
rs762564991 Chr.1: 3405680-3405680 on Build GRCh38
rs577134367 Chr.1: 3405773-3405773 on Build GRCh38
rs367771660 Chr.1: 3405697-3405697 on Build GRCh38
rs879187684 Chr.1: 3405402-3405402 on Build GRCh38
rs752145993 Chr.1: 3405672-3405672 on Build GRCh38
rs372181792 Chr.1: 3405671-3405671 on Build GRCh38
rs1009187651 Chr.1: 3405405-3405405 on Build GRCh38
rs1001899924 Chr.1: 3405440-3405440 on Build GRCh38
rs967364639 Chr.1: 3405818-3405818 on Build GRCh38
rs992948020 Chr.1: 3405469-3405469 on Build GRCh38
rs976239157 Chr.1: 3405687-3405687 on Build GRCh38
rs200562747 Chr.1: 3405555-3405555 on Build GRCh38
rs754579975 Chr.1: 3405480-3405480 on Build GRCh38
rs769778096 Chr.1: 3405593-3405593 on Build GRCh38
rs1056295149 Chr.1: 3405777-3405777 on Build GRCh38
rs944421182 Chr.1: 3405740-3405740 on Build GRCh38
rs771381295 Chr.1: 3405633-3405633 on Build GRCh38
rs1020177340 Chr.1: 3405412-3405412 on Build GRCh38
rs777390254 Chr.1: 3405702-3405702 on Build GRCh38
rs549681722 Chr.1: 3405407-3405407 on Build GRCh38
rs1001805709 Chr.1: 3405570-3405570 on Build GRCh38
rs377761563 Chr.1: 3405452-3405452 on Build GRCh38
rs757093442 Chr.1: 3405578-3405578 on Build GRCh38
rs371240361 Chr.1: 3405477-3405477 on Build GRCh38
rs986088535 Chr.1: 3405706-3405706 on Build GRCh38
rs749912933 Chr.1: 3405681-3405682 on Build GRCh38
rs61745281 Chr.1: 3405575-3405575 on Build GRCh38
rs374947232 Chr.1: 3405478-3405478 on Build GRCh38
rs777550737 Chr.1: 3405572-3405572 on Build GRCh38
rs977613443 Chr.1: 3405465-3405465 on Build GRCh38
rs376937143 Chr.1: 3405479-3405479 on Build GRCh38
rs754343098 Chr.1: 3405549-3405549 on Build GRCh38
rs374258269 Chr.1: 3405472-3405472 on Build GRCh38
rs369196369 Chr.1: 3405380-3405380 on Build GRCh38
rs767835445 Chr.1: 3405536-3405536 on Build GRCh38
rs752263127 Chr.1: 3405561-3405561 on Build GRCh38
rs2483221 Chr.1: 3405533-3405533 on Build GRCh38
rs914803985 Chr.1: 3405427-3405427 on Build GRCh38
rs913613194 Chr.1: 3405746-3405746 on Build GRCh38
rs763713785 Chr.1: 3405681-3405681 on Build GRCh38
rs374263452 Chr.1: 3405632-3405632 on Build GRCh38
rs749348281 Chr.1: 3405524-3405524 on Build GRCh38
rs569792747 Chr.1: 3405664-3405664 on Build GRCh38
rs747159959 Chr.1: 3405700-3405700 on Build GRCh38
rs368744559 Chr.1: 3405658-3405658 on Build GRCh38
rs968701973 Chr.1: 3405439-3405439 on Build GRCh38
rs776595154 Chr.1: 3405523-3405523 on Build GRCh38
rs891992060 Chr.1: 3405418-3405418 on Build GRCh38
rs780076547 Chr.1: 3405694-3405694 on Build GRCh38
rs555638718 Chr.1: 3405707-3405707 on Build GRCh38
rs755999342 Chr.1: 3405691-3405691 on Build GRCh38
rs1012053945 Chr.1: 3405801-3405801 on Build GRCh38
rs780901939 Chr.1: 3405688-3405688 on Build GRCh38
rs1048744417 Chr.1: 3405703-3405703 on Build GRCh38
rs766969882 Chr.1: 3405547-3405547 on Build GRCh38
rs764350754 Chr.1: 3405461-3405461 on Build GRCh38
rs907050620 Chr.1: 3405765-3405765 on Build GRCh38
rs879168008 Chr.1: 3405413-3405413 on Build GRCh38
rs946454669 Chr.1: 3405752-3405752 on Build GRCh38
rs960373627 Chr.1: 3405459-3405459 on Build GRCh38
rs572364957 Chr.1: 3405711-3405711 on Build GRCh38
rs911533400 Chr.1: 3405714-3405714 on Build GRCh38
rs534869994 Chr.1: 3405715-3405715 on Build GRCh38
rs772150656 Chr.1: 3405600-3405600 on Build GRCh38
rs876657957 Chr.1: 3405543-3405543 on Build GRCh38
rs1052179221 Chr.1: 3405741-3405741 on Build GRCh38
rs778030621 Chr.1: 3405698-3405698 on Build GRCh38
rs187151343 Chr.1: 3405772-3405772 on Build GRCh38
rs1002178015 Chr.1: 3405804-3405804 on Build GRCh38
rs1021620686 Chr.1: 3405577-3405577 on Build GRCh38
rs751168823 Chr.1: 3405682-3405682 on Build GRCh38
rs143311879 Chr.1: 3405406-3405407 on Build GRCh38
rs200169663 Chr.1: 3405596-3405596 on Build GRCh38
rs777375331 Chr.1: 3405490-3405490 on Build GRCh38
rs764582197 Chr.1: 3405560-3405560 on Build GRCh38
rs965946410 Chr.1: 3405824-3405824 on Build GRCh38
rs1010446434 Chr.1: 3405544-3405544 on Build GRCh38
rs748323433 Chr.1: 3405599-3405599 on Build GRCh38
rs981548692 Chr.1: 3405398-3405398 on Build GRCh38
rs368011527 Chr.1: 3405398-3405399 on Build GRCh38
rs549821991 Chr.1: 3405587-3405587 on Build GRCh38
rs1003379939 Chr.1: 3405548-3405548 on Build GRCh38
rs759901012 Chr.1: 3405666-3405666 on Build GRCh38
rs1034800925 Chr.1: 3405399-3405399 on Build GRCh38
rs747712501 Chr.1: 3405484-3405484 on Build GRCh38
rs1014743652 Chr.1: 3405614-3405614 on Build GRCh38
rs2493305 Chr.1: 3405419-3405419 on Build GRCh38
rs958700624 Chr.1: 3405407-3405407 on Build GRCh38
rs773614460 Chr.1: 3405626-3405626 on Build GRCh38
rs79350670 Chr.1: 3405819-3405819 on Build GRCh38
rs761160322 Chr.1: 3405542-3405542 on Build GRCh38
rs1021916959 Chr.1: 3405438-3405438 on Build GRCh38
rs548409318 Chr.1: 3405421-3405421 on Build GRCh38

View Full Product Details