Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49540730-49541235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH3
Entrez Gene ID
23416
Gene Name
potassium voltage-gated channel subfamily H member 3
Gene Aliases
BEC1, ELK2, Kv12.2
UniGene
Hs.64064
Gene Chromosome Location
Chr.12: 49538541-49558312 on Build GRCh38
Transcripts
NM_001314030.1 NM_012284.2 XM_011538085.2 XM_011538086.2 XM_017019096.1 XM_017019097.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7247491 c.235G>C p.V79L Chr.12: 49541057-49541057 on Build GRCh38
6690908 c.154C>T p.L52F Chr.12: 49540976-49540976 on Build GRCh38
3461670 c.306G>A p.K102K Chr.12: 49541128-49541128 on Build GRCh38
3376279 c.104A>G p.Q35R Chr.12: 49540926-49540926 on Build GRCh38
7231891 c.173C>A p.A58D Chr.12: 49540995-49540995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770488678 Chr.12: 49541009-49541009 on Build GRCh38
rs139875282 Chr.12: 49540921-49540921 on Build GRCh38
rs781484924 Chr.12: 49540863-49540864 on Build GRCh38
rs764983879 Chr.12: 49541072-49541072 on Build GRCh38
rs147651812 Chr.12: 49541010-49541010 on Build GRCh38
rs940010053 Chr.12: 49540873-49540873 on Build GRCh38
rs528653367 Chr.12: 49540915-49540915 on Build GRCh38
rs138969562 Chr.12: 49540855-49540855 on Build GRCh38
rs984168188 Chr.12: 49540840-49540840 on Build GRCh38
rs751284625 Chr.12: 49541104-49541104 on Build GRCh38
rs771480897 Chr.12: 49541174-49541174 on Build GRCh38
rs7132792 Chr.12: 49540782-49540782 on Build GRCh38
rs753455353 Chr.12: 49541038-49541038 on Build GRCh38
rs748321656 Chr.12: 49541145-49541145 on Build GRCh38
rs758883771 Chr.12: 49540856-49540856 on Build GRCh38
rs777669265 Chr.12: 49541094-49541094 on Build GRCh38
rs773615115 Chr.12: 49540992-49540992 on Build GRCh38
rs768926424 Chr.12: 49540945-49540945 on Build GRCh38
rs746718845 Chr.12: 49540836-49540836 on Build GRCh38
rs759056953 Chr.12: 49541061-49541061 on Build GRCh38
rs1020277386 Chr.12: 49540767-49540767 on Build GRCh38
rs767884315 Chr.12: 49540991-49540991 on Build GRCh38
rs965694966 Chr.12: 49540776-49540776 on Build GRCh38
rs747967354 Chr.12: 49540882-49540882 on Build GRCh38
rs753367266 Chr.12: 49540870-49540870 on Build GRCh38
rs768658333 Chr.12: 49541140-49541140 on Build GRCh38
rs1037907213 Chr.12: 49540790-49540790 on Build GRCh38
rs773693245 Chr.12: 49541166-49541166 on Build GRCh38
rs1056130934 Chr.12: 49541050-49541050 on Build GRCh38
rs762171007 Chr.12: 49540987-49540987 on Build GRCh38
rs753141258 Chr.12: 49540947-49540959 on Build GRCh38
rs372100238 Chr.12: 49540837-49540837 on Build GRCh38
rs760934136 Chr.12: 49541172-49541172 on Build GRCh38
rs777196024 Chr.12: 49541153-49541153 on Build GRCh38
rs779137840 Chr.12: 49541142-49541142 on Build GRCh38
rs151191732 Chr.12: 49541131-49541131 on Build GRCh38
rs1011814880 Chr.12: 49541086-49541086 on Build GRCh38
rs762154452 Chr.12: 49540854-49540854 on Build GRCh38
rs996070317 Chr.12: 49541167-49541167 on Build GRCh38
rs1046650598 Chr.12: 49540858-49540858 on Build GRCh38
rs879591990 Chr.12: 49540897-49540898 on Build GRCh38
rs754743462 Chr.12: 49540871-49540871 on Build GRCh38
rs758197823 Chr.12: 49540907-49540907 on Build GRCh38
rs144443412 Chr.12: 49541124-49541124 on Build GRCh38
rs563717479 Chr.12: 49540766-49540766 on Build GRCh38
rs781096480 Chr.12: 49541128-49541128 on Build GRCh38
rs893932884 Chr.12: 49541017-49541017 on Build GRCh38
rs759923166 Chr.12: 49541189-49541189 on Build GRCh38
rs999492222 Chr.12: 49540875-49540875 on Build GRCh38
rs765864732 Chr.12: 49540866-49540866 on Build GRCh38
rs150257048 Chr.12: 49540839-49540839 on Build GRCh38
rs982175196 Chr.12: 49540887-49540887 on Build GRCh38
rs377314805 Chr.12: 49540865-49540865 on Build GRCh38
rs770756585 Chr.12: 49540932-49540932 on Build GRCh38
rs975966497 Chr.12: 49540781-49540781 on Build GRCh38
rs145328322 Chr.12: 49540934-49540934 on Build GRCh38
rs140606804 Chr.12: 49541073-49541073 on Build GRCh38
rs571808041 Chr.12: 49540933-49540933 on Build GRCh38

View Full Product Details