Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51820268-51820770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIGNL2
Entrez Gene ID
401720
Gene Name
fidgetin like 2
Gene Aliases
-
UniGene
Hs.648218
Gene Chromosome Location
Chr.12: 51817892-51848753 on Build GRCh38
Transcripts
NM_001013690.4 XM_005268870.3 XM_017019298.1 XM_017019299.1

SNP Detail

SNP ID Chromosome Location
rs775709935 Chr.12: 51820471-51820471 on Build GRCh38
rs749932323 Chr.12: 51820710-51820710 on Build GRCh38
rs756016764 Chr.12: 51820512-51820512 on Build GRCh38
rs763435303 Chr.12: 51820473-51820473 on Build GRCh38
rs930621084 Chr.12: 51820499-51820499 on Build GRCh38
rs754733766 Chr.12: 51820522-51820522 on Build GRCh38
rs763877456 Chr.12: 51820418-51820418 on Build GRCh38
rs766965271 Chr.12: 51820505-51820505 on Build GRCh38
rs749985378 Chr.12: 51820506-51820506 on Build GRCh38
rs545868458 Chr.12: 51820462-51820462 on Build GRCh38
rs760118288 Chr.12: 51820715-51820715 on Build GRCh38
rs971677548 Chr.12: 51820399-51820399 on Build GRCh38
rs575900984 Chr.12: 51820573-51820573 on Build GRCh38
rs1022370784 Chr.12: 51820722-51820722 on Build GRCh38
rs189075046 Chr.12: 51820424-51820424 on Build GRCh38
rs565526347 Chr.12: 51820557-51820557 on Build GRCh38
rs1041563786 Chr.12: 51820721-51820721 on Build GRCh38
rs931185080 Chr.12: 51820305-51820305 on Build GRCh38
rs370766286 Chr.12: 51820460-51820460 on Build GRCh38
rs918494144 Chr.12: 51820298-51820298 on Build GRCh38
rs778604083 Chr.12: 51820523-51820523 on Build GRCh38
rs754892409 Chr.12: 51820732-51820732 on Build GRCh38
rs1042696119 Chr.12: 51820398-51820398 on Build GRCh38
rs765809952 Chr.12: 51820716-51820716 on Build GRCh38
rs202036803 Chr.12: 51820451-51820451 on Build GRCh38
rs746008117 Chr.12: 51820558-51820558 on Build GRCh38
rs760147863 Chr.12: 51820408-51820408 on Build GRCh38
rs192804276 Chr.12: 51820687-51820687 on Build GRCh38
rs771323852 Chr.12: 51820540-51820540 on Build GRCh38
rs1005372679 Chr.12: 51820563-51820563 on Build GRCh38
rs966526953 Chr.12: 51820724-51820724 on Build GRCh38
rs900528013 Chr.12: 51820498-51820498 on Build GRCh38
rs768224939 Chr.12: 51820402-51820402 on Build GRCh38
rs1041385807 Chr.12: 51820591-51820591 on Build GRCh38
rs773423756 Chr.12: 51820445-51820445 on Build GRCh38
rs889914461 Chr.12: 51820693-51820693 on Build GRCh38
rs756938218 Chr.12: 51820503-51820503 on Build GRCh38
rs763378368 Chr.12: 51820579-51820579 on Build GRCh38
rs776819148 Chr.12: 51820547-51820547 on Build GRCh38
rs765882327 Chr.12: 51820467-51820467 on Build GRCh38
rs756068123 Chr.12: 51820434-51820434 on Build GRCh38
rs751022483 Chr.12: 51820420-51820420 on Build GRCh38
rs989654668 Chr.12: 51820419-51820419 on Build GRCh38
rs747176075 Chr.12: 51820538-51820538 on Build GRCh38
rs369718908 Chr.12: 51820728-51820728 on Build GRCh38
rs959030536 Chr.12: 51820598-51820598 on Build GRCh38
rs748795066 Chr.12: 51820303-51820303 on Build GRCh38
rs766578150 Chr.12: 51820405-51820405 on Build GRCh38
rs772371192 Chr.12: 51820466-51820466 on Build GRCh38
rs764382002 Chr.12: 51820488-51820488 on Build GRCh38
rs764483463 Chr.12: 51820583-51820583 on Build GRCh38
rs780106689 Chr.12: 51820518-51820518 on Build GRCh38
rs868639340 Chr.12: 51820529-51820529 on Build GRCh38
rs1027714543 Chr.12: 51820341-51820341 on Build GRCh38
rs561403471 Chr.12: 51820625-51820625 on Build GRCh38
rs774601582 Chr.12: 51820584-51820584 on Build GRCh38
rs772169289 Chr.12: 51820440-51820440 on Build GRCh38
rs117108951 Chr.12: 51820316-51820316 on Build GRCh38
rs751161297 Chr.12: 51820492-51820492 on Build GRCh38
rs972587016 Chr.12: 51820297-51820298 on Build GRCh38
rs757953247 Chr.12: 51820415-51820415 on Build GRCh38
rs779693203 Chr.12: 51820437-51820437 on Build GRCh38
rs1005017028 Chr.12: 51820385-51820385 on Build GRCh38
rs544957811 Chr.12: 51820599-51820599 on Build GRCh38
rs749194996 Chr.12: 51820519-51820519 on Build GRCh38
rs553700551 Chr.12: 51820409-51820409 on Build GRCh38
rs1045920717 Chr.12: 51820566-51820566 on Build GRCh38
rs965343878 Chr.12: 51820293-51820293 on Build GRCh38
rs776009801 Chr.12: 51820575-51820575 on Build GRCh38
rs1056817365 Chr.12: 51820343-51820343 on Build GRCh38
rs924218988 Chr.12: 51820635-51820635 on Build GRCh38
rs745766990 Chr.12: 51820433-51820433 on Build GRCh38
rs749150037 Chr.12: 51820439-51820439 on Build GRCh38
rs1020006885 Chr.12: 51820289-51820289 on Build GRCh38
rs927090678 Chr.12: 51820318-51820319 on Build GRCh38
rs778743599 Chr.12: 51820742-51820742 on Build GRCh38
rs780689277 Chr.12: 51820432-51820432 on Build GRCh38
rs895430080 Chr.12: 51820353-51820353 on Build GRCh38
rs1024883678 Chr.12: 51820397-51820397 on Build GRCh38
rs376892638 Chr.12: 51820431-51820431 on Build GRCh38
rs150720486 Chr.12: 51820355-51820355 on Build GRCh38
rs753152237 Chr.12: 51820410-51820410 on Build GRCh38
rs754328780 Chr.12: 51820407-51820407 on Build GRCh38
rs761433757 Chr.12: 51820660-51820660 on Build GRCh38

View Full Product Details