Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52948786-52949293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT18
Entrez Gene ID
3875
Gene Name
keratin 18
Gene Aliases
CK-18, CYK18, K18
UniGene
Hs.406013
Gene Chromosome Location
Chr.12: 52948871-52952901 on Build GRCh38
Transcripts
NM_000224.2 NM_199187.1 NM_001256293.1 NR_045962.1

Gene Symbol
KRT8
Entrez Gene ID
3856
Gene Name
keratin 8
Gene Aliases
CARD2, CK-8, CK8, CYK8, K2C8, K8, KO
UniGene
Hs.533782
Gene Chromosome Location
Chr.12: 52897187-52949866 on Build GRCh38
Transcripts
NM_000224.2 NM_199187.1 NM_001256293.1 NR_045962.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1128552 c.11C>T p.T4I Chr.12: 52949184-52949184 on Build GRCh38
1128551 c.48G>A p.L16L Chr.12: 52949221-52949221 on Build GRCh38
1932135 c.13A>G p.T5A Chr.12: 52949186-52949186 on Build GRCh38
1128550 c.50G>A p.G17D Chr.12: 52949223-52949223 on Build GRCh38
3871921 c.83C>T p.P28L Chr.12: 52949256-52949256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368939913 Chr.12: 52949120-52949120 on Build GRCh38
rs750200705 Chr.12: 52949246-52949246 on Build GRCh38
rs775669589 Chr.12: 52948830-52948830 on Build GRCh38
rs1012004485 Chr.12: 52948825-52948825 on Build GRCh38
rs891320036 Chr.12: 52949111-52949111 on Build GRCh38
rs534114392 Chr.12: 52948941-52948941 on Build GRCh38
rs779623179 Chr.12: 52949152-52949153 on Build GRCh38
rs949911682 Chr.12: 52948998-52948998 on Build GRCh38
rs778594717 Chr.12: 52949269-52949269 on Build GRCh38
rs200362147 Chr.12: 52949158-52949158 on Build GRCh38
rs1007449580 Chr.12: 52949112-52949112 on Build GRCh38
rs147350452 Chr.12: 52949226-52949226 on Build GRCh38
rs79477652 Chr.12: 52948934-52948934 on Build GRCh38
rs766809169 Chr.12: 52949213-52949213 on Build GRCh38
rs374064321 Chr.12: 52949265-52949265 on Build GRCh38
rs1029793988 Chr.12: 52948846-52948846 on Build GRCh38
rs758566910 Chr.12: 52949126-52949126 on Build GRCh38
rs907711794 Chr.12: 52949043-52949043 on Build GRCh38
rs964352053 Chr.12: 52948935-52948935 on Build GRCh38
rs775547844 Chr.12: 52949193-52949193 on Build GRCh38
rs955723570 Chr.12: 52948891-52948891 on Build GRCh38
rs141066547 Chr.12: 52949242-52949242 on Build GRCh38
rs763632933 Chr.12: 52949151-52949151 on Build GRCh38
rs372353986 Chr.12: 52949123-52949123 on Build GRCh38
rs76301931 Chr.12: 52949184-52949184 on Build GRCh38
rs754487715 Chr.12: 52949171-52949176 on Build GRCh38
rs1018759767 Chr.12: 52948903-52948903 on Build GRCh38
rs777157164 Chr.12: 52949214-52949214 on Build GRCh38
rs780863496 Chr.12: 52949188-52949189 on Build GRCh38
rs373418568 Chr.12: 52949148-52949148 on Build GRCh38
rs935685098 Chr.12: 52949026-52949026 on Build GRCh38
rs755980643 Chr.12: 52949251-52949251 on Build GRCh38
rs1030132027 Chr.12: 52948969-52948969 on Build GRCh38
rs199517515 Chr.12: 52949142-52949142 on Build GRCh38
rs267607653 Chr.12: 52949159-52949159 on Build GRCh38
rs1025610446 Chr.12: 52949139-52949139 on Build GRCh38
rs920275137 Chr.12: 52949059-52949059 on Build GRCh38
rs774897980 Chr.12: 52949149-52949149 on Build GRCh38
rs906301801 Chr.12: 52949069-52949069 on Build GRCh38
rs376727372 Chr.12: 52949163-52949163 on Build GRCh38
rs753311520 Chr.12: 52949216-52949216 on Build GRCh38
rs78343594 Chr.12: 52949275-52949275 on Build GRCh38
rs1044175897 Chr.12: 52949017-52949017 on Build GRCh38
rs761560727 Chr.12: 52949212-52949212 on Build GRCh38
rs945689746 Chr.12: 52949199-52949199 on Build GRCh38
rs79476176 Chr.12: 52949223-52949223 on Build GRCh38
rs369198778 Chr.12: 52949178-52949178 on Build GRCh38
rs1010524694 Chr.12: 52949062-52949062 on Build GRCh38
rs892020611 Chr.12: 52948831-52948831 on Build GRCh38
rs74953757 Chr.12: 52949267-52949267 on Build GRCh38
rs1030479293 Chr.12: 52948979-52948979 on Build GRCh38
rs917125121 Chr.12: 52948947-52948947 on Build GRCh38
rs115941399 Chr.12: 52948896-52948896 on Build GRCh38
rs74379840 Chr.12: 52949256-52949256 on Build GRCh38
rs962725599 Chr.12: 52948907-52948907 on Build GRCh38
rs774095001 Chr.12: 52949153-52949153 on Build GRCh38
rs905598458 Chr.12: 52949023-52949023 on Build GRCh38
rs760023196 Chr.12: 52949215-52949215 on Build GRCh38
rs555437485 Chr.12: 52949013-52949013 on Build GRCh38
rs768280963 Chr.12: 52949206-52949206 on Build GRCh38
rs973259445 Chr.12: 52949056-52949056 on Build GRCh38
rs920524702 Chr.12: 52948924-52948924 on Build GRCh38
rs200298277 Chr.12: 52949138-52949138 on Build GRCh38
rs935714772 Chr.12: 52949164-52949164 on Build GRCh38
rs906718656 Chr.12: 52948841-52948841 on Build GRCh38
rs758542679 Chr.12: 52949169-52949169 on Build GRCh38
rs11551634 Chr.12: 52949250-52949250 on Build GRCh38
rs77825282 Chr.12: 52949252-52949252 on Build GRCh38
rs748718547 Chr.12: 52949204-52949204 on Build GRCh38
rs1044832297 Chr.12: 52949066-52949066 on Build GRCh38
rs762618085 Chr.12: 52949150-52949150 on Build GRCh38
rs769641289 Chr.12: 52949185-52949185 on Build GRCh38
rs940645676 Chr.12: 52949044-52949044 on Build GRCh38
rs376817240 Chr.12: 52949134-52949134 on Build GRCh38
rs545237524 Chr.12: 52949133-52949133 on Build GRCh38
rs751717313 Chr.12: 52949224-52949224 on Build GRCh38
rs898818722 Chr.12: 52948820-52948820 on Build GRCh38
rs1010369938 Chr.12: 52948839-52948839 on Build GRCh38
rs765308206 Chr.12: 52949166-52949166 on Build GRCh38
rs78514003 Chr.12: 52949249-52949249 on Build GRCh38
rs201934812 Chr.12: 52949155-52949155 on Build GRCh38
rs780920567 Chr.12: 52949230-52949230 on Build GRCh38
rs141170056 Chr.12: 52949179-52949179 on Build GRCh38
rs1003682897 Chr.12: 52948882-52948882 on Build GRCh38
rs7978102 Chr.12: 52949088-52949088 on Build GRCh38
rs974045746 Chr.12: 52948921-52948921 on Build GRCh38
rs528903225 Chr.12: 52949257-52949257 on Build GRCh38
rs756991398 Chr.12: 52949181-52949181 on Build GRCh38
rs201179094 Chr.12: 52949167-52949167 on Build GRCh38
rs751255280 Chr.12: 52949156-52949159 on Build GRCh38
rs80354424 Chr.12: 52949221-52949221 on Build GRCh38
rs1054017201 Chr.12: 52949058-52949058 on Build GRCh38
rs745822356 Chr.12: 52949147-52949147 on Build GRCh38
rs574125666 Chr.12: 52949060-52949060 on Build GRCh38

View Full Product Details