Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55028652-55029153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEUROD4
Entrez Gene ID
58158
Gene Name
neuronal differentiation 4
Gene Aliases
ATH-3, ATH3, Atoh3, MATH-3, MATH3, bHLHa4
UniGene
Hs.591024
Gene Chromosome Location
Chr.12: 55019945-55030017 on Build GRCh38
Transcripts
NM_021191.2

SNP Detail

SNP ID Chromosome Location
rs143102516 Chr.12: 55028704-55028704 on Build GRCh38
rs747532142 Chr.12: 55028725-55028725 on Build GRCh38
rs772261893 Chr.12: 55029115-55029115 on Build GRCh38
rs748323202 Chr.12: 55029088-55029088 on Build GRCh38
rs144737291 Chr.12: 55028888-55028889 on Build GRCh38
rs573948691 Chr.12: 55028951-55028951 on Build GRCh38
rs1022685391 Chr.12: 55028838-55028838 on Build GRCh38
rs897351706 Chr.12: 55029038-55029038 on Build GRCh38
rs545621122 Chr.12: 55029035-55029035 on Build GRCh38
rs115998690 Chr.12: 55028771-55028771 on Build GRCh38
rs746169956 Chr.12: 55028778-55028778 on Build GRCh38
rs186629634 Chr.12: 55028867-55028867 on Build GRCh38
rs866962102 Chr.12: 55029107-55029107 on Build GRCh38
rs112645813 Chr.12: 55028821-55028822 on Build GRCh38
rs1015735427 Chr.12: 55028683-55028683 on Build GRCh38
rs981248296 Chr.12: 55028846-55028846 on Build GRCh38
rs562806075 Chr.12: 55028985-55028985 on Build GRCh38
rs922663040 Chr.12: 55028907-55028907 on Build GRCh38
rs1049731417 Chr.12: 55028944-55028944 on Build GRCh38
rs914497894 Chr.12: 55028926-55028926 on Build GRCh38
rs998944661 Chr.12: 55028678-55028678 on Build GRCh38
rs894341178 Chr.12: 55029006-55029006 on Build GRCh38
rs990512338 Chr.12: 55028881-55028881 on Build GRCh38
rs386763221 Chr.12: 55028770-55028771 on Build GRCh38
rs934069571 Chr.12: 55028908-55028908 on Build GRCh38
rs1049080817 Chr.12: 55029069-55029069 on Build GRCh38
rs1038291978 Chr.12: 55029002-55029002 on Build GRCh38
rs951839470 Chr.12: 55028725-55028725 on Build GRCh38
rs149030113 Chr.12: 55028685-55028685 on Build GRCh38
rs540912483 Chr.12: 55028843-55028843 on Build GRCh38
rs544701324 Chr.12: 55028964-55028964 on Build GRCh38
rs897388520 Chr.12: 55028684-55028684 on Build GRCh38
rs568414859 Chr.12: 55029019-55029019 on Build GRCh38
rs563921958 Chr.12: 55029046-55029046 on Build GRCh38
rs934581892 Chr.12: 55028879-55028879 on Build GRCh38
rs868177106 Chr.12: 55029106-55029106 on Build GRCh38
rs539614056 Chr.12: 55028736-55028736 on Build GRCh38
rs964174392 Chr.12: 55028809-55028809 on Build GRCh38
rs957030772 Chr.12: 55029128-55029128 on Build GRCh38
rs138824901 Chr.12: 55028770-55028770 on Build GRCh38
rs895406445 Chr.12: 55029083-55029083 on Build GRCh38
rs558513594 Chr.12: 55028739-55028739 on Build GRCh38
rs956957426 Chr.12: 55028702-55028702 on Build GRCh38
rs932611387 Chr.12: 55028943-55028943 on Build GRCh38
rs530754793 Chr.12: 55028997-55028997 on Build GRCh38

View Full Product Details