Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57768873-57769377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL1
Entrez Gene ID
4234
Gene Name
methyltransferase like 1
Gene Aliases
C12orf1, TRM8, TRMT8, YDL201w
UniGene
Hs.42957
Gene Chromosome Location
Chr.12: 57768568-57772249 on Build GRCh38
Transcripts
NM_005371.5 NM_023033.3 XM_005268873.2 XM_017019305.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3737808 c.675+9G>C p.? Chr.12: 57769294-57769294 on Build GRCh38
6711575 c.769C>T p.R257* Chr.12: 57769058-57769058 on Build GRCh38
124416 c.718G>A p.E240K Chr.12: 57769109-57769109 on Build GRCh38
4043954 c.717G>T p.E239D Chr.12: 57769110-57769110 on Build GRCh38
6840841 c.787G>A p.V263I Chr.12: 57769040-57769040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911314496 Chr.12: 57769025-57769025 on Build GRCh38
rs996068793 Chr.12: 57769211-57769211 on Build GRCh38
rs772355553 Chr.12: 57769115-57769115 on Build GRCh38
rs201484091 Chr.12: 57769160-57769160 on Build GRCh38
rs978406943 Chr.12: 57769097-57769097 on Build GRCh38
rs930650442 Chr.12: 57769057-57769057 on Build GRCh38
rs534101281 Chr.12: 57768989-57768994 on Build GRCh38
rs374376984 Chr.12: 57768971-57768971 on Build GRCh38
rs200265154 Chr.12: 57768969-57768969 on Build GRCh38
rs762473406 Chr.12: 57769037-57769037 on Build GRCh38
rs200703167 Chr.12: 57769058-57769058 on Build GRCh38
rs933521736 Chr.12: 57769226-57769226 on Build GRCh38
rs764669573 Chr.12: 57769016-57769016 on Build GRCh38
rs77858092 Chr.12: 57769208-57769208 on Build GRCh38
rs752547385 Chr.12: 57769077-57769077 on Build GRCh38
rs562644428 Chr.12: 57769137-57769137 on Build GRCh38
rs759805058 Chr.12: 57769008-57769008 on Build GRCh38
rs564363792 Chr.12: 57769318-57769318 on Build GRCh38
rs776656881 Chr.12: 57769183-57769183 on Build GRCh38
rs779713965 Chr.12: 57768958-57768958 on Build GRCh38
rs370602083 Chr.12: 57769023-57769023 on Build GRCh38
rs778940217 Chr.12: 57769277-57769277 on Build GRCh38
rs768893540 Chr.12: 57769084-57769085 on Build GRCh38
rs550296848 Chr.12: 57768905-57768905 on Build GRCh38
rs917507482 Chr.12: 57768993-57768993 on Build GRCh38
rs761134567 Chr.12: 57769130-57769130 on Build GRCh38
rs767201145 Chr.12: 57769159-57769159 on Build GRCh38
rs774732743 Chr.12: 57769004-57769004 on Build GRCh38
rs754260658 Chr.12: 57769028-57769028 on Build GRCh38
rs775254763 Chr.12: 57769344-57769344 on Build GRCh38
rs140194153 Chr.12: 57769090-57769090 on Build GRCh38
rs768855649 Chr.12: 57769133-57769133 on Build GRCh38
rs975057178 Chr.12: 57769143-57769143 on Build GRCh38
rs187530858 Chr.12: 57769281-57769281 on Build GRCh38
rs370350550 Chr.12: 57768982-57768982 on Build GRCh38
rs368285948 Chr.12: 57769139-57769139 on Build GRCh38
rs374416908 Chr.12: 57769129-57769129 on Build GRCh38
rs747177585 Chr.12: 57769320-57769320 on Build GRCh38
rs777245297 Chr.12: 57769136-57769136 on Build GRCh38
rs944570396 Chr.12: 57769132-57769132 on Build GRCh38
rs745948904 Chr.12: 57769280-57769280 on Build GRCh38
rs760676072 Chr.12: 57769004-57769007 on Build GRCh38
rs199711056 Chr.12: 57769125-57769125 on Build GRCh38
rs993005230 Chr.12: 57768991-57768991 on Build GRCh38
rs983506626 Chr.12: 57769240-57769240 on Build GRCh38
rs753898222 Chr.12: 57769178-57769178 on Build GRCh38
rs754619955 Chr.12: 57769046-57769046 on Build GRCh38
rs145819242 Chr.12: 57769014-57769014 on Build GRCh38
rs770192581 Chr.12: 57769259-57769259 on Build GRCh38
rs1021926812 Chr.12: 57768938-57768938 on Build GRCh38
rs978216346 Chr.12: 57768959-57768959 on Build GRCh38
rs900411046 Chr.12: 57768909-57768909 on Build GRCh38
rs757251387 Chr.12: 57769252-57769252 on Build GRCh38
rs703842 Chr.12: 57768956-57768956 on Build GRCh38
rs760209741 Chr.12: 57769345-57769345 on Build GRCh38
rs770298061 Chr.12: 57769329-57769329 on Build GRCh38
rs758987779 Chr.12: 57769142-57769142 on Build GRCh38
rs777511586 Chr.12: 57768987-57768987 on Build GRCh38
rs773837002 Chr.12: 57769140-57769140 on Build GRCh38
rs746153660 Chr.12: 57769110-57769110 on Build GRCh38
rs558669975 Chr.12: 57769274-57769274 on Build GRCh38
rs766886635 Chr.12: 57769341-57769341 on Build GRCh38
rs749966401 Chr.12: 57768943-57768943 on Build GRCh38
rs776975890 Chr.12: 57769323-57769323 on Build GRCh38
rs199565614 Chr.12: 57768985-57768985 on Build GRCh38
rs779026533 Chr.12: 57768952-57768952 on Build GRCh38
rs763769990 Chr.12: 57769177-57769177 on Build GRCh38
rs568628584 Chr.12: 57768937-57768937 on Build GRCh38
rs903545347 Chr.12: 57769253-57769253 on Build GRCh38
rs907965088 Chr.12: 57769246-57769246 on Build GRCh38
rs753206974 Chr.12: 57768923-57768923 on Build GRCh38
rs774677732 Chr.12: 57768913-57768913 on Build GRCh38
rs528482986 Chr.12: 57769322-57769322 on Build GRCh38
rs376356409 Chr.12: 57769005-57769005 on Build GRCh38
rs773640593 Chr.12: 57769330-57769330 on Build GRCh38
rs751252853 Chr.12: 57769041-57769041 on Build GRCh38
rs765853566 Chr.12: 57769040-57769040 on Build GRCh38
rs770886393 Chr.12: 57769107-57769107 on Build GRCh38
rs749184039 Chr.12: 57769091-57769091 on Build GRCh38
rs886416572 Chr.12: 57768933-57768933 on Build GRCh38
rs201673849 Chr.12: 57768994-57768994 on Build GRCh38
rs755956057 Chr.12: 57769172-57769172 on Build GRCh38
rs751164864 Chr.12: 57768968-57768968 on Build GRCh38
rs900420861 Chr.12: 57769184-57769184 on Build GRCh38
rs368265719 Chr.12: 57769166-57769166 on Build GRCh38
rs147949367 Chr.12: 57769039-57769039 on Build GRCh38
rs368069805 Chr.12: 57769010-57769010 on Build GRCh38
rs771091009 Chr.12: 57769000-57769000 on Build GRCh38
rs149829610 Chr.12: 57769035-57769035 on Build GRCh38
rs549726296 Chr.12: 57768983-57768984 on Build GRCh38
rs771179004 Chr.12: 57769104-57769104 on Build GRCh38
rs532070975 Chr.12: 57768985-57768986 on Build GRCh38
rs747182796 Chr.12: 57768992-57768992 on Build GRCh38
rs372333605 Chr.12: 57769326-57769326 on Build GRCh38
rs761210130 Chr.12: 57768940-57768940 on Build GRCh38
rs1021982227 Chr.12: 57769242-57769242 on Build GRCh38
rs780905387 Chr.12: 57769049-57769049 on Build GRCh38
rs923415051 Chr.12: 57769176-57769176 on Build GRCh38
rs759986559 Chr.12: 57768917-57768917 on Build GRCh38
rs759115113 Chr.12: 57769335-57769335 on Build GRCh38
rs748280610 Chr.12: 57769203-57769203 on Build GRCh38

View Full Product Details