Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 62508197-62508700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MON2
Entrez Gene ID
23041
Gene Name
MON2 homolog, regulator of endosome-to-Golgi trafficking
Gene Aliases
-
UniGene
Hs.389378
Gene Chromosome Location
Chr.12: 62466817-62598956 on Build GRCh38
Transcripts
NM_001278469.1 NM_001278470.1 NM_001278471.1 NM_001278472.1 NM_015026.2 XM_017019041.1 XM_017019042.1 XM_017019043.1 XM_017019044.1 XM_017019045.1 XM_017019046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6567850 c.907G>A p.E303K Chr.12: 62508403-62508403 on Build GRCh38
4886888 c.935G>A p.R312H Chr.12: 62508431-62508431 on Build GRCh38
6567851 c.907G>A p.E303K Chr.12: 62508403-62508403 on Build GRCh38
4702864 c.884C>T p.T295I Chr.12: 62508380-62508380 on Build GRCh38
1178234 c.880A>G p.S294G Chr.12: 62508376-62508376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765357229 Chr.12: 62508513-62508513 on Build GRCh38
rs770285723 Chr.12: 62508531-62508532 on Build GRCh38
rs777815730 Chr.12: 62508244-62508244 on Build GRCh38
rs771950518 Chr.12: 62508465-62508465 on Build GRCh38
rs576710508 Chr.12: 62508612-62508612 on Build GRCh38
rs773797285 Chr.12: 62508457-62508457 on Build GRCh38
rs747836084 Chr.12: 62508296-62508296 on Build GRCh38
rs1036483591 Chr.12: 62508429-62508429 on Build GRCh38
rs377752807 Chr.12: 62508258-62508258 on Build GRCh38
rs551638999 Chr.12: 62508298-62508298 on Build GRCh38
rs531539589 Chr.12: 62508280-62508280 on Build GRCh38
rs777604480 Chr.12: 62508438-62508438 on Build GRCh38
rs750528726 Chr.12: 62508525-62508525 on Build GRCh38
rs772500447 Chr.12: 62508277-62508277 on Build GRCh38
rs376188370 Chr.12: 62508385-62508385 on Build GRCh38
rs545287509 Chr.12: 62508620-62508620 on Build GRCh38
rs774356034 Chr.12: 62508356-62508356 on Build GRCh38
rs532347079 Chr.12: 62508537-62508537 on Build GRCh38
rs760494195 Chr.12: 62508498-62508498 on Build GRCh38
rs755801169 Chr.12: 62508430-62508430 on Build GRCh38
rs558760761 Chr.12: 62508624-62508624 on Build GRCh38
rs192904006 Chr.12: 62508668-62508668 on Build GRCh38
rs373491785 Chr.12: 62508447-62508447 on Build GRCh38
rs747385917 Chr.12: 62508427-62508427 on Build GRCh38
rs762596564 Chr.12: 62508330-62508330 on Build GRCh38
rs549276653 Chr.12: 62508237-62508237 on Build GRCh38
rs1035898639 Chr.12: 62508588-62508588 on Build GRCh38
rs745802976 Chr.12: 62508462-62508462 on Build GRCh38
rs1013253050 Chr.12: 62508669-62508669 on Build GRCh38
rs553872896 Chr.12: 62508511-62508511 on Build GRCh38
rs762149035 Chr.12: 62508256-62508256 on Build GRCh38
rs758896492 Chr.12: 62508417-62508417 on Build GRCh38
rs758919793 Chr.12: 62508526-62508526 on Build GRCh38
rs759183178 Chr.12: 62508367-62508367 on Build GRCh38
rs770939562 Chr.12: 62508336-62508336 on Build GRCh38
rs571449218 Chr.12: 62508380-62508380 on Build GRCh38
rs779427475 Chr.12: 62508257-62508257 on Build GRCh38
rs776853315 Chr.12: 62508499-62508499 on Build GRCh38
rs1014617050 Chr.12: 62508621-62508621 on Build GRCh38
rs775323148 Chr.12: 62508469-62508469 on Build GRCh38
rs985417361 Chr.12: 62508306-62508306 on Build GRCh38
rs183705841 Chr.12: 62508387-62508387 on Build GRCh38
rs1010899454 Chr.12: 62508554-62508554 on Build GRCh38
rs188531907 Chr.12: 62508663-62508663 on Build GRCh38
rs889596479 Chr.12: 62508558-62508558 on Build GRCh38
rs969619163 Chr.12: 62508288-62508288 on Build GRCh38
rs556817946 Chr.12: 62508605-62508605 on Build GRCh38
rs535830648 Chr.12: 62508557-62508557 on Build GRCh38
rs572284270 Chr.12: 62508630-62508630 on Build GRCh38
rs142321979 Chr.12: 62508556-62508556 on Build GRCh38
rs7294404 Chr.12: 62508640-62508640 on Build GRCh38
rs150194547 Chr.12: 62508643-62508643 on Build GRCh38
rs935379024 Chr.12: 62508386-62508386 on Build GRCh38
rs374317455 Chr.12: 62508384-62508384 on Build GRCh38
rs753966844 Chr.12: 62508246-62508246 on Build GRCh38
rs780455923 Chr.12: 62508421-62508421 on Build GRCh38
rs764339428 Chr.12: 62508381-62508381 on Build GRCh38
rs190446545 Chr.12: 62508271-62508271 on Build GRCh38
rs868285744 Chr.12: 62508649-62508649 on Build GRCh38
rs748745817 Chr.12: 62508445-62508445 on Build GRCh38
rs1052338744 Chr.12: 62508283-62508283 on Build GRCh38
rs757025321 Chr.12: 62508564-62508564 on Build GRCh38
rs777040861 Chr.12: 62508241-62508241 on Build GRCh38
rs765675781 Chr.12: 62508394-62508394 on Build GRCh38
rs748941871 Chr.12: 62508559-62508559 on Build GRCh38
rs999587540 Chr.12: 62508651-62508651 on Build GRCh38
rs751936979 Chr.12: 62508529-62508529 on Build GRCh38
rs757729463 Chr.12: 62508255-62508255 on Build GRCh38
rs766838053 Chr.12: 62508528-62508528 on Build GRCh38
rs767336295 Chr.12: 62508375-62508375 on Build GRCh38
rs369425233 Chr.12: 62508411-62508411 on Build GRCh38
rs1030788232 Chr.12: 62508664-62508664 on Build GRCh38
rs769631514 Chr.12: 62508297-62508297 on Build GRCh38
rs749555936 Chr.12: 62508426-62508426 on Build GRCh38

View Full Product Details