Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64713815-64714337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNS
Entrez Gene ID
2799
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Gene Aliases
G6S
UniGene
Hs.334534
Gene Chromosome Location
Chr.12: 64713442-64759446 on Build GRCh38
Transcripts
NM_002076.3

SNP Detail

SNP ID Chromosome Location
rs979409758 Chr.12: 64714073-64714073 on Build GRCh38
rs962269307 Chr.12: 64713858-64713858 on Build GRCh38
rs534118009 Chr.12: 64713897-64713897 on Build GRCh38
rs529418480 Chr.12: 64714252-64714252 on Build GRCh38
rs886049762 Chr.12: 64714181-64714181 on Build GRCh38
rs538136456 Chr.12: 64713874-64713874 on Build GRCh38
rs867153534 Chr.12: 64714042-64714042 on Build GRCh38
rs1052696851 Chr.12: 64713873-64713873 on Build GRCh38
rs1028250406 Chr.12: 64714199-64714199 on Build GRCh38
rs554197342 Chr.12: 64714028-64714028 on Build GRCh38
rs770812812 Chr.12: 64714078-64714078 on Build GRCh38
rs949157944 Chr.12: 64714149-64714149 on Build GRCh38
rs942611699 Chr.12: 64713853-64713853 on Build GRCh38
rs749062392 Chr.12: 64714075-64714075 on Build GRCh38
rs995044057 Chr.12: 64714170-64714170 on Build GRCh38
rs950126830 Chr.12: 64714062-64714062 on Build GRCh38
rs937274607 Chr.12: 64714001-64714001 on Build GRCh38
rs1018300414 Chr.12: 64714020-64714020 on Build GRCh38
rs137943279 Chr.12: 64713990-64713990 on Build GRCh38
rs945943039 Chr.12: 64713964-64713964 on Build GRCh38
rs976506035 Chr.12: 64713955-64713955 on Build GRCh38
rs145906720 Chr.12: 64714103-64714103 on Build GRCh38
rs188119987 Chr.12: 64713932-64713932 on Build GRCh38
rs1027929587 Chr.12: 64713888-64713888 on Build GRCh38
rs201933913 Chr.12: 64714129-64714130 on Build GRCh38
rs750602693 Chr.12: 64713860-64713860 on Build GRCh38
rs994378631 Chr.12: 64713978-64713978 on Build GRCh38
rs925179626 Chr.12: 64713859-64713859 on Build GRCh38
rs577545481 Chr.12: 64713898-64713898 on Build GRCh38
rs898738577 Chr.12: 64713957-64713957 on Build GRCh38
rs976908243 Chr.12: 64714287-64714287 on Build GRCh38
rs547788908 Chr.12: 64714037-64714037 on Build GRCh38
rs1017920778 Chr.12: 64714234-64714234 on Build GRCh38
rs577212739 Chr.12: 64714085-64714085 on Build GRCh38
rs149013255 Chr.12: 64713854-64713854 on Build GRCh38
rs955486433 Chr.12: 64714002-64714002 on Build GRCh38
rs935238610 Chr.12: 64714113-64714113 on Build GRCh38
rs1007819273 Chr.12: 64714228-64714228 on Build GRCh38
rs1044766506 Chr.12: 64714230-64714230 on Build GRCh38
rs573404754 Chr.12: 64713958-64713958 on Build GRCh38
rs147105773 Chr.12: 64713900-64713900 on Build GRCh38
rs148217767 Chr.12: 64714203-64714203 on Build GRCh38
rs886049761 Chr.12: 64714001-64714001 on Build GRCh38
rs751642036 Chr.12: 64713947-64713947 on Build GRCh38
rs889348909 Chr.12: 64714217-64714217 on Build GRCh38
rs73323439 Chr.12: 64714117-64714117 on Build GRCh38
rs567930294 Chr.12: 64714294-64714294 on Build GRCh38
rs965076343 Chr.12: 64714244-64714244 on Build GRCh38
rs143396704 Chr.12: 64714006-64714006 on Build GRCh38
rs886049760 Chr.12: 64713954-64713954 on Build GRCh38
rs1036485392 Chr.12: 64714115-64714115 on Build GRCh38
rs904941118 Chr.12: 64714293-64714293 on Build GRCh38
rs547034900 Chr.12: 64714139-64714139 on Build GRCh38
rs895827504 Chr.12: 64714051-64714051 on Build GRCh38
rs533328232 Chr.12: 64714129-64714129 on Build GRCh38
rs549205368 Chr.12: 64713852-64713852 on Build GRCh38

View Full Product Details