Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 68294441-68294940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MDM1
Entrez Gene ID
56890
Gene Name
Mdm1 nuclear protein
Gene Aliases
-
UniGene
Hs.655702
Gene Chromosome Location
Chr.12: 68294566-68332381 on Build GRCh38
Transcripts
NM_001205028.1 NM_017440.4 XM_005269026.1 XM_017019646.1 XM_017019647.1 XM_017019648.1 XM_017019649.1 XM_017019650.1

SNP Detail

SNP ID Chromosome Location
rs899329601 Chr.12: 68294729-68294729 on Build GRCh38
rs906547585 Chr.12: 68294624-68294624 on Build GRCh38
rs1009173004 Chr.12: 68294869-68294869 on Build GRCh38
rs985498854 Chr.12: 68294507-68294507 on Build GRCh38
rs115983270 Chr.12: 68294485-68294485 on Build GRCh38
rs977790945 Chr.12: 68294887-68294887 on Build GRCh38
rs948004628 Chr.12: 68294616-68294616 on Build GRCh38
rs912233698 Chr.12: 68294509-68294509 on Build GRCh38
rs1053703662 Chr.12: 68294578-68294578 on Build GRCh38
rs773901764 Chr.12: 68294602-68294602 on Build GRCh38
rs1054499227 Chr.12: 68294775-68294775 on Build GRCh38
rs748128790 Chr.12: 68294872-68294872 on Build GRCh38
rs957889025 Chr.12: 68294897-68294897 on Build GRCh38
rs981543084 Chr.12: 68294706-68294706 on Build GRCh38
rs181789363 Chr.12: 68294684-68294684 on Build GRCh38
rs569891729 Chr.12: 68294548-68294548 on Build GRCh38
rs1004958598 Chr.12: 68294625-68294625 on Build GRCh38
rs368245709 Chr.12: 68294800-68294800 on Build GRCh38
rs907710705 Chr.12: 68294646-68294646 on Build GRCh38
rs563801828 Chr.12: 68294752-68294752 on Build GRCh38
rs541366212 Chr.12: 68294650-68294650 on Build GRCh38
rs780781787 Chr.12: 68294638-68294638 on Build GRCh38
rs891310136 Chr.12: 68294905-68294905 on Build GRCh38
rs965041042 Chr.12: 68294875-68294875 on Build GRCh38
rs974407771 Chr.12: 68294599-68294599 on Build GRCh38
rs995450536 Chr.12: 68294771-68294771 on Build GRCh38
rs547124945 Chr.12: 68294473-68294473 on Build GRCh38
rs537280408 Chr.12: 68294566-68294566 on Build GRCh38
rs557902680 Chr.12: 68294519-68294519 on Build GRCh38
rs893338306 Chr.12: 68294826-68294826 on Build GRCh38
rs531521113 Chr.12: 68294833-68294833 on Build GRCh38
rs543684346 Chr.12: 68294865-68294865 on Build GRCh38
rs36012261 Chr.12: 68294810-68294811 on Build GRCh38
rs545543939 Chr.12: 68294709-68294709 on Build GRCh38
rs1030744799 Chr.12: 68294891-68294891 on Build GRCh38
rs565723759 Chr.12: 68294484-68294484 on Build GRCh38
rs555956216 Chr.12: 68294620-68294620 on Build GRCh38
rs937312802 Chr.12: 68294759-68294759 on Build GRCh38
rs953719781 Chr.12: 68294792-68294792 on Build GRCh38
rs1029172861 Chr.12: 68294785-68294785 on Build GRCh38
rs953732398 Chr.12: 68294492-68294492 on Build GRCh38
rs555629185 Chr.12: 68294575-68294575 on Build GRCh38
rs1027518993 Chr.12: 68294600-68294601 on Build GRCh38
rs553099309 Chr.12: 68294667-68294667 on Build GRCh38
rs936530539 Chr.12: 68294576-68294576 on Build GRCh38

View Full Product Details