Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 91156983-91157494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCN
Entrez Gene ID
1634
Gene Name
decorin
Gene Aliases
CSCD, DSPG2, PG40, PGII, PGS2, SLRR1B
UniGene
Hs.156316
Gene Chromosome Location
Chr.12: 91143277-91183124 on Build GRCh38
Transcripts
NM_001920.4 NM_133503.3 NM_133504.3 NM_133505.3 NM_133506.3 NM_133507.3 XM_005268693.1 XM_006719270.1 XM_017018917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4045359 c.612C>T p.Y204Y Chr.12: 91157115-91157115 on Build GRCh38
944388 c.606C>A p.L202L Chr.12: 91157121-91157121 on Build GRCh38
4045358 c.616C>T p.R206C Chr.12: 91157111-91157111 on Build GRCh38
1706025 c.593G>A p.G198E Chr.12: 91157134-91157134 on Build GRCh38
3359959 c.597G>T p.M199I Chr.12: 91157130-91157130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761253178 Chr.12: 91157125-91157125 on Build GRCh38
rs375956828 Chr.12: 91157025-91157025 on Build GRCh38
rs958279810 Chr.12: 91157336-91157336 on Build GRCh38
rs759169003 Chr.12: 91157028-91157028 on Build GRCh38
rs748901311 Chr.12: 91157110-91157110 on Build GRCh38
rs769180321 Chr.12: 91157221-91157221 on Build GRCh38
rs777158190 Chr.12: 91157225-91157225 on Build GRCh38
rs370036360 Chr.12: 91157065-91157065 on Build GRCh38
rs3138246 Chr.12: 91157052-91157052 on Build GRCh38
rs142198841 Chr.12: 91157445-91157445 on Build GRCh38
rs972016650 Chr.12: 91157195-91157195 on Build GRCh38
rs759359940 Chr.12: 91157337-91157337 on Build GRCh38
rs1045542873 Chr.12: 91157072-91157072 on Build GRCh38
rs779649768 Chr.12: 91157211-91157211 on Build GRCh38
rs765248137 Chr.12: 91157068-91157068 on Build GRCh38
rs748505613 Chr.12: 91157239-91157239 on Build GRCh38
rs997946920 Chr.12: 91157399-91157399 on Build GRCh38
rs981172580 Chr.12: 91157440-91157440 on Build GRCh38
rs887702686 Chr.12: 91157109-91157109 on Build GRCh38
rs1014711304 Chr.12: 91157142-91157142 on Build GRCh38
rs879186231 Chr.12: 91157007-91157008 on Build GRCh38
rs757186299 Chr.12: 91157201-91157201 on Build GRCh38
rs764534812 Chr.12: 91157061-91157061 on Build GRCh38
rs909842116 Chr.12: 91157316-91157316 on Build GRCh38
rs147765043 Chr.12: 91157172-91157172 on Build GRCh38
rs753654634 Chr.12: 91157198-91157198 on Build GRCh38
rs988274947 Chr.12: 91157341-91157341 on Build GRCh38
rs777073492 Chr.12: 91157158-91157158 on Build GRCh38
rs772675673 Chr.12: 91157032-91157032 on Build GRCh38
rs925548866 Chr.12: 91157023-91157023 on Build GRCh38
rs762209864 Chr.12: 91157166-91157166 on Build GRCh38
rs892115529 Chr.12: 91157423-91157423 on Build GRCh38
rs199553162 Chr.12: 91157030-91157030 on Build GRCh38
rs776132393 Chr.12: 91157033-91157033 on Build GRCh38
rs201740609 Chr.12: 91157089-91157089 on Build GRCh38
rs184220156 Chr.12: 91157026-91157026 on Build GRCh38
rs1032572633 Chr.12: 91157294-91157294 on Build GRCh38
rs745531761 Chr.12: 91157203-91157203 on Build GRCh38
rs1052211711 Chr.12: 91157420-91157420 on Build GRCh38
rs777628468 Chr.12: 91157108-91157108 on Build GRCh38
rs888264658 Chr.12: 91157010-91157010 on Build GRCh38
rs778696927 Chr.12: 91157202-91157202 on Build GRCh38
rs745368707 Chr.12: 91157112-91157112 on Build GRCh38
rs77520130 Chr.12: 91157047-91157047 on Build GRCh38
rs796178231 Chr.12: 91157019-91157019 on Build GRCh38
rs561316543 Chr.12: 91157371-91157371 on Build GRCh38
rs60125162 Chr.12: 91157008-91157008 on Build GRCh38
rs541473594 Chr.12: 91157310-91157310 on Build GRCh38
rs575760721 Chr.12: 91157356-91157356 on Build GRCh38
rs752509888 Chr.12: 91157179-91157179 on Build GRCh38
rs775805766 Chr.12: 91157117-91157117 on Build GRCh38
rs573167363 Chr.12: 91157212-91157212 on Build GRCh38
rs980996530 Chr.12: 91157118-91157118 on Build GRCh38
rs960780189 Chr.12: 91157192-91157192 on Build GRCh38
rs1030689445 Chr.12: 91157071-91157071 on Build GRCh38
rs1039548349 Chr.12: 91157342-91157342 on Build GRCh38
rs914067824 Chr.12: 91157406-91157406 on Build GRCh38
rs149901877 Chr.12: 91157178-91157178 on Build GRCh38
rs780984671 Chr.12: 91157102-91157102 on Build GRCh38
rs758510607 Chr.12: 91157095-91157095 on Build GRCh38
rs144972644 Chr.12: 91157181-91157181 on Build GRCh38
rs555705830 Chr.12: 91157167-91157167 on Build GRCh38
rs139057073 Chr.12: 91157173-91157173 on Build GRCh38
rs1010613436 Chr.12: 91157431-91157431 on Build GRCh38
rs373676217 Chr.12: 91157106-91157106 on Build GRCh38
rs953386337 Chr.12: 91157270-91157270 on Build GRCh38
rs969674625 Chr.12: 91157466-91157466 on Build GRCh38
rs752737077 Chr.12: 91157105-91157105 on Build GRCh38
rs867691361 Chr.12: 91157285-91157285 on Build GRCh38
rs912985958 Chr.12: 91157397-91157413 on Build GRCh38
rs757783232 Chr.12: 91157204-91157204 on Build GRCh38
rs1048282418 Chr.12: 91157304-91157304 on Build GRCh38
rs1558593 Chr.12: 91157059-91157059 on Build GRCh38
rs768866022 Chr.12: 91157143-91157143 on Build GRCh38
rs970926090 Chr.12: 91157434-91157434 on Build GRCh38
rs763988753 Chr.12: 91157182-91157182 on Build GRCh38
rs771673142 Chr.12: 91157115-91157115 on Build GRCh38
rs778750338 Chr.12: 91157111-91157111 on Build GRCh38
rs1008000641 Chr.12: 91157282-91157282 on Build GRCh38
rs368745396 Chr.12: 91157240-91157240 on Build GRCh38
rs778965623 Chr.12: 91157115-91157115 on Build GRCh38

View Full Product Details