Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 96001043-96001553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTA4H
Entrez Gene ID
4048
Gene Name
leukotriene A4 hydrolase
Gene Aliases
-
UniGene
Hs.524648
Gene Chromosome Location
Chr.12: 96000753-96043520 on Build GRCh38
Transcripts
NM_000895.2 NM_001256643.1 NM_001256644.1 NR_132659.1 XM_005268871.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7139496 c.1735G>T p.D579Y Chr.12: 96001090-96001090 on Build GRCh38
5922980 c.1744C>T p.H582Y Chr.12: 96001081-96001081 on Build GRCh38
6704774 c.1759C>T p.R587* Chr.12: 96001066-96001066 on Build GRCh38
1214063 c.1756G>A p.V586I Chr.12: 96001069-96001069 on Build GRCh38
3793054 c.1745A>G p.H582R Chr.12: 96001080-96001080 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs909549006 Chr.12: 96001397-96001397 on Build GRCh38
rs909610754 Chr.12: 96001157-96001157 on Build GRCh38
rs1048113079 Chr.12: 96001340-96001340 on Build GRCh38
rs529013701 Chr.12: 96001394-96001394 on Build GRCh38
rs966688488 Chr.12: 96001421-96001421 on Build GRCh38
rs754858865 Chr.12: 96001465-96001465 on Build GRCh38
rs369712805 Chr.12: 96001371-96001371 on Build GRCh38
rs999944484 Chr.12: 96001073-96001073 on Build GRCh38
rs755759231 Chr.12: 96001072-96001072 on Build GRCh38
rs541706119 Chr.12: 96001178-96001181 on Build GRCh38
rs982276709 Chr.12: 96001322-96001322 on Build GRCh38
rs1029991148 Chr.12: 96001467-96001467 on Build GRCh38
rs563736079 Chr.12: 96001225-96001225 on Build GRCh38
rs776601875 Chr.12: 96001139-96001139 on Build GRCh38
rs535853377 Chr.12: 96001246-96001246 on Build GRCh38
rs748793205 Chr.12: 96001080-96001080 on Build GRCh38
rs201072252 Chr.12: 96001121-96001121 on Build GRCh38
rs985680499 Chr.12: 96001505-96001505 on Build GRCh38
rs200755127 Chr.12: 96001079-96001079 on Build GRCh38
rs561600218 Chr.12: 96001338-96001338 on Build GRCh38
rs771347255 Chr.12: 96001089-96001089 on Build GRCh38
rs1050916456 Chr.12: 96001497-96001497 on Build GRCh38
rs941142745 Chr.12: 96001405-96001405 on Build GRCh38
rs909735929 Chr.12: 96001519-96001519 on Build GRCh38
rs374987362 Chr.12: 96001330-96001330 on Build GRCh38
rs779336106 Chr.12: 96001094-96001094 on Build GRCh38
rs1055325093 Chr.12: 96001336-96001336 on Build GRCh38
rs954016353 Chr.12: 96001468-96001468 on Build GRCh38
rs762717809 Chr.12: 96001142-96001142 on Build GRCh38
rs760858860 Chr.12: 96001125-96001125 on Build GRCh38
rs899558758 Chr.12: 96001451-96001451 on Build GRCh38
rs531162252 Chr.12: 96001287-96001287 on Build GRCh38
rs893872669 Chr.12: 96001353-96001353 on Build GRCh38
rs751532654 Chr.12: 96001359-96001359 on Build GRCh38
rs752358414 Chr.12: 96001071-96001071 on Build GRCh38
rs560320858 Chr.12: 96001143-96001143 on Build GRCh38
rs765344927 Chr.12: 96001129-96001132 on Build GRCh38
rs574092921 Chr.12: 96001124-96001124 on Build GRCh38
rs933832254 Chr.12: 96001480-96001480 on Build GRCh38
rs574334193 Chr.12: 96001127-96001131 on Build GRCh38
rs145248834 Chr.12: 96001409-96001409 on Build GRCh38
rs181102738 Chr.12: 96001163-96001163 on Build GRCh38
rs541629909 Chr.12: 96001138-96001138 on Build GRCh38
rs930208712 Chr.12: 96001368-96001368 on Build GRCh38
rs929512299 Chr.12: 96001240-96001242 on Build GRCh38
rs888129931 Chr.12: 96001447-96001447 on Build GRCh38
rs777197120 Chr.12: 96001074-96001074 on Build GRCh38
rs772494802 Chr.12: 96001123-96001123 on Build GRCh38
rs1047090476 Chr.12: 96001300-96001300 on Build GRCh38
rs1036676140 Chr.12: 96001221-96001221 on Build GRCh38
rs145327577 Chr.12: 96001223-96001223 on Build GRCh38
rs879084865 Chr.12: 96001244-96001244 on Build GRCh38
rs113724669 Chr.12: 96001196-96001196 on Build GRCh38

View Full Product Details