Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102196567-102197082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARPBP
Entrez Gene ID
55010
Gene Name
PARP1 binding protein
Gene Aliases
AROM, C12orf48, PARI
UniGene
Hs.330663
Gene Chromosome Location
Chr.12: 102120171-102197520 on Build GRCh38
Transcripts
NM_001319988.1 NM_001319993.1 NM_001319994.1 NM_017915.4 NR_135123.1 XM_011538509.1 XM_011538514.2 XM_011538515.1 XM_017019534.1 XM_017019535.1 XM_017019536.1 XM_017019537.1 XM_017019538.1 XM_017019539.1 XM_017019540.1 XM_017019542.1 XM_017019543.1 XM_017019544.1 NM_002674.3 XM_017019483.1

Gene Symbol
PMCH
Entrez Gene ID
5367
Gene Name
pro-melanin concentrating hormone
Gene Aliases
MCH, ppMCH
UniGene
Hs.707990
Gene Chromosome Location
Chr.12: 102196459-102199540 on Build GRCh38
Transcripts
NM_001319988.1 NM_001319993.1 NM_001319994.1 NM_017915.4 NR_135123.1 XM_011538509.1 XM_011538514.2 XM_011538515.1 XM_017019534.1 XM_017019535.1 XM_017019536.1 XM_017019537.1 XM_017019538.1 XM_017019539.1 XM_017019540.1 XM_017019542.1 XM_017019543.1 XM_017019544.1 NM_002674.3 XM_017019483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
125463 c.402T>A p.D134E Chr.12: 102197019-102197019 on Build GRCh38
4454036 c.393A>C p.E131D Chr.12: 102197028-102197028 on Build GRCh38
691587 c.462G>A p.M154I Chr.12: 102196688-102196688 on Build GRCh38
3980706 c.411C>G p.N137K Chr.12: 102197010-102197010 on Build GRCh38
5753840 c.350A>T p.E117V Chr.12: 102197071-102197071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772349415 Chr.12: 102196686-102196686 on Build GRCh38
rs529197874 Chr.12: 102196610-102196610 on Build GRCh38
rs747308628 Chr.12: 102196991-102196991 on Build GRCh38
rs754440818 Chr.12: 102196711-102196711 on Build GRCh38
rs778904142 Chr.12: 102196951-102196951 on Build GRCh38
rs530771530 Chr.12: 102196993-102196993 on Build GRCh38
rs757313851 Chr.12: 102196948-102196948 on Build GRCh38
rs753141212 Chr.12: 102196785-102196785 on Build GRCh38
rs765629077 Chr.12: 102197043-102197044 on Build GRCh38
rs905199758 Chr.12: 102196680-102196680 on Build GRCh38
rs761128374 Chr.12: 102196692-102196692 on Build GRCh38
rs766088615 Chr.12: 102196743-102196743 on Build GRCh38
rs751263754 Chr.12: 102197037-102197037 on Build GRCh38
rs761407744 Chr.12: 102196717-102196717 on Build GRCh38
rs956578881 Chr.12: 102196769-102196769 on Build GRCh38
rs374157468 Chr.12: 102196649-102196649 on Build GRCh38
rs1036901529 Chr.12: 102197042-102197042 on Build GRCh38
rs192810118 Chr.12: 102196850-102196850 on Build GRCh38
rs754724347 Chr.12: 102197048-102197048 on Build GRCh38
rs1025697443 Chr.12: 102196823-102196823 on Build GRCh38
rs772142314 Chr.12: 102196964-102196964 on Build GRCh38
rs528570123 Chr.12: 102196877-102196877 on Build GRCh38
rs757705217 Chr.12: 102196744-102196745 on Build GRCh38
rs200281750 Chr.12: 102196672-102196672 on Build GRCh38
rs749691464 Chr.12: 102196876-102196876 on Build GRCh38
rs764692903 Chr.12: 102196742-102196743 on Build GRCh38
rs764490356 Chr.12: 102196709-102196709 on Build GRCh38
rs969954286 Chr.12: 102196854-102196854 on Build GRCh38
rs752266364 Chr.12: 102196739-102196739 on Build GRCh38
rs781075906 Chr.12: 102196931-102196931 on Build GRCh38
rs1009777132 Chr.12: 102196820-102196820 on Build GRCh38
rs960724715 Chr.12: 102196831-102196831 on Build GRCh38
rs559560894 Chr.12: 102196872-102196872 on Build GRCh38
rs138736999 Chr.12: 102196980-102196980 on Build GRCh38
rs980009165 Chr.12: 102196859-102196859 on Build GRCh38
rs746825153 Chr.12: 102196635-102196635 on Build GRCh38
rs578029835 Chr.12: 102196667-102196667 on Build GRCh38
rs994664978 Chr.12: 102196809-102196809 on Build GRCh38
rs940870183 Chr.12: 102197041-102197041 on Build GRCh38
rs992138879 Chr.12: 102196834-102196834 on Build GRCh38
rs551401488 Chr.12: 102196958-102196958 on Build GRCh38
rs377553179 Chr.12: 102196658-102196658 on Build GRCh38
rs759732671 Chr.12: 102196676-102196676 on Build GRCh38
rs917207923 Chr.12: 102196844-102196844 on Build GRCh38
rs150610425 Chr.12: 102196766-102196766 on Build GRCh38
rs781675279 Chr.12: 102196695-102196695 on Build GRCh38
rs141233921 Chr.12: 102197016-102197016 on Build GRCh38
rs187611964 Chr.12: 102196810-102196810 on Build GRCh38
rs1017514123 Chr.12: 102196775-102196775 on Build GRCh38
rs1038580620 Chr.12: 102197038-102197038 on Build GRCh38
rs767355648 Chr.12: 102197053-102197053 on Build GRCh38
rs138083104 Chr.12: 102197026-102197026 on Build GRCh38
rs11558437 Chr.12: 102197007-102197007 on Build GRCh38
rs912968384 Chr.12: 102196960-102196960 on Build GRCh38
rs111468854 Chr.12: 102196871-102196871 on Build GRCh38
rs764900337 Chr.12: 102196724-102196724 on Build GRCh38
rs774549475 Chr.12: 102196675-102196675 on Build GRCh38
rs199546666 Chr.12: 102196749-102196749 on Build GRCh38
rs750170320 Chr.12: 102196607-102196607 on Build GRCh38
rs779658098 Chr.12: 102196611-102196611 on Build GRCh38
rs769213731 Chr.12: 102197000-102197000 on Build GRCh38
rs748708182 Chr.12: 102197039-102197039 on Build GRCh38
rs754850490 Chr.12: 102196924-102196924 on Build GRCh38
rs762546638 Chr.12: 102197034-102197034 on Build GRCh38
rs543740530 Chr.12: 102196737-102196737 on Build GRCh38
rs768941642 Chr.12: 102196945-102196945 on Build GRCh38
rs150362881 Chr.12: 102197022-102197022 on Build GRCh38
rs369329505 Chr.12: 102196942-102196942 on Build GRCh38
rs975325292 Chr.12: 102196878-102196878 on Build GRCh38
rs962877827 Chr.12: 102196795-102196795 on Build GRCh38
rs757922645 Chr.12: 102196740-102196740 on Build GRCh38

View Full Product Details