Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 105189900-105190398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APPL2
Entrez Gene ID
55198
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
Gene Aliases
DIP13B
UniGene
Hs.506603
Gene Chromosome Location
Chr.12: 105173297-105236230 on Build GRCh38
Transcripts
NM_001251904.1 NM_001251905.1 NM_018171.3 XM_006719472.1 XM_011538530.2 XM_011538531.2 XM_011538532.2 XM_017019551.1 XM_017019552.1 XM_017019553.1 XM_017019554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2064882 c.1372G>C p.A458P Chr.12: 105190025-105190025 on Build GRCh38
691481 c.1321G>C p.E441Q Chr.12: 105190076-105190076 on Build GRCh38
5904379 c.1295A>C p.K432T Chr.12: 105190102-105190102 on Build GRCh38
3752860 c.1298C>T p.A433V Chr.12: 105190099-105190099 on Build GRCh38
238823 c.1364T>C p.V455A Chr.12: 105190033-105190033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768318729 Chr.12: 105190252-105190252 on Build GRCh38
rs771971297 Chr.12: 105190363-105190363 on Build GRCh38
rs977070238 Chr.12: 105190019-105190019 on Build GRCh38
rs568041214 Chr.12: 105190262-105190262 on Build GRCh38
rs144258618 Chr.12: 105189958-105189958 on Build GRCh38
rs771272779 Chr.12: 105190070-105190070 on Build GRCh38
rs148713024 Chr.12: 105190343-105190343 on Build GRCh38
rs769255127 Chr.12: 105190198-105190198 on Build GRCh38
rs199959711 Chr.12: 105190005-105190005 on Build GRCh38
rs753935134 Chr.12: 105190147-105190147 on Build GRCh38
rs769178183 Chr.12: 105190176-105190176 on Build GRCh38
rs762768742 Chr.12: 105190107-105190107 on Build GRCh38
rs747539200 Chr.12: 105190173-105190173 on Build GRCh38
rs752141990 Chr.12: 105189942-105189942 on Build GRCh38
rs747722750 Chr.12: 105190309-105190309 on Build GRCh38
rs767251315 Chr.12: 105190134-105190134 on Build GRCh38
rs753450244 Chr.12: 105190046-105190046 on Build GRCh38
rs749454237 Chr.12: 105190024-105190024 on Build GRCh38
rs769870095 Chr.12: 105190020-105190020 on Build GRCh38
rs766932250 Chr.12: 105190165-105190165 on Build GRCh38
rs758329938 Chr.12: 105189994-105189994 on Build GRCh38
rs376711138 Chr.12: 105190065-105190065 on Build GRCh38
rs1000601188 Chr.12: 105190195-105190195 on Build GRCh38
rs775110087 Chr.12: 105190031-105190031 on Build GRCh38
rs867942790 Chr.12: 105190119-105190119 on Build GRCh38
rs576659482 Chr.12: 105190190-105190190 on Build GRCh38
rs777888513 Chr.12: 105190022-105190022 on Build GRCh38
rs761495598 Chr.12: 105190100-105190100 on Build GRCh38
rs977503369 Chr.12: 105189961-105189961 on Build GRCh38
rs375871461 Chr.12: 105189944-105189944 on Build GRCh38
rs73179996 Chr.12: 105189969-105189969 on Build GRCh38
rs779456671 Chr.12: 105190075-105190075 on Build GRCh38
rs757599181 Chr.12: 105190149-105190149 on Build GRCh38
rs750188627 Chr.12: 105190053-105190053 on Build GRCh38
rs200542741 Chr.12: 105190102-105190102 on Build GRCh38
rs764145042 Chr.12: 105190138-105190138 on Build GRCh38
rs761189842 Chr.12: 105190092-105190092 on Build GRCh38
rs149910163 Chr.12: 105190063-105190063 on Build GRCh38
rs780457933 Chr.12: 105190172-105190172 on Build GRCh38
rs544933408 Chr.12: 105189949-105189949 on Build GRCh38
rs560274275 Chr.12: 105190027-105190027 on Build GRCh38
rs760184598 Chr.12: 105190028-105190028 on Build GRCh38
rs763628746 Chr.12: 105189956-105189956 on Build GRCh38
rs199855187 Chr.12: 105190025-105190025 on Build GRCh38
rs778625962 Chr.12: 105189980-105189980 on Build GRCh38
rs200578180 Chr.12: 105190097-105190097 on Build GRCh38
rs1049462170 Chr.12: 105190199-105190199 on Build GRCh38
rs781375212 Chr.12: 105189995-105189995 on Build GRCh38
rs371085355 Chr.12: 105190050-105190050 on Build GRCh38
rs371012317 Chr.12: 105190089-105190089 on Build GRCh38
rs2272495 Chr.12: 105190099-105190099 on Build GRCh38
rs369034385 Chr.12: 105190041-105190041 on Build GRCh38
rs768411686 Chr.12: 105189986-105189986 on Build GRCh38
rs958568591 Chr.12: 105189926-105189926 on Build GRCh38
rs138410860 Chr.12: 105190095-105190096 on Build GRCh38
rs754928718 Chr.12: 105190131-105190131 on Build GRCh38
rs377078499 Chr.12: 105189992-105189992 on Build GRCh38
rs773817389 Chr.12: 105190091-105190091 on Build GRCh38
rs562507472 Chr.12: 105190064-105190064 on Build GRCh38
rs746211065 Chr.12: 105190080-105190080 on Build GRCh38
rs772803222 Chr.12: 105190105-105190105 on Build GRCh38
rs977220825 Chr.12: 105190374-105190374 on Build GRCh38
rs747681991 Chr.12: 105190196-105190196 on Build GRCh38
rs201971126 Chr.12: 105190040-105190040 on Build GRCh38
rs766131370 Chr.12: 105190115-105190115 on Build GRCh38
rs758381619 Chr.12: 105190054-105190054 on Build GRCh38
rs746044580 Chr.12: 105190160-105190160 on Build GRCh38
rs772673476 Chr.12: 105190030-105190030 on Build GRCh38
rs762967345 Chr.12: 105189975-105189976 on Build GRCh38
rs143474178 Chr.12: 105189932-105189932 on Build GRCh38
rs773173237 Chr.12: 105189945-105189946 on Build GRCh38
rs751464567 Chr.12: 105190116-105190116 on Build GRCh38
rs201323570 Chr.12: 105190086-105190086 on Build GRCh38
rs375608143 Chr.12: 105190062-105190062 on Build GRCh38
rs758707220 Chr.12: 105190163-105190163 on Build GRCh38
rs765086628 Chr.12: 105190051-105190051 on Build GRCh38
rs779296120 Chr.12: 105190156-105190156 on Build GRCh38
rs866975263 Chr.12: 105189990-105189990 on Build GRCh38
rs570052615 Chr.12: 105190368-105190368 on Build GRCh38
rs748154763 Chr.12: 105189996-105189996 on Build GRCh38

View Full Product Details