Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113316447-113316954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC8B1
Entrez Gene ID
80024
Gene Name
solute carrier family 8 member B1
Gene Aliases
NCKX6, NCLX, SLC24A6
UniGene
Hs.286194
Gene Chromosome Location
Chr.12: 113298766-113335142 on Build GRCh38
Transcripts
NM_024959.2 XM_006719607.2 XM_011538749.2 XM_011538750.2 XM_011538752.2 XM_017019978.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6780463 c.902C>T p.T301M Chr.12: 113316617-113316617 on Build GRCh38
252817 c.919C>T p.R307W Chr.12: 113316600-113316600 on Build GRCh38
6780462 c.903G>A p.T301T Chr.12: 113316616-113316616 on Build GRCh38
430400 c.885C>T p.F295F Chr.12: 113316634-113316634 on Build GRCh38
5970523 c.867T>C p.D289D Chr.12: 113316652-113316652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140152775 Chr.12: 113316660-113316660 on Build GRCh38
rs894270512 Chr.12: 113316729-113316729 on Build GRCh38
rs199723765 Chr.12: 113316586-113316586 on Build GRCh38
rs544145829 Chr.12: 113316767-113316767 on Build GRCh38
rs775526957 Chr.12: 113316903-113316903 on Build GRCh38
rs1014355807 Chr.12: 113316769-113316769 on Build GRCh38
rs78925531 Chr.12: 113316682-113316682 on Build GRCh38
rs770767646 Chr.12: 113316693-113316693 on Build GRCh38
rs774510628 Chr.12: 113316775-113316775 on Build GRCh38
rs1054261447 Chr.12: 113316650-113316650 on Build GRCh38
rs781091225 Chr.12: 113316599-113316599 on Build GRCh38
rs755090268 Chr.12: 113316598-113316598 on Build GRCh38
rs780392077 Chr.12: 113316490-113316490 on Build GRCh38
rs760243661 Chr.12: 113316644-113316644 on Build GRCh38
rs375299683 Chr.12: 113316686-113316686 on Build GRCh38
rs1034877881 Chr.12: 113316469-113316469 on Build GRCh38
rs77481393 Chr.12: 113316887-113316887 on Build GRCh38
rs986301203 Chr.12: 113316645-113316645 on Build GRCh38
rs748301456 Chr.12: 113316536-113316536 on Build GRCh38
rs746770728 Chr.12: 113316628-113316629 on Build GRCh38
rs766581444 Chr.12: 113316667-113316667 on Build GRCh38
rs986862643 Chr.12: 113316842-113316842 on Build GRCh38
rs972050520 Chr.12: 113316837-113316837 on Build GRCh38
rs1036756660 Chr.12: 113316893-113316893 on Build GRCh38
rs200679896 Chr.12: 113316707-113316707 on Build GRCh38
rs778536727 Chr.12: 113316701-113316701 on Build GRCh38
rs1027879525 Chr.12: 113316700-113316700 on Build GRCh38
rs1031924740 Chr.12: 113316776-113316776 on Build GRCh38
rs775561138 Chr.12: 113316706-113316706 on Build GRCh38
rs758828313 Chr.12: 113316594-113316594 on Build GRCh38
rs115328394 Chr.12: 113316468-113316468 on Build GRCh38
rs756184409 Chr.12: 113316691-113316691 on Build GRCh38
rs201484722 Chr.12: 113316708-113316708 on Build GRCh38
rs763902737 Chr.12: 113316652-113316652 on Build GRCh38
rs760366615 Chr.12: 113316585-113316585 on Build GRCh38
rs750855322 Chr.12: 113316590-113316590 on Build GRCh38
rs991584383 Chr.12: 113316552-113316552 on Build GRCh38
rs535181226 Chr.12: 113316496-113316496 on Build GRCh38
rs771648224 Chr.12: 113316640-113316640 on Build GRCh38
rs776922174 Chr.12: 113316509-113316509 on Build GRCh38
rs753524846 Chr.12: 113316658-113316658 on Build GRCh38
rs769606106 Chr.12: 113316547-113316547 on Build GRCh38
rs752138278 Chr.12: 113316671-113316671 on Build GRCh38
rs375836359 Chr.12: 113316597-113316597 on Build GRCh38
rs192939316 Chr.12: 113316692-113316692 on Build GRCh38
rs189446912 Chr.12: 113316639-113316639 on Build GRCh38
rs774625879 Chr.12: 113316914-113316914 on Build GRCh38
rs868420261 Chr.12: 113316777-113316777 on Build GRCh38
rs190342887 Chr.12: 113316871-113316871 on Build GRCh38
rs766788388 Chr.12: 113316909-113316909 on Build GRCh38
rs990040740 Chr.12: 113316787-113316787 on Build GRCh38
rs749150438 Chr.12: 113316617-113316617 on Build GRCh38
rs772941024 Chr.12: 113316551-113316551 on Build GRCh38
rs755389759 Chr.12: 113316672-113316672 on Build GRCh38
rs200050012 Chr.12: 113316924-113316924 on Build GRCh38
rs759143683 Chr.12: 113316574-113316574 on Build GRCh38
rs746411682 Chr.12: 113316478-113316478 on Build GRCh38
rs768990376 Chr.12: 113316901-113316902 on Build GRCh38
rs774152586 Chr.12: 113316561-113316561 on Build GRCh38
rs752402153 Chr.12: 113316578-113316578 on Build GRCh38
rs1028350292 Chr.12: 113316852-113316852 on Build GRCh38
rs150909186 Chr.12: 113316641-113316641 on Build GRCh38
rs759912262 Chr.12: 113316923-113316923 on Build GRCh38
rs768306742 Chr.12: 113316616-113316616 on Build GRCh38
rs932343879 Chr.12: 113316835-113316835 on Build GRCh38
rs771158950 Chr.12: 113316743-113316743 on Build GRCh38
rs562443808 Chr.12: 113316784-113316784 on Build GRCh38
rs912193736 Chr.12: 113316880-113316880 on Build GRCh38
rs1050831554 Chr.12: 113316679-113316679 on Build GRCh38
rs774360124 Chr.12: 113316633-113316633 on Build GRCh38
rs183659475 Chr.12: 113316595-113316595 on Build GRCh38
rs201732490 Chr.12: 113316703-113316703 on Build GRCh38
rs952310996 Chr.12: 113316853-113316853 on Build GRCh38
rs766884597 Chr.12: 113316575-113316575 on Build GRCh38
rs747177229 Chr.12: 113316491-113316491 on Build GRCh38
rs758928434 Chr.12: 113316486-113316486 on Build GRCh38
rs143888280 Chr.12: 113316841-113316841 on Build GRCh38
rs768602112 Chr.12: 113316501-113316501 on Build GRCh38
rs549421723 Chr.12: 113316722-113316722 on Build GRCh38
rs762715752 Chr.12: 113316555-113316555 on Build GRCh38
rs770943015 Chr.12: 113316557-113316557 on Build GRCh38
rs201016489 Chr.12: 113316908-113316908 on Build GRCh38
rs770619577 Chr.12: 113316628-113316628 on Build GRCh38
rs770049033 Chr.12: 113316612-113316612 on Build GRCh38
rs540306423 Chr.12: 113316797-113316797 on Build GRCh38
rs748389337 Chr.12: 113316600-113316600 on Build GRCh38
rs185099421 Chr.12: 113316721-113316721 on Build GRCh38
rs866590476 Chr.12: 113316718-113316718 on Build GRCh38
rs748770332 Chr.12: 113316535-113316535 on Build GRCh38
rs995105326 Chr.12: 113316696-113316696 on Build GRCh38

View Full Product Details