Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 26758459-26758962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR12
Entrez Gene ID
2835
Gene Name
G protein-coupled receptor 12
Gene Aliases
GPCR12, GPCR21, PPP1R84
UniGene
Hs.123034
Gene Chromosome Location
Chr.13: 26755200-26760786 on Build GRCh38
Transcripts
NM_005288.3 XM_005266360.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
432256 c.962C>T p.P321L Chr.13: 26758866-26758866 on Build GRCh38
946384 c.956G>A p.C319Y Chr.13: 26758872-26758872 on Build GRCh38
6074354 c.894T>C p.P298P Chr.13: 26758934-26758934 on Build GRCh38
7369951 c.976C>A p.Q326K Chr.13: 26758852-26758852 on Build GRCh38
2069909 c.990G>A p.S330S Chr.13: 26758838-26758838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536262480 Chr.13: 26758816-26758816 on Build GRCh38
rs542611211 Chr.13: 26758701-26758701 on Build GRCh38
rs1036428945 Chr.13: 26758618-26758618 on Build GRCh38
rs770089394 Chr.13: 26758929-26758929 on Build GRCh38
rs772580044 Chr.13: 26758804-26758804 on Build GRCh38
rs775896560 Chr.13: 26758933-26758933 on Build GRCh38
rs1016382544 Chr.13: 26758722-26758722 on Build GRCh38
rs778560633 Chr.13: 26758851-26758851 on Build GRCh38
rs767460900 Chr.13: 26758836-26758836 on Build GRCh38
rs781631956 Chr.13: 26758895-26758895 on Build GRCh38
rs370232717 Chr.13: 26758808-26758808 on Build GRCh38
rs550450670 Chr.13: 26758832-26758832 on Build GRCh38
rs759172517 Chr.13: 26758684-26758684 on Build GRCh38
rs747367348 Chr.13: 26758807-26758807 on Build GRCh38
rs951959483 Chr.13: 26758641-26758641 on Build GRCh38
rs376937208 Chr.13: 26758800-26758800 on Build GRCh38
rs532712067 Chr.13: 26758617-26758618 on Build GRCh38
rs760738450 Chr.13: 26758932-26758932 on Build GRCh38
rs754587158 Chr.13: 26758844-26758844 on Build GRCh38
rs542551076 Chr.13: 26758648-26758648 on Build GRCh38
rs373528373 Chr.13: 26758786-26758786 on Build GRCh38
rs948078104 Chr.13: 26758483-26758483 on Build GRCh38
rs770423995 Chr.13: 26758612-26758612 on Build GRCh38
rs746264322 Chr.13: 26758917-26758917 on Build GRCh38
rs1043475368 Chr.13: 26758707-26758707 on Build GRCh38
rs551572566 Chr.13: 26758681-26758681 on Build GRCh38
rs763088466 Chr.13: 26758937-26758937 on Build GRCh38
rs752179748 Chr.13: 26758866-26758866 on Build GRCh38
rs766031054 Chr.13: 26758842-26758842 on Build GRCh38
rs1018636414 Chr.13: 26758691-26758691 on Build GRCh38
rs925084377 Chr.13: 26758710-26758710 on Build GRCh38
rs775667623 Chr.13: 26758827-26758827 on Build GRCh38
rs557365729 Chr.13: 26758649-26758649 on Build GRCh38
rs775424334 Chr.13: 26758867-26758867 on Build GRCh38
rs201409616 Chr.13: 26758596-26758596 on Build GRCh38
rs200921336 Chr.13: 26758774-26758774 on Build GRCh38
rs547416630 Chr.13: 26758703-26758703 on Build GRCh38
rs778338916 Chr.13: 26758886-26758886 on Build GRCh38
rs748771175 Chr.13: 26758600-26758600 on Build GRCh38
rs779755956 Chr.13: 26758776-26758776 on Build GRCh38
rs776895704 Chr.13: 26758811-26758811 on Build GRCh38
rs760509127 Chr.13: 26758838-26758838 on Build GRCh38
rs905856786 Chr.13: 26758806-26758806 on Build GRCh38
rs747527142 Chr.13: 26758889-26758889 on Build GRCh38
rs970067833 Chr.13: 26758526-26758526 on Build GRCh38
rs771478789 Chr.13: 26758894-26758894 on Build GRCh38
rs777301206 Chr.13: 26758509-26758509 on Build GRCh38
rs564815325 Chr.13: 26758695-26758695 on Build GRCh38
rs773639587 Chr.13: 26758805-26758805 on Build GRCh38
rs765558647 Chr.13: 26758820-26758820 on Build GRCh38
rs748998493 Chr.13: 26758778-26758778 on Build GRCh38
rs980745667 Chr.13: 26758593-26758593 on Build GRCh38
rs897978652 Chr.13: 26758647-26758649 on Build GRCh38
rs187921171 Chr.13: 26758673-26758673 on Build GRCh38
rs936137231 Chr.13: 26758715-26758715 on Build GRCh38
rs1053307261 Chr.13: 26758739-26758739 on Build GRCh38
rs747689547 Chr.13: 26758802-26758802 on Build GRCh38
rs759770137 Chr.13: 26758818-26758818 on Build GRCh38
rs570350461 Chr.13: 26758860-26758860 on Build GRCh38
rs753635977 Chr.13: 26758843-26758843 on Build GRCh38
rs983317456 Chr.13: 26758657-26758657 on Build GRCh38
rs975712467 Chr.13: 26758686-26758686 on Build GRCh38
rs868045258 Chr.13: 26758537-26758537 on Build GRCh38
rs1927522 Chr.13: 26758699-26758699 on Build GRCh38

View Full Product Details