Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43108976-43109473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC15
Entrez Gene ID
29103
Gene Name
DnaJ heat shock protein family (Hsp40) member C15
Gene Aliases
DNAJD1, HSD18, MCJ
UniGene
Hs.438830
Gene Chromosome Location
Chr.13: 43023226-43109170 on Build GRCh38
Transcripts
NM_013238.2

SNP Detail

SNP ID Chromosome Location
rs34285426 Chr.13: 43109367-43109368 on Build GRCh38
rs531121967 Chr.13: 43109212-43109212 on Build GRCh38
rs767937200 Chr.13: 43109104-43109104 on Build GRCh38
rs988302887 Chr.13: 43109035-43109035 on Build GRCh38
rs762108144 Chr.13: 43109101-43109101 on Build GRCh38
rs145569779 Chr.13: 43109364-43109364 on Build GRCh38
rs886616020 Chr.13: 43109242-43109242 on Build GRCh38
rs1034447585 Chr.13: 43109007-43109007 on Build GRCh38
rs140163760 Chr.13: 43109275-43109275 on Build GRCh38
rs575947807 Chr.13: 43109247-43109247 on Build GRCh38
rs147531115 Chr.13: 43109233-43109233 on Build GRCh38
rs185648380 Chr.13: 43109100-43109100 on Build GRCh38
rs752068694 Chr.13: 43108998-43108998 on Build GRCh38
rs545793000 Chr.13: 43109223-43109223 on Build GRCh38
rs183030338 Chr.13: 43109373-43109373 on Build GRCh38
rs538407938 Chr.13: 43109044-43109044 on Build GRCh38
rs554011587 Chr.13: 43109129-43109129 on Build GRCh38
rs893208934 Chr.13: 43109344-43109344 on Build GRCh38
rs953484628 Chr.13: 43109320-43109320 on Build GRCh38
rs918093585 Chr.13: 43109050-43109050 on Build GRCh38
rs1004137760 Chr.13: 43109226-43109226 on Build GRCh38
rs1000912791 Chr.13: 43109326-43109326 on Build GRCh38
rs997571256 Chr.13: 43109269-43109269 on Build GRCh38
rs956920066 Chr.13: 43109449-43109449 on Build GRCh38
rs1048774418 Chr.13: 43109199-43109199 on Build GRCh38
rs931114763 Chr.13: 43109209-43109209 on Build GRCh38
rs9590742 Chr.13: 43109108-43109108 on Build GRCh38
rs777057341 Chr.13: 43109345-43109346 on Build GRCh38
rs1039401300 Chr.13: 43109113-43109113 on Build GRCh38
rs1046031398 Chr.13: 43109271-43109271 on Build GRCh38
rs1034374243 Chr.13: 43109289-43109289 on Build GRCh38
rs913105488 Chr.13: 43109059-43109059 on Build GRCh38
rs527526350 Chr.13: 43109343-43109343 on Build GRCh38
rs907173136 Chr.13: 43109272-43109272 on Build GRCh38
rs1011696778 Chr.13: 43109356-43109356 on Build GRCh38
rs370546497 Chr.13: 43109083-43109083 on Build GRCh38
rs188947435 Chr.13: 43109431-43109431 on Build GRCh38
rs990371569 Chr.13: 43109052-43109052 on Build GRCh38
rs957090626 Chr.13: 43109015-43109016 on Build GRCh38
rs931024828 Chr.13: 43109149-43109149 on Build GRCh38
rs190750400 Chr.13: 43109250-43109250 on Build GRCh38
rs142477201 Chr.13: 43109082-43109082 on Build GRCh38
rs117520797 Chr.13: 43109206-43109206 on Build GRCh38

View Full Product Details