Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49145638-49146156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC3A
Entrez Gene ID
22862
Gene Name
fibronectin type III domain containing 3A
Gene Aliases
FNDC3, HUGO, bA203I16.1, bA203I16.5
UniGene
Hs.508010
Gene Chromosome Location
Chr.13: 48975241-49209779 on Build GRCh38
Transcripts
NM_001079673.1 NM_001278438.1 NM_014923.4 NR_103528.1 XM_011534997.2 XM_017020440.1 XM_017020441.1 XM_017020442.1 XM_017020443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5743618 c.826G>A p.D276N Chr.13: 49145784-49145784 on Build GRCh38
4685240 c.891_892delAG p.D298fs*1 Chr.13: 49145849-49145850 on Build GRCh38
5342360 c.947G>A p.G316E Chr.13: 49145905-49145905 on Build GRCh38
5733810 c.839G>C p.R280T Chr.13: 49145797-49145797 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs866545229 Chr.13: 49145782-49145782 on Build GRCh38
rs887209671 Chr.13: 49146084-49146084 on Build GRCh38
rs370952260 Chr.13: 49145870-49145870 on Build GRCh38
rs1029678596 Chr.13: 49146001-49146001 on Build GRCh38
rs771849342 Chr.13: 49145766-49145767 on Build GRCh38
rs1019065352 Chr.13: 49146041-49146041 on Build GRCh38
rs763286271 Chr.13: 49145923-49145924 on Build GRCh38
rs758699776 Chr.13: 49145939-49145939 on Build GRCh38
rs767857297 Chr.13: 49145866-49145866 on Build GRCh38
rs79927363 Chr.13: 49146030-49146030 on Build GRCh38
rs769649405 Chr.13: 49145980-49145980 on Build GRCh38
rs1056972586 Chr.13: 49145845-49145845 on Build GRCh38
rs1047058615 Chr.13: 49146081-49146081 on Build GRCh38
rs937194145 Chr.13: 49145830-49145830 on Build GRCh38
rs747741674 Chr.13: 49145688-49145689 on Build GRCh38
rs946722123 Chr.13: 49146090-49146090 on Build GRCh38
rs745333503 Chr.13: 49145964-49145964 on Build GRCh38
rs753925868 Chr.13: 49145922-49145922 on Build GRCh38
rs984310344 Chr.13: 49145752-49145752 on Build GRCh38
rs928205315 Chr.13: 49145805-49145805 on Build GRCh38
rs376979601 Chr.13: 49145719-49145719 on Build GRCh38
rs998206293 Chr.13: 49146075-49146075 on Build GRCh38
rs370781473 Chr.13: 49145961-49145961 on Build GRCh38
rs987578268 Chr.13: 49145701-49145701 on Build GRCh38
rs905752214 Chr.13: 49145792-49145792 on Build GRCh38
rs755491898 Chr.13: 49145928-49145928 on Build GRCh38
rs778094808 Chr.13: 49145944-49145944 on Build GRCh38
rs765196205 Chr.13: 49145839-49145839 on Build GRCh38
rs912002286 Chr.13: 49145711-49145711 on Build GRCh38
rs866850738 Chr.13: 49145762-49145762 on Build GRCh38
rs770028803 Chr.13: 49145740-49145742 on Build GRCh38
rs748525776 Chr.13: 49145740-49145740 on Build GRCh38
rs201004322 Chr.13: 49145784-49145784 on Build GRCh38
rs779189418 Chr.13: 49145736-49145736 on Build GRCh38
rs946424076 Chr.13: 49145912-49145912 on Build GRCh38
rs565086058 Chr.13: 49145984-49145984 on Build GRCh38
rs772782633 Chr.13: 49145774-49145774 on Build GRCh38
rs530068609 Chr.13: 49145724-49145724 on Build GRCh38
rs771737432 Chr.13: 49145754-49145754 on Build GRCh38
rs1043475881 Chr.13: 49145930-49145930 on Build GRCh38
rs956826134 Chr.13: 49146066-49146066 on Build GRCh38
rs901732763 Chr.13: 49146063-49146063 on Build GRCh38
rs74965684 Chr.13: 49145783-49145783 on Build GRCh38
rs201547692 Chr.13: 49145779-49145779 on Build GRCh38
rs9591241 Chr.13: 49146057-49146057 on Build GRCh38
rs775169432 Chr.13: 49145842-49145842 on Build GRCh38
rs1004631389 Chr.13: 49145903-49145903 on Build GRCh38
rs779430516 Chr.13: 49145931-49145931 on Build GRCh38
rs145148334 Chr.13: 49145886-49145886 on Build GRCh38
rs762797019 Chr.13: 49145848-49145848 on Build GRCh38
rs561376641 Chr.13: 49145668-49145669 on Build GRCh38
rs960373295 Chr.13: 49145818-49145818 on Build GRCh38
rs987629650 Chr.13: 49146031-49146031 on Build GRCh38
rs112927362 Chr.13: 49145843-49145843 on Build GRCh38
rs769204993 Chr.13: 49145733-49145733 on Build GRCh38
rs759431009 Chr.13: 49145813-49145813 on Build GRCh38
rs913653607 Chr.13: 49145906-49145906 on Build GRCh38
rs367881475 Chr.13: 49145953-49145953 on Build GRCh38
rs148351632 Chr.13: 49145934-49145934 on Build GRCh38

View Full Product Details