Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 75284949-75285447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D4
Entrez Gene ID
9882
Gene Name
TBC1 domain family member 4
Gene Aliases
AS160, NIDDM5
UniGene
Hs.210891
Gene Chromosome Location
Chr.13: 75284664-75482646 on Build GRCh38
Transcripts
NM_001286658.2 NM_001286659.2 NM_014832.4 XM_005266603.1 XM_005266605.2 XM_006719903.3 XM_011535331.1 XM_017020882.1 XM_017020883.1 XM_017020884.1

SNP Detail

SNP ID Chromosome Location
rs150881540 Chr.13: 75285010-75285010 on Build GRCh38
rs1050957735 Chr.13: 75285383-75285383 on Build GRCh38
rs15167 Chr.13: 75284973-75284973 on Build GRCh38
rs553181027 Chr.13: 75285267-75285267 on Build GRCh38
rs910353763 Chr.13: 75285386-75285386 on Build GRCh38
rs888966774 Chr.13: 75285122-75285122 on Build GRCh38
rs906664822 Chr.13: 75285001-75285001 on Build GRCh38
rs1010513748 Chr.13: 75285034-75285034 on Build GRCh38
rs944540896 Chr.13: 75285396-75285396 on Build GRCh38
rs1058830 Chr.13: 75285009-75285009 on Build GRCh38
rs973498189 Chr.13: 75285403-75285403 on Build GRCh38
rs930724032 Chr.13: 75285365-75285365 on Build GRCh38
rs920671771 Chr.13: 75285028-75285028 on Build GRCh38
rs920619483 Chr.13: 75285327-75285327 on Build GRCh38
rs916757469 Chr.13: 75284976-75284976 on Build GRCh38
rs529077533 Chr.13: 75285078-75285078 on Build GRCh38
rs892237229 Chr.13: 75285016-75285016 on Build GRCh38
rs74333807 Chr.13: 75285150-75285150 on Build GRCh38
rs1036400803 Chr.13: 75285024-75285024 on Build GRCh38
rs1013098572 Chr.13: 75285242-75285242 on Build GRCh38
rs1014998811 Chr.13: 75285285-75285285 on Build GRCh38
rs902818053 Chr.13: 75285207-75285207 on Build GRCh38
rs867718120 Chr.13: 75285268-75285268 on Build GRCh38
rs903094744 Chr.13: 75285414-75285414 on Build GRCh38
rs552001242 Chr.13: 75285233-75285233 on Build GRCh38
rs41306652 Chr.13: 75285279-75285279 on Build GRCh38
rs535650329 Chr.13: 75285325-75285325 on Build GRCh38
rs1050254325 Chr.13: 75285093-75285093 on Build GRCh38
rs563813986 Chr.13: 75285420-75285420 on Build GRCh38
rs546338395 Chr.13: 75285029-75285029 on Build GRCh38
rs570464054 Chr.13: 75285228-75285228 on Build GRCh38
rs568798682 Chr.13: 75285142-75285142 on Build GRCh38
rs545829869 Chr.13: 75284992-75284992 on Build GRCh38
rs746103379 Chr.13: 75285167-75285167 on Build GRCh38
rs564043763 Chr.13: 75285002-75285002 on Build GRCh38
rs60811184 Chr.13: 75285276-75285276 on Build GRCh38
rs495945 Chr.13: 75285162-75285162 on Build GRCh38
rs982344696 Chr.13: 75285278-75285278 on Build GRCh38
rs192901349 Chr.13: 75285084-75285084 on Build GRCh38
rs575339926 Chr.13: 75284984-75284984 on Build GRCh38
rs561960973 Chr.13: 75285037-75285037 on Build GRCh38
rs962324007 Chr.13: 75285319-75285319 on Build GRCh38
rs535126888 Chr.13: 75285229-75285229 on Build GRCh38
rs928196905 Chr.13: 75285284-75285284 on Build GRCh38

View Full Product Details