Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 99200542-99201041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBAC2
Entrez Gene ID
337867
Gene Name
UBA domain containing 2
Gene Aliases
PHGDHL1
UniGene
Hs.508545
Gene Chromosome Location
Chr.13: 99200425-99386499 on Build GRCh38
Transcripts
NM_001144072.1 NM_177967.3 NR_026644.1 XM_006719948.3 XM_011521082.2 XM_011521083.2 NR_036531.1 NR_036532.1

Gene Symbol
UBAC2-AS1
Entrez Gene ID
100289373
Gene Name
UBAC2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.225914
Gene Chromosome Location
Chr.13: 99196374-99200757 on Build GRCh38
Transcripts
NM_001144072.1 NM_177967.3 NR_026644.1 XM_006719948.3 XM_011521082.2 XM_011521083.2 NR_036531.1 NR_036532.1

SNP Detail

SNP ID Chromosome Location
rs527978131 Chr.13: 99200782-99200782 on Build GRCh38
rs763181920 Chr.13: 99200798-99200798 on Build GRCh38
rs973652872 Chr.13: 99201022-99201022 on Build GRCh38
rs774234534 Chr.13: 99200775-99200775 on Build GRCh38
rs543238763 Chr.13: 99200637-99200637 on Build GRCh38
rs758693215 Chr.13: 99200642-99200642 on Build GRCh38
rs761305716 Chr.13: 99200957-99200957 on Build GRCh38
rs760650640 Chr.13: 99200871-99200871 on Build GRCh38
rs774631614 Chr.13: 99200859-99200859 on Build GRCh38
rs986682666 Chr.13: 99200680-99200680 on Build GRCh38
rs1003294870 Chr.13: 99200918-99200918 on Build GRCh38
rs746237524 Chr.13: 99200890-99200890 on Build GRCh38
rs780337848 Chr.13: 99200655-99200655 on Build GRCh38
rs985112458 Chr.13: 99200735-99200735 on Build GRCh38
rs756499102 Chr.13: 99200845-99200845 on Build GRCh38
rs959776315 Chr.13: 99201003-99201003 on Build GRCh38
rs1044492487 Chr.13: 99200739-99200739 on Build GRCh38
rs771052019 Chr.13: 99200924-99200924 on Build GRCh38
rs965038852 Chr.13: 99200959-99200959 on Build GRCh38
rs755341903 Chr.13: 99200896-99200896 on Build GRCh38
rs545322501 Chr.13: 99200712-99200712 on Build GRCh38
rs769582705 Chr.13: 99200948-99200948 on Build GRCh38
rs1040531001 Chr.13: 99200657-99200657 on Build GRCh38
rs1012897567 Chr.13: 99200783-99200783 on Build GRCh38
rs948487903 Chr.13: 99200728-99200728 on Build GRCh38
rs534305294 Chr.13: 99200776-99200790 on Build GRCh38
rs754612679 Chr.13: 99200803-99200803 on Build GRCh38
rs572717844 Chr.13: 99200612-99200612 on Build GRCh38
rs751124248 Chr.13: 99200956-99200956 on Build GRCh38
rs768783605 Chr.13: 99200772-99200772 on Build GRCh38
rs759297846 Chr.13: 99200947-99200947 on Build GRCh38
rs976319229 Chr.13: 99200861-99200861 on Build GRCh38
rs768815068 Chr.13: 99200856-99200856 on Build GRCh38
rs1006636247 Chr.13: 99200949-99200949 on Build GRCh38
rs114014993 Chr.13: 99200659-99200659 on Build GRCh38
rs776512241 Chr.13: 99200875-99200875 on Build GRCh38
rs772611996 Chr.13: 99200896-99200897 on Build GRCh38
rs375852830 Chr.13: 99200770-99200770 on Build GRCh38
rs948412694 Chr.13: 99200571-99200571 on Build GRCh38
rs773314899 Chr.13: 99200789-99200789 on Build GRCh38
rs1022858677 Chr.13: 99200694-99200694 on Build GRCh38
rs117807058 Chr.13: 99200865-99200865 on Build GRCh38
rs955473937 Chr.13: 99200679-99200679 on Build GRCh38
rs1021403743 Chr.13: 99200939-99200939 on Build GRCh38
rs989819837 Chr.13: 99200723-99200723 on Build GRCh38
rs755324448 Chr.13: 99200965-99200965 on Build GRCh38
rs757869105 Chr.13: 99200888-99200888 on Build GRCh38
rs1041058330 Chr.13: 99200761-99200761 on Build GRCh38
rs370106352 Chr.13: 99200792-99200792 on Build GRCh38
rs544676429 Chr.13: 99200993-99200993 on Build GRCh38
rs775708614 Chr.13: 99200768-99200768 on Build GRCh38
rs554550718 Chr.13: 99200832-99200832 on Build GRCh38
rs996614882 Chr.13: 99200677-99200677 on Build GRCh38
rs932323958 Chr.13: 99200606-99200606 on Build GRCh38
rs932292949 Chr.13: 99200732-99200732 on Build GRCh38
rs868768990 Chr.13: 99200935-99200935 on Build GRCh38
rs752213731 Chr.13: 99200820-99200820 on Build GRCh38
rs766914013 Chr.13: 99200960-99200960 on Build GRCh38
rs750929509 Chr.13: 99200889-99200889 on Build GRCh38
rs891755318 Chr.13: 99200969-99200969 on Build GRCh38
rs953779973 Chr.13: 99200815-99200815 on Build GRCh38
rs914332525 Chr.13: 99200725-99200725 on Build GRCh38
rs182889692 Chr.13: 99200752-99200752 on Build GRCh38
rs1047886593 Chr.13: 99200629-99200629 on Build GRCh38
rs914236202 Chr.13: 99200603-99200603 on Build GRCh38
rs748357810 Chr.13: 99200902-99200902 on Build GRCh38
rs749722415 Chr.13: 99200894-99200894 on Build GRCh38
rs898019253 Chr.13: 99200658-99200658 on Build GRCh38
rs758044713 Chr.13: 99200785-99200785 on Build GRCh38
rs758636662 Chr.13: 99200972-99200972 on Build GRCh38
rs775383089 Chr.13: 99200970-99200970 on Build GRCh38
rs944848206 Chr.13: 99200651-99200651 on Build GRCh38
rs749523896 Chr.13: 99200854-99200854 on Build GRCh38
rs748236156 Chr.13: 99200780-99200780 on Build GRCh38
rs373364683 Chr.13: 99200863-99200863 on Build GRCh38
rs7332516 Chr.13: 99200971-99200971 on Build GRCh38
rs560006493 Chr.13: 99200721-99200721 on Build GRCh38
rs767424226 Chr.13: 99200786-99200786 on Build GRCh38
rs187489387 Chr.13: 99200903-99200903 on Build GRCh38
rs779057665 Chr.13: 99200898-99200898 on Build GRCh38
rs548027687 Chr.13: 99200787-99200787 on Build GRCh38
rs778128954 Chr.13: 99200979-99200979 on Build GRCh38
rs527531328 Chr.13: 99200747-99200747 on Build GRCh38
rs747155396 Chr.13: 99200982-99200982 on Build GRCh38
rs1012928722 Chr.13: 99200688-99200688 on Build GRCh38
rs571045666 Chr.13: 99200801-99200801 on Build GRCh38
rs764817547 Chr.13: 99200808-99200808 on Build GRCh38
rs71437996 Chr.13: 99200610-99200610 on Build GRCh38
rs573804464 Chr.13: 99200890-99200890 on Build GRCh38
rs891733535 Chr.13: 99200936-99200936 on Build GRCh38
rs989893231 Chr.13: 99200597-99200597 on Build GRCh38
rs746957543 Chr.13: 99200908-99200908 on Build GRCh38
rs771286084 Chr.13: 99200722-99200722 on Build GRCh38
rs1047945726 Chr.13: 99200766-99200766 on Build GRCh38
rs770122317 Chr.13: 99200765-99200765 on Build GRCh38
rs985070207 Chr.13: 99200991-99200991 on Build GRCh38
rs879722849 Chr.13: 99200776-99200776 on Build GRCh38
rs780325778 Chr.13: 99200849-99200849 on Build GRCh38
rs752371279 Chr.13: 99200881-99200881 on Build GRCh38
rs776677891 Chr.13: 99200929-99200929 on Build GRCh38
rs929430402 Chr.13: 99200866-99200866 on Build GRCh38
rs969886950 Chr.13: 99200983-99200983 on Build GRCh38
rs754350232 Chr.13: 99200961-99200961 on Build GRCh38
rs762595604 Chr.13: 99200953-99200953 on Build GRCh38
rs1044452443 Chr.13: 99200998-99200998 on Build GRCh38
rs773308226 Chr.13: 99200869-99200869 on Build GRCh38
rs900635550 Chr.13: 99200897-99200897 on Build GRCh38
rs7331894 Chr.13: 99200698-99200698 on Build GRCh38
rs1011378908 Chr.13: 99200794-99200794 on Build GRCh38
rs544326107 Chr.13: 99200892-99200892 on Build GRCh38
rs983120395 Chr.13: 99201004-99201004 on Build GRCh38
rs1011473809 Chr.13: 99200938-99200938 on Build GRCh38
rs376502594 Chr.13: 99200882-99200882 on Build GRCh38
rs546492801 Chr.13: 99200825-99200825 on Build GRCh38
rs115307678 Chr.13: 99200706-99200706 on Build GRCh38
rs566328001 Chr.13: 99200841-99200841 on Build GRCh38
rs763603437 Chr.13: 99200954-99200954 on Build GRCh38
rs1003122361 Chr.13: 99200607-99200607 on Build GRCh38
rs989962655 Chr.13: 99200709-99200709 on Build GRCh38
rs772018668 Chr.13: 99200823-99200823 on Build GRCh38
rs948770399 Chr.13: 99200900-99200900 on Build GRCh38
rs775036733 Chr.13: 99200951-99200951 on Build GRCh38
rs766282657 Chr.13: 99200873-99200873 on Build GRCh38
rs767889622 Chr.13: 99200668-99200668 on Build GRCh38
rs549985156 Chr.13: 99200788-99200788 on Build GRCh38
rs747534851 Chr.13: 99200676-99200676 on Build GRCh38
rs113860034 Chr.13: 99201016-99201016 on Build GRCh38
rs1044052006 Chr.13: 99200587-99200587 on Build GRCh38
rs781598886 Chr.13: 99200836-99200836 on Build GRCh38
rs555163358 Chr.13: 99200880-99200880 on Build GRCh38
rs192537681 Chr.13: 99200653-99200653 on Build GRCh38
rs942295778 Chr.13: 99201020-99201020 on Build GRCh38
rs12428890 Chr.13: 99200757-99200757 on Build GRCh38
rs909372897 Chr.13: 99200992-99200992 on Build GRCh38

View Full Product Details