Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 108208082-108208592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIG4
Entrez Gene ID
3981
Gene Name
DNA ligase 4
Gene Aliases
LIG4S
UniGene
Hs.166091
Gene Chromosome Location
Chr.13: 108207442-108218368 on Build GRCh38
Transcripts
NM_001098268.1 NM_002312.3 NM_206937.1 XM_005254056.1 XM_005254057.4 XM_005254058.3 XM_006719951.3 XM_006719952.1 XM_011521091.2 XM_011521092.2 XM_017020563.1 XM_017020564.1 XM_017020565.1 XM_017020566.1 XM_017020567.1 XM_017020568.1 XM_017020569.1 XM_017020570.1 XM_017020571.1 XM_017020572.1 XM_017020573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5896207 c.2725T>C p.Y909H Chr.13: 108208544-108208544 on Build GRCh38
945237 c.2733T>G p.I911M Chr.13: 108208536-108208536 on Build GRCh38
3793099 c.2716G>A p.E906K Chr.13: 108208553-108208553 on Build GRCh38
6138602 c.2678G>T p.W893L Chr.13: 108208591-108208591 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757019157 Chr.13: 108208256-108208256 on Build GRCh38
rs971052887 Chr.13: 108208227-108208227 on Build GRCh38
rs567266202 Chr.13: 108208234-108208234 on Build GRCh38
rs2232643 Chr.13: 108208479-108208479 on Build GRCh38
rs3093769 Chr.13: 108208329-108208329 on Build GRCh38
rs192044163 Chr.13: 108208223-108208223 on Build GRCh38
rs774731069 Chr.13: 108208556-108208556 on Build GRCh38
rs974443784 Chr.13: 108208359-108208359 on Build GRCh38
rs766679114 Chr.13: 108208533-108208533 on Build GRCh38
rs763778059 Chr.13: 108208212-108208212 on Build GRCh38
rs578115401 Chr.13: 108208406-108208406 on Build GRCh38
rs1001607543 Chr.13: 108208154-108208154 on Build GRCh38
rs748776641 Chr.13: 108208248-108208248 on Build GRCh38
rs200695712 Chr.13: 108208505-108208505 on Build GRCh38
rs911866772 Chr.13: 108208317-108208317 on Build GRCh38
rs3093767 Chr.13: 108208404-108208404 on Build GRCh38
rs911705145 Chr.13: 108208308-108208308 on Build GRCh38
rs386774524 Chr.13: 108208402-108208404 on Build GRCh38
rs886049945 Chr.13: 108208227-108208227 on Build GRCh38
rs553852969 Chr.13: 108208440-108208440 on Build GRCh38
rs536126735 Chr.13: 108208303-108208303 on Build GRCh38
rs1010011811 Chr.13: 108208166-108208166 on Build GRCh38
rs551263548 Chr.13: 108208530-108208530 on Build GRCh38
rs778616787 Chr.13: 108208266-108208266 on Build GRCh38
rs3093768 Chr.13: 108208402-108208402 on Build GRCh38
rs538355612 Chr.13: 108208367-108208367 on Build GRCh38
rs886049944 Chr.13: 108208181-108208181 on Build GRCh38
rs1038928844 Chr.13: 108208335-108208335 on Build GRCh38
rs1034008680 Chr.13: 108208489-108208489 on Build GRCh38
rs1024893369 Chr.13: 108208124-108208124 on Build GRCh38
rs959885828 Chr.13: 108208498-108208498 on Build GRCh38
rs907319991 Chr.13: 108208139-108208139 on Build GRCh38
rs886049946 Chr.13: 108208291-108208291 on Build GRCh38
rs940934198 Chr.13: 108208427-108208427 on Build GRCh38
rs199988074 Chr.13: 108208529-108208529 on Build GRCh38
rs369320737 Chr.13: 108208499-108208499 on Build GRCh38
rs1033897436 Chr.13: 108208242-108208242 on Build GRCh38
rs906839898 Chr.13: 108208477-108208477 on Build GRCh38
rs930322513 Chr.13: 108208377-108208377 on Build GRCh38
rs773391515 Chr.13: 108208507-108208507 on Build GRCh38
rs748160595 Chr.13: 108208503-108208503 on Build GRCh38
rs886049947 Chr.13: 108208477-108208477 on Build GRCh38
rs751588140 Chr.13: 108208363-108208363 on Build GRCh38
rs983583039 Chr.13: 108208315-108208315 on Build GRCh38
rs767764977 Chr.13: 108208458-108208458 on Build GRCh38
rs573957541 Chr.13: 108208443-108208443 on Build GRCh38
rs886049948 Chr.13: 108208507-108208507 on Build GRCh38
rs966989318 Chr.13: 108208298-108208298 on Build GRCh38
rs143680726 Chr.13: 108208306-108208306 on Build GRCh38
rs1028222636 Chr.13: 108208522-108208522 on Build GRCh38
rs2232644 Chr.13: 108208429-108208429 on Build GRCh38
rs1007379195 Chr.13: 108208326-108208326 on Build GRCh38

View Full Product Details