Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 108230609-108231111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD13
Entrez Gene ID
84945
Gene Name
abhydrolase domain containing 13
Gene Aliases
BEM46L1, C13orf6, bA153I24.2
UniGene
Hs.183528
Gene Chromosome Location
Chr.13: 108218415-108235440 on Build GRCh38
Transcripts
NM_032859.2 XM_011521128.2

SNP Detail

SNP ID Chromosome Location
rs771472353 Chr.13: 108230657-108230657 on Build GRCh38
rs769768184 Chr.13: 108230641-108230642 on Build GRCh38
rs776956063 Chr.13: 108230751-108230751 on Build GRCh38
rs1022637583 Chr.13: 108231064-108231064 on Build GRCh38
rs773409046 Chr.13: 108230789-108230790 on Build GRCh38
rs942930026 Chr.13: 108231056-108231056 on Build GRCh38
rs1032033665 Chr.13: 108231054-108231054 on Build GRCh38
rs907063241 Chr.13: 108230900-108230901 on Build GRCh38
rs543387171 Chr.13: 108230772-108230772 on Build GRCh38
rs943316548 Chr.13: 108230917-108230917 on Build GRCh38
rs957123744 Chr.13: 108230936-108230936 on Build GRCh38
rs989865944 Chr.13: 108230992-108230992 on Build GRCh38
rs546899670 Chr.13: 108230705-108230705 on Build GRCh38
rs1036909645 Chr.13: 108230755-108230755 on Build GRCh38
rs77167645 Chr.13: 108231063-108231063 on Build GRCh38
rs887068032 Chr.13: 108230853-108230853 on Build GRCh38
rs577584668 Chr.13: 108230709-108230709 on Build GRCh38
rs1039785074 Chr.13: 108230715-108230715 on Build GRCh38
rs931366993 Chr.13: 108230828-108230828 on Build GRCh38
rs774057432 Chr.13: 108230938-108230938 on Build GRCh38
rs1019952473 Chr.13: 108230997-108230997 on Build GRCh38
rs773164746 Chr.13: 108231049-108231049 on Build GRCh38
rs889239101 Chr.13: 108230912-108230912 on Build GRCh38
rs987096787 Chr.13: 108231033-108231033 on Build GRCh38
rs919421407 Chr.13: 108230793-108230793 on Build GRCh38
rs983761597 Chr.13: 108230684-108230684 on Build GRCh38
rs570300506 Chr.13: 108230851-108230851 on Build GRCh38
rs998794797 Chr.13: 108231050-108231050 on Build GRCh38
rs967448594 Chr.13: 108231007-108231007 on Build GRCh38
rs912771921 Chr.13: 108230689-108230689 on Build GRCh38
rs866556627 Chr.13: 108230896-108230896 on Build GRCh38
rs1024635248 Chr.13: 108230898-108230898 on Build GRCh38
rs142572031 Chr.13: 108231070-108231070 on Build GRCh38
rs879114453 Chr.13: 108231052-108231053 on Build GRCh38
rs1047057914 Chr.13: 108230843-108230843 on Build GRCh38
rs148022496 Chr.13: 108231057-108231057 on Build GRCh38
rs190636809 Chr.13: 108230837-108230837 on Build GRCh38
rs530075512 Chr.13: 108230633-108230633 on Build GRCh38
rs566325467 Chr.13: 108230999-108230999 on Build GRCh38
rs763480751 Chr.13: 108230919-108230919 on Build GRCh38
rs904413039 Chr.13: 108230922-108230922 on Build GRCh38
rs969689444 Chr.13: 108230642-108230642 on Build GRCh38
rs933508068 Chr.13: 108230862-108230862 on Build GRCh38
rs762575109 Chr.13: 108230910-108230910 on Build GRCh38
rs563478948 Chr.13: 108230923-108230923 on Build GRCh38

View Full Product Details