Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110910493-110911011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD10
Entrez Gene ID
55608
Gene Name
ankyrin repeat domain 10
Gene Aliases
-
UniGene
Hs.525163
Gene Chromosome Location
Chr.13: 110878540-110915107 on Build GRCh38
Transcripts
NM_001286721.1 NM_017664.3 NR_104586.1 NR_104587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7240079 c.214G>C p.E72Q Chr.13: 110910767-110910767 on Build GRCh38
5052057 c.240G>A p.A80A Chr.13: 110910741-110910741 on Build GRCh38
4045998 c.239C>T p.A80V Chr.13: 110910742-110910742 on Build GRCh38
7154364 c.333G>A p.L111L Chr.13: 110910648-110910648 on Build GRCh38
7240080 c.214G>C p.E72Q Chr.13: 110910767-110910767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374687918 Chr.13: 110910516-110910516 on Build GRCh38
rs1051088477 Chr.13: 110910961-110910961 on Build GRCh38
rs765147230 Chr.13: 110910726-110910726 on Build GRCh38
rs1015546482 Chr.13: 110910669-110910669 on Build GRCh38
rs148355794 Chr.13: 110910841-110910841 on Build GRCh38
rs761519788 Chr.13: 110910725-110910725 on Build GRCh38
rs577635206 Chr.13: 110910970-110910970 on Build GRCh38
rs767031712 Chr.13: 110910698-110910699 on Build GRCh38
rs1018976317 Chr.13: 110910747-110910747 on Build GRCh38
rs780009979 Chr.13: 110910689-110910689 on Build GRCh38
rs113322614 Chr.13: 110910969-110910969 on Build GRCh38
rs373572644 Chr.13: 110910578-110910578 on Build GRCh38
rs953479837 Chr.13: 110910707-110910707 on Build GRCh38
rs753327697 Chr.13: 110910781-110910782 on Build GRCh38
rs200594052 Chr.13: 110910723-110910723 on Build GRCh38
rs746331293 Chr.13: 110910814-110910814 on Build GRCh38
rs776095441 Chr.13: 110910569-110910581 on Build GRCh38
rs1015425196 Chr.13: 110910864-110910864 on Build GRCh38
rs773755248 Chr.13: 110910583-110910583 on Build GRCh38
rs151032452 Chr.13: 110910708-110910708 on Build GRCh38
rs573286734 Chr.13: 110910901-110910901 on Build GRCh38
rs567653926 Chr.13: 110910775-110910775 on Build GRCh38
rs768032030 Chr.13: 110910717-110910717 on Build GRCh38
rs146756849 Chr.13: 110910678-110910678 on Build GRCh38
rs762150209 Chr.13: 110910976-110910976 on Build GRCh38
rs141005738 Chr.13: 110910716-110910716 on Build GRCh38
rs751437692 Chr.13: 110910754-110910754 on Build GRCh38
rs771188210 Chr.13: 110910800-110910800 on Build GRCh38
rs540758326 Chr.13: 110910910-110910910 on Build GRCh38
rs778067900 Chr.13: 110910706-110910706 on Build GRCh38
rs140379452 Chr.13: 110910699-110910699 on Build GRCh38
rs761303042 Chr.13: 110910577-110910578 on Build GRCh38
rs1055721901 Chr.13: 110910568-110910568 on Build GRCh38
rs977255837 Chr.13: 110910533-110910533 on Build GRCh38
rs9515310 Chr.13: 110910523-110910523 on Build GRCh38
rs761571172 Chr.13: 110910818-110910818 on Build GRCh38
rs919694717 Chr.13: 110910953-110910953 on Build GRCh38
rs34628205 Chr.13: 110910705-110910705 on Build GRCh38
rs548394085 Chr.13: 110910618-110910618 on Build GRCh38
rs753530921 Chr.13: 110910644-110910644 on Build GRCh38
rs192056146 Chr.13: 110910577-110910577 on Build GRCh38
rs752944118 Chr.13: 110910769-110910769 on Build GRCh38
rs766964358 Chr.13: 110910605-110910605 on Build GRCh38
rs376198510 Chr.13: 110910730-110910730 on Build GRCh38
rs201123888 Chr.13: 110910657-110910657 on Build GRCh38
rs766388108 Chr.13: 110910937-110910937 on Build GRCh38
rs781449379 Chr.13: 110910700-110910700 on Build GRCh38
rs369883132 Chr.13: 110910757-110910757 on Build GRCh38
rs747216678 Chr.13: 110910567-110910567 on Build GRCh38
rs768039455 Chr.13: 110910808-110910808 on Build GRCh38
rs748577354 Chr.13: 110910579-110910579 on Build GRCh38
rs528628084 Chr.13: 110910786-110910786 on Build GRCh38
rs748718492 Chr.13: 110910976-110910981 on Build GRCh38
rs201363046 Chr.13: 110910684-110910684 on Build GRCh38
rs772222727 Chr.13: 110910518-110910518 on Build GRCh38
rs769504011 Chr.13: 110910820-110910820 on Build GRCh38
rs1008837781 Chr.13: 110910983-110910983 on Build GRCh38
rs747044030 Chr.13: 110910697-110910697 on Build GRCh38
rs9559904 Chr.13: 110910842-110910842 on Build GRCh38
rs763875761 Chr.13: 110910635-110910635 on Build GRCh38
rs1005035263 Chr.13: 110910659-110910659 on Build GRCh38
rs766404124 Chr.13: 110910742-110910742 on Build GRCh38
rs768315066 Chr.13: 110910815-110910815 on Build GRCh38
rs939785152 Chr.13: 110910873-110910873 on Build GRCh38
rs977224852 Chr.13: 110910787-110910787 on Build GRCh38
rs543023607 Chr.13: 110910536-110910536 on Build GRCh38
rs554897390 Chr.13: 110910553-110910553 on Build GRCh38
rs554984417 Chr.13: 110910885-110910885 on Build GRCh38
rs760254438 Chr.13: 110910620-110910620 on Build GRCh38
rs144260529 Chr.13: 110910759-110910759 on Build GRCh38
rs75790696 Chr.13: 110910850-110910850 on Build GRCh38
rs763415492 Chr.13: 110910594-110910594 on Build GRCh38
rs775299558 Chr.13: 110910557-110910557 on Build GRCh38
rs1038133138 Chr.13: 110910875-110910875 on Build GRCh38
rs771405371 Chr.13: 110910712-110910712 on Build GRCh38
rs935957322 Chr.13: 110910552-110910552 on Build GRCh38
rs546753251 Chr.13: 110910817-110910817 on Build GRCh38
rs995034248 Chr.13: 110910890-110910890 on Build GRCh38
rs73621198 Chr.13: 110910869-110910869 on Build GRCh38
rs186013252 Chr.13: 110910871-110910871 on Build GRCh38
rs143331283 Chr.13: 110910619-110910619 on Build GRCh38
rs868086308 Chr.13: 110910651-110910651 on Build GRCh38
rs774720088 Chr.13: 110910714-110910714 on Build GRCh38
rs777043399 Chr.13: 110910876-110910876 on Build GRCh38
rs775756178 Chr.13: 110910804-110910804 on Build GRCh38
rs754030090 Chr.13: 110910813-110910814 on Build GRCh38
rs757169482 Chr.13: 110910656-110910656 on Build GRCh38
rs770128455 Chr.13: 110910581-110910581 on Build GRCh38
rs945857020 Chr.13: 110910532-110910532 on Build GRCh38
rs750186687 Chr.13: 110910729-110910729 on Build GRCh38
rs578140050 Chr.13: 110910782-110910782 on Build GRCh38

View Full Product Details