Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23276490-23276996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOMEZ
Entrez Gene ID
57594
Gene Name
homeobox and leucine zipper encoding
Gene Aliases
KIAA1443
UniGene
Hs.632332
Gene Chromosome Location
Chr.14: 23273635-23286100 on Build GRCh38
Transcripts
NM_020834.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4049995 c.619A>G p.T207A Chr.14: 23276609-23276609 on Build GRCh38
6021257 c.459G>C p.Q153H Chr.14: 23276697-23276697 on Build GRCh38
4950443 c.668T>G p.L223R Chr.14: 23276560-23276560 on Build GRCh38
4049999 c.373C>T p.R125* Chr.14: 23276855-23276855 on Build GRCh38
3885887 c.160C>T p.P54S Chr.14: 23276996-23276996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756332834 Chr.14: 23276949-23276949 on Build GRCh38
rs1047460773 Chr.14: 23276909-23276909 on Build GRCh38
rs775173637 Chr.14: 23276840-23276840 on Build GRCh38
rs765570451 Chr.14: 23276880-23276880 on Build GRCh38
rs765619841 Chr.14: 23276773-23276773 on Build GRCh38
rs752462244 Chr.14: 23276608-23276608 on Build GRCh38
rs754275626 Chr.14: 23276639-23276639 on Build GRCh38
rs141391198 Chr.14: 23276952-23276952 on Build GRCh38
rs995531431 Chr.14: 23276521-23276521 on Build GRCh38
rs1010894258 Chr.14: 23276734-23276734 on Build GRCh38
rs750934524 Chr.14: 23276789-23276789 on Build GRCh38
rs913673921 Chr.14: 23276670-23276670 on Build GRCh38
rs555718213 Chr.14: 23276747-23276747 on Build GRCh38
rs151224767 Chr.14: 23276646-23276646 on Build GRCh38
rs772969776 Chr.14: 23276565-23276565 on Build GRCh38
rs200890395 Chr.14: 23276765-23276765 on Build GRCh38
rs751449532 Chr.14: 23276586-23276586 on Build GRCh38
rs763640428 Chr.14: 23276854-23276854 on Build GRCh38
rs200575967 Chr.14: 23276842-23276842 on Build GRCh38
rs746328110 Chr.14: 23276650-23276650 on Build GRCh38
rs747231137 Chr.14: 23276653-23276653 on Build GRCh38
rs772155577 Chr.14: 23276843-23276843 on Build GRCh38
rs370288175 Chr.14: 23276569-23276569 on Build GRCh38
rs754986781 Chr.14: 23276808-23276808 on Build GRCh38
rs775115082 Chr.14: 23276743-23276743 on Build GRCh38
rs775700857 Chr.14: 23276540-23276540 on Build GRCh38
rs1029872144 Chr.14: 23276798-23276798 on Build GRCh38
rs745807747 Chr.14: 23276797-23276797 on Build GRCh38
rs11844797 Chr.14: 23276514-23276514 on Build GRCh38
rs745703035 Chr.14: 23276956-23276956 on Build GRCh38
rs1037886712 Chr.14: 23276600-23276600 on Build GRCh38
rs758984751 Chr.14: 23276592-23276592 on Build GRCh38
rs867223331 Chr.14: 23276771-23276771 on Build GRCh38
rs755113250 Chr.14: 23276902-23276902 on Build GRCh38
rs757601886 Chr.14: 23276641-23276641 on Build GRCh38
rs771924835 Chr.14: 23276957-23276957 on Build GRCh38
rs201848325 Chr.14: 23276580-23276580 on Build GRCh38
rs767037610 Chr.14: 23276606-23276606 on Build GRCh38
rs752898380 Chr.14: 23276916-23276916 on Build GRCh38
rs1026986951 Chr.14: 23276788-23276788 on Build GRCh38
rs772122394 Chr.14: 23276522-23276522 on Build GRCh38
rs894468935 Chr.14: 23276754-23276754 on Build GRCh38
rs374149068 Chr.14: 23276786-23276786 on Build GRCh38
rs766879279 Chr.14: 23276898-23276898 on Build GRCh38
rs747282762 Chr.14: 23276694-23276694 on Build GRCh38
rs776204768 Chr.14: 23276855-23276855 on Build GRCh38
rs778327194 Chr.14: 23276630-23276630 on Build GRCh38
rs1056074764 Chr.14: 23276714-23276714 on Build GRCh38
rs920907477 Chr.14: 23276624-23276624 on Build GRCh38
rs775526551 Chr.14: 23276847-23276847 on Build GRCh38
rs201309405 Chr.14: 23276560-23276560 on Build GRCh38
rs946159070 Chr.14: 23276675-23276675 on Build GRCh38
rs779910230 Chr.14: 23276884-23276884 on Build GRCh38
rs761496469 Chr.14: 23276861-23276861 on Build GRCh38
rs375469673 Chr.14: 23276689-23276689 on Build GRCh38
rs774216330 Chr.14: 23276712-23276712 on Build GRCh38
rs368363259 Chr.14: 23276792-23276792 on Build GRCh38
rs201024374 Chr.14: 23276701-23276701 on Build GRCh38
rs767171943 Chr.14: 23276730-23276730 on Build GRCh38
rs776798725 Chr.14: 23276729-23276729 on Build GRCh38
rs748392753 Chr.14: 23276839-23276839 on Build GRCh38
rs868334703 Chr.14: 23276769-23276769 on Build GRCh38
rs758724170 Chr.14: 23276897-23276897 on Build GRCh38
rs760884609 Chr.14: 23276554-23276554 on Build GRCh38
rs780491384 Chr.14: 23276793-23276793 on Build GRCh38
rs375408085 Chr.14: 23276913-23276913 on Build GRCh38
rs548943423 Chr.14: 23276695-23276695 on Build GRCh38
rs759509939 Chr.14: 23276713-23276713 on Build GRCh38
rs527590138 Chr.14: 23276613-23276613 on Build GRCh38
rs752007684 Chr.14: 23276901-23276901 on Build GRCh38
rs750823744 Chr.14: 23276892-23276892 on Build GRCh38
rs111256939 Chr.14: 23276794-23276794 on Build GRCh38
rs755792976 Chr.14: 23276609-23276609 on Build GRCh38
rs746507331 Chr.14: 23276959-23276960 on Build GRCh38
rs867868645 Chr.14: 23276654-23276654 on Build GRCh38
rs1003447662 Chr.14: 23276681-23276681 on Build GRCh38
rs758670338 Chr.14: 23276515-23276515 on Build GRCh38
rs17128364 Chr.14: 23276826-23276826 on Build GRCh38
rs866326509 Chr.14: 23276762-23276762 on Build GRCh38
rs866952670 Chr.14: 23276582-23276582 on Build GRCh38
rs898888064 Chr.14: 23276618-23276618 on Build GRCh38
rs753471767 Chr.14: 23276766-23276766 on Build GRCh38
rs372297156 Chr.14: 23276768-23276768 on Build GRCh38
rs577666081 Chr.14: 23276852-23276852 on Build GRCh38
rs778097861 Chr.14: 23276574-23276574 on Build GRCh38
rs929035363 Chr.14: 23276867-23276867 on Build GRCh38

View Full Product Details